The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001322123.1 | NP_001309052.1 | 4 - 5 | 680 | 65 |
NM_001322124.1 | NP_001309053.1 | 4 - 5 | 549 | 65 | |
NM_001322127.1 | NP_001309056.1 | 4 - 5 | 737 | 65 | |
NM_144709.3 | NP_653310.2 | 4 - 5 | 737 | 65 | |
XM_011532568.2 | XP_011530870.1 | 5 - 6 | 808 | 65 | |
XM_011532570.1 | XP_011530872.1 | 4 - 5 | 515 | 65 | |
XM_011532571.1 | XP_011530873.1 | 4 - 5 | 689 | 65 | |
XM_011532572.1 | XP_011530874.1 | 4 - 5 | 609 | 65 | |
XM_011532573.2 | XP_011530875.1 | 4 - 5 | 569 | 65 | |
XM_011532574.2 | XP_011530876.1 | 4 - 5 | 630 | 65 | |
XM_011532576.2 | XP_011530878.1 | 4 - 5 | 689 | 65 | |
XM_011532578.1 | XP_011530880.1 | 4 - 5 | 667 | 65 | |
XM_017003428.1 | XP_016858917.1 | 3 - 4 | 506 | 65 | |
XM_017003429.1 | XP_016858918.1 | 3 - 4 | 302 | 65 | |
GenBank mRNA | AK056874.1 | - | 4 - 5 | 551 | 65 |
AK291729.1 | - | 4 - 5 | 525 | 65 | |
AL832208.1 | - | 4 - 5 | 738 | 65 | |
BC101680.1 | - | 4 - 5 | 529 | 65 | |
BC101706.1 | - | 4 - 5 | 529 | 65 | |
BC143480.1 | - | 4 - 5 | 529 | 65 |