Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 45855900-45856396 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRY2
Entrez Gene ID
1408
Gene Name
cryptochrome circadian clock 2
Gene Aliases
HCRY2, PHLL2
UniGene
Hs.532491
Gene Chromosome Location
Chr.11: 45847118-45883248 on Build GRCh38
Transcripts
NM_001127457.2 NM_021117.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
189415 c.286C>T p.R96W Chr.11: 45856052-45856052 on Build GRCh38
1979677 c.304G>A p.V102M Chr.11: 45856070-45856070 on Build GRCh38
4168590 c.349C>T p.R117W Chr.11: 45856052-45856052 on Build GRCh38
3998406 c.279-4A>G p.? Chr.11: 45855978-45855978 on Build GRCh38
3998409 c.266A>C p.N89T Chr.11: 45856032-45856032 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144480697 Chr.11: 45856053-45856053 on Build GRCh38
rs753048345 Chr.11: 45856138-45856138 on Build GRCh38
rs750684818 Chr.11: 45855944-45855944 on Build GRCh38
rs375524683 Chr.11: 45856098-45856098 on Build GRCh38
rs888546903 Chr.11: 45856177-45856177 on Build GRCh38
rs768698727 Chr.11: 45856112-45856112 on Build GRCh38
rs2292912 Chr.11: 45856137-45856137 on Build GRCh38
rs778411772 Chr.11: 45856096-45856096 on Build GRCh38
rs1005516482 Chr.11: 45856181-45856181 on Build GRCh38
rs755311944 Chr.11: 45856067-45856067 on Build GRCh38
rs947227419 Chr.11: 45856257-45856257 on Build GRCh38
rs775311434 Chr.11: 45856286-45856286 on Build GRCh38
rs1007622322 Chr.11: 45856340-45856340 on Build GRCh38
rs1030329445 Chr.11: 45856320-45856320 on Build GRCh38
rs200067613 Chr.11: 45856087-45856087 on Build GRCh38
rs772188459 Chr.11: 45856043-45856043 on Build GRCh38
rs747704173 Chr.11: 45855964-45855964 on Build GRCh38
rs911975826 Chr.11: 45856187-45856187 on Build GRCh38
rs756977453 Chr.11: 45856095-45856095 on Build GRCh38
rs760646362 Chr.11: 45856305-45856305 on Build GRCh38
rs948300992 Chr.11: 45856342-45856342 on Build GRCh38
rs935654682 Chr.11: 45856063-45856063 on Build GRCh38
rs991134530 Chr.11: 45856290-45856290 on Build GRCh38
rs1047166129 Chr.11: 45856171-45856171 on Build GRCh38
rs571428136 Chr.11: 45856238-45856238 on Build GRCh38
rs777080865 Chr.11: 45855977-45855977 on Build GRCh38
rs771233272 Chr.11: 45856133-45856133 on Build GRCh38
rs963901755 Chr.11: 45856152-45856152 on Build GRCh38
rs929612431 Chr.11: 45856173-45856173 on Build GRCh38
rs747505736 Chr.11: 45855948-45855948 on Build GRCh38
rs188542192 Chr.11: 45856256-45856256 on Build GRCh38
rs374052307 Chr.11: 45855934-45855934 on Build GRCh38
rs760652914 Chr.11: 45856052-45856052 on Build GRCh38
rs967225901 Chr.11: 45856230-45856230 on Build GRCh38
rs766776238 Chr.11: 45855950-45855950 on Build GRCh38
rs763290948 Chr.11: 45856060-45856060 on Build GRCh38
rs551290356 Chr.11: 45856186-45856186 on Build GRCh38
rs977268711 Chr.11: 45856243-45856243 on Build GRCh38
rs755544149 Chr.11: 45855952-45855952 on Build GRCh38
rs185632883 Chr.11: 45856097-45856097 on Build GRCh38
rs2292913 Chr.11: 45855978-45855978 on Build GRCh38
rs200945082 Chr.11: 45856070-45856070 on Build GRCh38
rs75521198 Chr.11: 45856161-45856161 on Build GRCh38
rs937100868 Chr.11: 45856212-45856212 on Build GRCh38
rs534865692 Chr.11: 45856170-45856170 on Build GRCh38
rs762909166 Chr.11: 45856131-45856131 on Build GRCh38
rs768608838 Chr.11: 45856333-45856333 on Build GRCh38
rs147827036 Chr.11: 45856162-45856162 on Build GRCh38
rs918231009 Chr.11: 45855933-45855933 on Build GRCh38
rs746953201 Chr.11: 45856108-45856108 on Build GRCh38
rs1016417826 Chr.11: 45856139-45856139 on Build GRCh38
rs767821278 Chr.11: 45856069-45856069 on Build GRCh38
rs557269726 Chr.11: 45856274-45856274 on Build GRCh38
rs781506593 Chr.11: 45856066-45856066 on Build GRCh38
rs774457567 Chr.11: 45856134-45856134 on Build GRCh38
rs1011479291 Chr.11: 45856034-45856034 on Build GRCh38
rs745620833 Chr.11: 45856037-45856037 on Build GRCh38
rs751853411 Chr.11: 45855951-45855951 on Build GRCh38
rs997472869 Chr.11: 45856302-45856302 on Build GRCh38
rs774270514 Chr.11: 45856016-45856016 on Build GRCh38
rs1029487695 Chr.11: 45856228-45856228 on Build GRCh38
rs773405279 Chr.11: 45856127-45856127 on Build GRCh38
rs1016777937 Chr.11: 45856353-45856353 on Build GRCh38
rs781640514 Chr.11: 45855963-45855963 on Build GRCh38
rs777401218 Chr.11: 45856080-45856080 on Build GRCh38
rs903103814 Chr.11: 45856288-45856288 on Build GRCh38
rs998135217 Chr.11: 45856220-45856220 on Build GRCh38
rs963547290 Chr.11: 45856369-45856369 on Build GRCh38
rs867577947 Chr.11: 45856336-45856336 on Build GRCh38
rs925588441 Chr.11: 45856024-45856024 on Build GRCh38
rs775593968 Chr.11: 45856046-45856046 on Build GRCh38
rs770478503 Chr.11: 45855988-45855988 on Build GRCh38
rs1055981862 Chr.11: 45856255-45856255 on Build GRCh38

View Full Product Details