Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SAMD13
Entrez Gene ID
148418
Gene Name
sterile alpha motif domain containing 13
Gene Aliases
HSD-41, HSD-42
Chromosome Location
Chr.1: 84298366 - 84350798 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001010971.2 NP_001010971.1 1 - 2 203 149
GenBank mRNA AK056937.1 - 1 - 2 203 149

View Full Product Details