Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 46656155-46656665 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG13
Entrez Gene ID
9776
Gene Name
autophagy related 13
Gene Aliases
KIAA0652, PARATARG8
UniGene
Hs.127403
Gene Chromosome Location
Chr.11: 46617276-46676019 on Build GRCh38
Transcripts
NM_001142673.2 NM_001205119.1 NM_001205120.1 NM_001205121.1 NM_001205122.1 NM_014741.4 XM_005253262.2 XM_005253263.2 XM_005253265.2 XM_005253266.2 XM_005253268.2 XM_006718394.2 XM_006718395.2 XM_006718396.2 XM_011520493.1 XM_011520496.2 XM_011520499.2 XM_011520500.1 XM_011520501.1 XM_011520502.1 XM_011520503.1 XM_017018599.1 XM_017018600.1 XM_017018601.1 XM_017018602.1 XM_017018603.1 XM_017018604.1 XM_017018605.1 XM_017018606.1 XM_017018607.1 XM_017018608.1 XM_017018609.1 XM_017018610.1 XM_017018611.1 XM_017018612.1 XM_017018613.1 XM_017018614.1 XM_017018615.1 XM_017018616.1 XM_017018617.1 XM_017018618.1 XM_017018619.1 XM_017018620.1 XM_017018621.1 XM_017018622.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7092988 c.475G>T p.V159F Chr.11: 46656249-46656249 on Build GRCh38
7092989 c.475G>T p.V159F Chr.11: 46656249-46656249 on Build GRCh38
6443405 c.493G>A p.G165R Chr.11: 46656267-46656267 on Build GRCh38
6068740 c.459-2A>G p.? Chr.11: 46656231-46656231 on Build GRCh38
6068741 c.459-2A>G p.? Chr.11: 46656231-46656231 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs891228127 Chr.11: 46656526-46656526 on Build GRCh38
rs913147437 Chr.11: 46656338-46656338 on Build GRCh38
rs1006617242 Chr.11: 46656514-46656514 on Build GRCh38
rs556019714 Chr.11: 46656559-46656559 on Build GRCh38
rs771113319 Chr.11: 46656332-46656333 on Build GRCh38
rs762296163 Chr.11: 46656296-46656298 on Build GRCh38
rs774933850 Chr.11: 46656215-46656216 on Build GRCh38
rs779900568 Chr.11: 46656504-46656504 on Build GRCh38
rs932071104 Chr.11: 46656381-46656381 on Build GRCh38
rs183665918 Chr.11: 46656303-46656303 on Build GRCh38
rs371927221 Chr.11: 46656269-46656269 on Build GRCh38
rs999232260 Chr.11: 46656532-46656532 on Build GRCh38
rs572589069 Chr.11: 46656624-46656624 on Build GRCh38
rs562458680 Chr.11: 46656203-46656203 on Build GRCh38
rs570971602 Chr.11: 46656299-46656299 on Build GRCh38
rs923060725 Chr.11: 46656377-46656377 on Build GRCh38
rs887813606 Chr.11: 46656502-46656502 on Build GRCh38
rs775958246 Chr.11: 46656206-46656206 on Build GRCh38
rs761193302 Chr.11: 46656208-46656208 on Build GRCh38
rs996698088 Chr.11: 46656479-46656479 on Build GRCh38
rs1030118711 Chr.11: 46656503-46656503 on Build GRCh38
rs187970248 Chr.11: 46656436-46656436 on Build GRCh38
rs970597601 Chr.11: 46656630-46656630 on Build GRCh38
rs202000661 Chr.11: 46656285-46656285 on Build GRCh38
rs549907718 Chr.11: 46656345-46656345 on Build GRCh38
rs975668332 Chr.11: 46656533-46656533 on Build GRCh38
rs879476684 Chr.11: 46656618-46656618 on Build GRCh38
rs569991768 Chr.11: 46656352-46656352 on Build GRCh38
rs945895447 Chr.11: 46656339-46656339 on Build GRCh38
rs990105608 Chr.11: 46656330-46656330 on Build GRCh38
rs561781278 Chr.11: 46656260-46656260 on Build GRCh38
rs772622681 Chr.11: 46656317-46656321 on Build GRCh38
rs4474394 Chr.11: 46656184-46656184 on Build GRCh38
rs1031666057 Chr.11: 46656537-46656537 on Build GRCh38
rs1013759357 Chr.11: 46656619-46656619 on Build GRCh38
rs778715519 Chr.11: 46656392-46656396 on Build GRCh38
rs771194852 Chr.11: 46656194-46656194 on Build GRCh38
rs551584742 Chr.11: 46656357-46656357 on Build GRCh38
rs1036656971 Chr.11: 46656578-46656578 on Build GRCh38
rs774604861 Chr.11: 46656300-46656300 on Build GRCh38
rs767627519 Chr.11: 46656205-46656205 on Build GRCh38
rs779732863 Chr.11: 46656277-46656277 on Build GRCh38
rs957383260 Chr.11: 46656558-46656560 on Build GRCh38
rs1023258978 Chr.11: 46656610-46656610 on Build GRCh38
rs990400575 Chr.11: 46656601-46656601 on Build GRCh38
rs764936045 Chr.11: 46656229-46656229 on Build GRCh38
rs141970718 Chr.11: 46656218-46656218 on Build GRCh38
rs547945733 Chr.11: 46656286-46656286 on Build GRCh38
rs746898312 Chr.11: 46656282-46656282 on Build GRCh38
rs775578066 Chr.11: 46656317-46656317 on Build GRCh38
rs772094954 Chr.11: 46656315-46656315 on Build GRCh38
rs368591554 Chr.11: 46656222-46656222 on Build GRCh38
rs981636751 Chr.11: 46656641-46656641 on Build GRCh38

View Full Product Details