Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 46679274-46679771 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP1
Entrez Gene ID
392
Gene Name
Rho GTPase activating protein 1
Gene Aliases
CDC42GAP, RHOGAP, RHOGAP1, p50rhoGAP
UniGene
Hs.138860
Gene Chromosome Location
Chr.11: 46677075-46700665 on Build GRCh38
Transcripts
NM_004308.3 XM_011520095.1 XM_017017720.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4033050 c.968G>A p.R323Q Chr.11: 46679707-46679707 on Build GRCh38
4033051 c.947T>A p.V316D Chr.11: 46679728-46679728 on Build GRCh38
6602995 c.969G>A p.R323R Chr.11: 46679706-46679706 on Build GRCh38
4662704 c.1078C>T p.L360L Chr.11: 46679418-46679418 on Build GRCh38
1353979 c.1083C>T p.P361P Chr.11: 46679413-46679413 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757043724 Chr.11: 46679445-46679445 on Build GRCh38
rs553048352 Chr.11: 46679573-46679573 on Build GRCh38
rs968301107 Chr.11: 46679516-46679516 on Build GRCh38
rs764099533 Chr.11: 46679379-46679379 on Build GRCh38
rs748217545 Chr.11: 46679509-46679509 on Build GRCh38
rs970350540 Chr.11: 46679373-46679373 on Build GRCh38
rs758274604 Chr.11: 46679685-46679685 on Build GRCh38
rs147210555 Chr.11: 46679390-46679390 on Build GRCh38
rs928565474 Chr.11: 46679383-46679383 on Build GRCh38
rs878966561 Chr.11: 46679540-46679540 on Build GRCh38
rs72912170 Chr.11: 46679558-46679558 on Build GRCh38
rs768735129 Chr.11: 46679467-46679467 on Build GRCh38
rs773769674 Chr.11: 46679413-46679413 on Build GRCh38
rs537308490 Chr.11: 46679323-46679324 on Build GRCh38
rs947849528 Chr.11: 46679387-46679387 on Build GRCh38
rs1047586241 Chr.11: 46679389-46679389 on Build GRCh38
rs1028755738 Chr.11: 46679527-46679527 on Build GRCh38
rs767250150 Chr.11: 46679430-46679430 on Build GRCh38
rs374411055 Chr.11: 46679370-46679370 on Build GRCh38
rs397759792 Chr.11: 46679487-46679487 on Build GRCh38
rs769879177 Chr.11: 46679510-46679510 on Build GRCh38
rs752632876 Chr.11: 46679329-46679329 on Build GRCh38
rs199616017 Chr.11: 46679721-46679721 on Build GRCh38
rs371270837 Chr.11: 46679443-46679443 on Build GRCh38
rs150718407 Chr.11: 46679465-46679465 on Build GRCh38
rs549354963 Chr.11: 46679523-46679523 on Build GRCh38
rs78561418 Chr.11: 46679622-46679622 on Build GRCh38
rs965818019 Chr.11: 46679299-46679299 on Build GRCh38
rs765265297 Chr.11: 46679676-46679676 on Build GRCh38
rs900005377 Chr.11: 46679483-46679483 on Build GRCh38
rs951755056 Chr.11: 46679309-46679309 on Build GRCh38
rs367590178 Chr.11: 46679403-46679403 on Build GRCh38
rs771280427 Chr.11: 46679610-46679610 on Build GRCh38
rs781548772 Chr.11: 46679487-46679487 on Build GRCh38
rs778600249 Chr.11: 46679460-46679460 on Build GRCh38
rs749090193 Chr.11: 46679368-46679368 on Build GRCh38
rs375437342 Chr.11: 46679633-46679633 on Build GRCh38
rs372580382 Chr.11: 46679624-46679624 on Build GRCh38
rs766660778 Chr.11: 46679416-46679416 on Build GRCh38
rs757162345 Chr.11: 46679664-46679664 on Build GRCh38
rs190139019 Chr.11: 46679640-46679640 on Build GRCh38
rs142672718 Chr.11: 46679441-46679441 on Build GRCh38
rs1023029450 Chr.11: 46679559-46679559 on Build GRCh38
rs773645063 Chr.11: 46679597-46679597 on Build GRCh38
rs775340418 Chr.11: 46679634-46679634 on Build GRCh38
rs747198102 Chr.11: 46679380-46679380 on Build GRCh38
rs201440837 Chr.11: 46679712-46679712 on Build GRCh38
rs775213363 Chr.11: 46679739-46679739 on Build GRCh38
rs748376853 Chr.11: 46679688-46679688 on Build GRCh38
rs774558378 Chr.11: 46679618-46679618 on Build GRCh38
rs544447260 Chr.11: 46679708-46679708 on Build GRCh38
rs969731654 Chr.11: 46679563-46679563 on Build GRCh38
rs749827642 Chr.11: 46679598-46679598 on Build GRCh38
rs974819576 Chr.11: 46679724-46679724 on Build GRCh38
rs761925290 Chr.11: 46679407-46679407 on Build GRCh38
rs909055867 Chr.11: 46679660-46679660 on Build GRCh38
rs752225067 Chr.11: 46679440-46679440 on Build GRCh38
rs750150951 Chr.11: 46679678-46679678 on Build GRCh38
rs760433907 Chr.11: 46679740-46679740 on Build GRCh38
rs907805944 Chr.11: 46679334-46679334 on Build GRCh38
rs892750625 Chr.11: 46679434-46679434 on Build GRCh38
rs756099087 Chr.11: 46679330-46679330 on Build GRCh38
rs753784390 Chr.11: 46679659-46679659 on Build GRCh38
rs961919483 Chr.11: 46679377-46679377 on Build GRCh38
rs35160532 Chr.11: 46679485-46679485 on Build GRCh38
rs777557410 Chr.11: 46679600-46679600 on Build GRCh38
rs1041273028 Chr.11: 46679471-46679471 on Build GRCh38
rs113094912 Chr.11: 46679694-46679694 on Build GRCh38
rs936701614 Chr.11: 46679411-46679411 on Build GRCh38
rs187238148 Chr.11: 46679574-46679574 on Build GRCh38
rs537182010 Chr.11: 46679305-46679305 on Build GRCh38
rs372150816 Chr.11: 46679328-46679328 on Build GRCh38
rs772498411 Chr.11: 46679733-46679733 on Build GRCh38
rs375008964 Chr.11: 46679475-46679475 on Build GRCh38
rs756149679 Chr.11: 46679691-46679691 on Build GRCh38
rs770285908 Chr.11: 46679412-46679412 on Build GRCh38
rs763646448 Chr.11: 46679744-46679744 on Build GRCh38
rs749420115 Chr.11: 46679707-46679707 on Build GRCh38
rs777896991 Chr.11: 46679348-46679348 on Build GRCh38
rs200568285 Chr.11: 46679715-46679715 on Build GRCh38
rs995925023 Chr.11: 46679300-46679300 on Build GRCh38
rs11822837 Chr.11: 46679391-46679391 on Build GRCh38
rs992036453 Chr.11: 46679596-46679596 on Build GRCh38
rs984837279 Chr.11: 46679311-46679311 on Build GRCh38
rs375751904 Chr.11: 46679639-46679639 on Build GRCh38
rs759134220 Chr.11: 46679420-46679420 on Build GRCh38
rs777839644 Chr.11: 46679442-46679442 on Build GRCh38
rs980988405 Chr.11: 46679635-46679635 on Build GRCh38
rs370106026 Chr.11: 46679706-46679706 on Build GRCh38
rs1032550984 Chr.11: 46679488-46679488 on Build GRCh38
rs969205219 Chr.11: 46679628-46679628 on Build GRCh38
rs751185981 Chr.11: 46679419-46679419 on Build GRCh38
rs539338698 Chr.11: 46679593-46679593 on Build GRCh38

View Full Product Details