Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 47466638-47467135 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CELF1
Entrez Gene ID
10658
Gene Name
CUGBP, Elav-like family member 1
Gene Aliases
BRUNOL2, CUG-BP, CUGBP, CUGBP1, EDEN-BP, NAB50, NAPOR, hNab50
UniGene
Hs.595333
Gene Chromosome Location
Chr.11: 47465937-47565569 on Build GRCh38
Transcripts
NM_001025596.2 NM_001172639.1 NM_001172640.1 NM_006560.3 NM_198700.2 XM_011519848.2 XM_011519850.2 XM_011519852.2 XM_011519854.1 XM_011519855.1 XM_011519856.1 XM_011519857.1 XM_011519858.1 XM_011519859.1 XM_017017101.1 XM_017017102.1 XM_017017103.1 XM_017017104.1 XM_017017105.1 XM_017017106.1 XM_017017107.1 XM_017017108.1 XM_017017109.1 XM_017017110.1 XM_017017111.1 XM_017017112.1 XM_017017113.1 XM_017017114.1 XM_017017115.1 XM_017017116.1 XM_017017117.1 XM_017017118.1 XM_017017119.1 XM_017017120.1 XM_017017121.1 XM_017017122.1 XM_017017123.1 XM_017017124.1 XM_017017125.1 XM_017017126.1 XM_017017127.1 XM_017017128.1 XM_017017129.1 XM_017017130.1 XM_017017131.1 XM_017017132.1 XM_017017133.1 XM_017017134.1 XM_017017135.1

SNP Detail

SNP ID Chromosome Location
rs36060230 Chr.11: 47466941-47466942 on Build GRCh38
rs751793248 Chr.11: 47466774-47466774 on Build GRCh38
rs973625988 Chr.11: 47466802-47466802 on Build GRCh38
rs766575306 Chr.11: 47466728-47466728 on Build GRCh38
rs980576236 Chr.11: 47466941-47466941 on Build GRCh38
rs1008564511 Chr.11: 47467096-47467096 on Build GRCh38
rs145694200 Chr.11: 47467038-47467038 on Build GRCh38
rs537472388 Chr.11: 47466702-47466702 on Build GRCh38
rs1021470255 Chr.11: 47466726-47466726 on Build GRCh38
rs537465879 Chr.11: 47466938-47466939 on Build GRCh38
rs964936408 Chr.11: 47466795-47466795 on Build GRCh38
rs568447899 Chr.11: 47466675-47466678 on Build GRCh38
rs914737414 Chr.11: 47466868-47466868 on Build GRCh38
rs929475514 Chr.11: 47466832-47466832 on Build GRCh38
rs572172045 Chr.11: 47466900-47466900 on Build GRCh38
rs1020254474 Chr.11: 47466831-47466831 on Build GRCh38
rs776093235 Chr.11: 47466863-47466863 on Build GRCh38
rs12054 Chr.11: 47466697-47466697 on Build GRCh38
rs571834471 Chr.11: 47466674-47466674 on Build GRCh38
rs927419627 Chr.11: 47466979-47466979 on Build GRCh38
rs956280113 Chr.11: 47466834-47466836 on Build GRCh38
rs540851882 Chr.11: 47466984-47466984 on Build GRCh38
rs557390114 Chr.11: 47466749-47466749 on Build GRCh38
rs1003968073 Chr.11: 47466734-47466734 on Build GRCh38
rs998718027 Chr.11: 47467017-47467017 on Build GRCh38
rs954470729 Chr.11: 47467058-47467058 on Build GRCh38
rs895107920 Chr.11: 47466932-47466932 on Build GRCh38
rs954589992 Chr.11: 47466773-47466773 on Build GRCh38
rs928087055 Chr.11: 47466847-47466847 on Build GRCh38
rs75336726 Chr.11: 47466783-47466783 on Build GRCh38
rs1048299295 Chr.11: 47466837-47466837 on Build GRCh38
rs11546837 Chr.11: 47466834-47466834 on Build GRCh38
rs572821940 Chr.11: 47466983-47466983 on Build GRCh38
rs977603371 Chr.11: 47466746-47466746 on Build GRCh38
rs920792635 Chr.11: 47466811-47466811 on Build GRCh38
rs1012804834 Chr.11: 47466707-47466707 on Build GRCh38
rs917793388 Chr.11: 47467079-47467079 on Build GRCh38
rs536704719 Chr.11: 47466810-47466810 on Build GRCh38
rs1802814 Chr.11: 47466740-47466740 on Build GRCh38
rs1007909730 Chr.11: 47466808-47466808 on Build GRCh38
rs1003525625 Chr.11: 47466687-47466687 on Build GRCh38
rs939387144 Chr.11: 47466892-47466892 on Build GRCh38
rs1028099748 Chr.11: 47466793-47466793 on Build GRCh38
rs555622363 Chr.11: 47466895-47466895 on Build GRCh38
rs759866488 Chr.11: 47466970-47466970 on Build GRCh38

View Full Product Details