Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 47467445-47467943 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CELF1
Entrez Gene ID
10658
Gene Name
CUGBP, Elav-like family member 1
Gene Aliases
BRUNOL2, CUG-BP, CUGBP, CUGBP1, EDEN-BP, NAB50, NAPOR, hNab50
UniGene
Hs.595333
Gene Chromosome Location
Chr.11: 47465937-47565569 on Build GRCh38
Transcripts
NM_001025596.2 NM_001172639.1 NM_001172640.1 NM_006560.3 NM_198700.2 XM_011519848.2 XM_011519850.2 XM_011519852.2 XM_011519854.1 XM_011519855.1 XM_011519856.1 XM_011519857.1 XM_011519858.1 XM_011519859.1 XM_017017101.1 XM_017017102.1 XM_017017103.1 XM_017017104.1 XM_017017105.1 XM_017017106.1 XM_017017107.1 XM_017017108.1 XM_017017109.1 XM_017017110.1 XM_017017111.1 XM_017017112.1 XM_017017113.1 XM_017017114.1 XM_017017115.1 XM_017017116.1 XM_017017117.1 XM_017017118.1 XM_017017119.1 XM_017017120.1 XM_017017121.1 XM_017017122.1 XM_017017123.1 XM_017017124.1 XM_017017125.1 XM_017017126.1 XM_017017127.1 XM_017017128.1 XM_017017129.1 XM_017017130.1 XM_017017131.1 XM_017017132.1 XM_017017133.1 XM_017017134.1 XM_017017135.1

SNP Detail

SNP ID Chromosome Location
rs979464119 Chr.11: 47467607-47467607 on Build GRCh38
rs566829188 Chr.11: 47467732-47467732 on Build GRCh38
rs886103000 Chr.11: 47467826-47467826 on Build GRCh38
rs1035257311 Chr.11: 47467902-47467902 on Build GRCh38
rs890313797 Chr.11: 47467703-47467703 on Build GRCh38
rs900276342 Chr.11: 47467805-47467805 on Build GRCh38
rs1005168995 Chr.11: 47467864-47467864 on Build GRCh38
rs1014392035 Chr.11: 47467905-47467905 on Build GRCh38
rs944526722 Chr.11: 47467738-47467738 on Build GRCh38
rs960857959 Chr.11: 47467574-47467574 on Build GRCh38
rs911045574 Chr.11: 47467754-47467754 on Build GRCh38
rs895964594 Chr.11: 47467893-47467893 on Build GRCh38
rs1001784764 Chr.11: 47467831-47467831 on Build GRCh38
rs1035235332 Chr.11: 47467573-47467573 on Build GRCh38
rs148955629 Chr.11: 47467700-47467700 on Build GRCh38
rs960924584 Chr.11: 47467908-47467908 on Build GRCh38
rs756638228 Chr.11: 47467481-47467481 on Build GRCh38
rs985816454 Chr.11: 47467753-47467753 on Build GRCh38
rs778493500 Chr.11: 47467806-47467806 on Build GRCh38
rs973385772 Chr.11: 47467639-47467639 on Build GRCh38
rs1034519835 Chr.11: 47467568-47467568 on Build GRCh38
rs951005679 Chr.11: 47467482-47467482 on Build GRCh38
rs557145148 Chr.11: 47467878-47467881 on Build GRCh38
rs567618681 Chr.11: 47467662-47467662 on Build GRCh38
rs536758852 Chr.11: 47467671-47467671 on Build GRCh38
rs756775198 Chr.11: 47467743-47467743 on Build GRCh38
rs943894166 Chr.11: 47467774-47467774 on Build GRCh38
rs1040792997 Chr.11: 47467783-47467783 on Build GRCh38
rs990927190 Chr.11: 47467577-47467577 on Build GRCh38
rs557655989 Chr.11: 47467812-47467812 on Build GRCh38
rs943796874 Chr.11: 47467467-47467467 on Build GRCh38
rs759253752 Chr.11: 47467564-47467564 on Build GRCh38
rs147051157 Chr.11: 47467742-47467742 on Build GRCh38
rs1009924704 Chr.11: 47467514-47467514 on Build GRCh38
rs891450287 Chr.11: 47467505-47467505 on Build GRCh38
rs181043887 Chr.11: 47467593-47467593 on Build GRCh38
rs946754305 Chr.11: 47467600-47467600 on Build GRCh38
rs531222846 Chr.11: 47467522-47467522 on Build GRCh38
rs1053434545 Chr.11: 47467701-47467701 on Build GRCh38
rs904780506 Chr.11: 47467551-47467551 on Build GRCh38
rs891564551 Chr.11: 47467793-47467793 on Build GRCh38

View Full Product Details