The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300940.1 | NP_001287869.1 | 2 - 3 | 581 | 84 |
NM_001300941.1 | NP_001287870.1 | 2 - 3 | 422 | 84 | |
NM_152440.4 | NP_689653.3 | 2 - 3 | 581 | 84 | |
GenBank mRNA | AK057111.1 | - | 2 - 3 | 548 | 84 |
AK129933.1 | - | 2 - 3 | 422 | 84 | |
BC036246.1 | - | 2 - 3 | 550 | 84 | |
BC071634.1 | - | 2 - 3 | 538 | 84 |