Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KAZN
Entrez Gene ID
23254
Gene Name
kazrin, periplakin interacting protein
Gene Aliases
KAZ
Chromosome Location
Chr.1: 13893387 - 15118048 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201628.2 NP_963922.2 15 - 15 4289 77
XM_005245795.4 XP_005245852.1 17 - 17 4544 77
XM_011541074.2 XP_011539376.1 16 - 16 6480 77
XM_011541075.2 XP_011539377.1 14 - 14 6136 77
XM_011541076.2 XP_011539378.1 14 - 14 6281 77
XM_011541077.2 XP_011539379.1 14 - 14 7260 77
XM_011541081.2 XP_011539383.1 8 - 8 4960 77
GenBank mRNA AK057155.1 - 1 - 1 849 77
AK125388.1 - 1 - 1 1464 77

Gene Symbol
TMEM51-AS1
Entrez Gene ID
200197
Gene Name
TMEM51 antisense RNA 1
Gene Aliases
C1orf126
Chromosome Location
Chr.1: 15111815 - 15152464 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_027136.1 5 - 5 77
GenBank mRNA AK074283.1 - 6 - 6 77
AK124431.1 - 5 - 5 77

View Full Product Details