Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- AHCTF1P1
- Entrez Gene ID
-
285116
- Gene Name
- AT-hook containing transcription factor 1 pseudogene 1
- Gene Aliases
-
AHCTF1P, ELYS-like
- Chromosome Location
-
Chr.2: 161499067 - 161507902 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_077058.1 |
|
1 - 1 |
419 |
100 |
GenBank mRNA |
AK057394.1 |
- |
1 - 1 |
253 |
100 |
- Gene Symbol
- AHCTF1
- Entrez Gene ID
-
25909
- Gene Name
- AT-hook containing transcription factor 1
- Gene Aliases
-
ELYS, MST108, MSTP108, TMBS62
- Chromosome Location
-
Chr.1: 246839098 - 246931978 on Build GRCh38
View Full Product Details