The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286808.1 | NP_001273737.1 | 4 - 5 | 843 | 68 |
NM_130809.4 | NP_570721.1 | 4 - 5 | 843 | 68 | |
XM_006714529.3 | XP_006714592.1 | 4 - 5 | 1051 | 68 | |
GenBank mRNA | AJ427339.1 | - | 4 - 5 | 796 | 68 |
AJ441110.3 | - | 4 - 5 | 796 | 68 | |
AJ496322.1 | - | 4 - 5 | 796 | 68 | |
AJ515158.1 | - | 4 - 5 | 796 | 68 | |
AJ515428.1 | - | 4 - 5 | 796 | 68 | |
AJ515429.1 | - | 4 - 5 | 796 | 68 | |
AK057437.1 | - | 4 - 5 | 780 | 68 | |
AK090486.1 | - | 3 - 4 | 668 | 68 | |
AL831963.1 | - | 4 - 5 | 805 | 68 | |
AL832056.1 | - | 4 - 5 | 806 | 68 | |
AL832600.2 | - | 4 - 5 | 802 | 68 | |
BC017066.1 | - | 4 - 5 | 780 | 68 | |
BX099255.1 | - | 1 - 2 | 39 | 68 |