Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62692295-62692795 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BSCL2
Entrez Gene ID
26580
Gene Name
BSCL2, seipin lipid droplet biogenesis associated
Gene Aliases
GNG3LG, HMN5, PELD, SPG17
UniGene
Hs.533709
Gene Chromosome Location
Chr.11: 62690262-62709619 on Build GRCh38
Transcripts
NM_001122955.3 NM_001130702.2 NM_032667.6 NR_037948.1 NR_037949.1 NR_037946.1

Gene Symbol
HNRNPUL2-BSCL2
Entrez Gene ID
100534595
Gene Name
HNRNPUL2-BSCL2 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.533709
Gene Chromosome Location
Chr.11: 62690262-62727384 on Build GRCh38
Transcripts
NM_001122955.3 NM_001130702.2 NM_032667.6 NR_037948.1 NR_037949.1 NR_037946.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6837952 c.580C>A p.P194T Chr.11: 62692467-62692467 on Build GRCh38
6837950 c.662C>G p.T221S Chr.11: 62692385-62692385 on Build GRCh38
6837951 c.772C>A p.P258T Chr.11: 62692467-62692467 on Build GRCh38
6837949 c.854C>G p.T285S Chr.11: 62692385-62692385 on Build GRCh38
5477738 c.759G>T p.E253D Chr.11: 62692669-62692669 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140896339 Chr.11: 62692454-62692454 on Build GRCh38
rs756196182 Chr.11: 62692677-62692677 on Build GRCh38
rs137852975 Chr.11: 62692671-62692671 on Build GRCh38
rs947254577 Chr.11: 62692636-62692636 on Build GRCh38
rs374765276 Chr.11: 62692481-62692481 on Build GRCh38
rs2850597 Chr.11: 62692522-62692522 on Build GRCh38
rs539700774 Chr.11: 62692420-62692420 on Build GRCh38
rs934296756 Chr.11: 62692652-62692652 on Build GRCh38
rs778378228 Chr.11: 62692739-62692739 on Build GRCh38
rs528070411 Chr.11: 62692649-62692649 on Build GRCh38
rs755623017 Chr.11: 62692762-62692762 on Build GRCh38
rs755200939 Chr.11: 62692687-62692687 on Build GRCh38
rs535602209 Chr.11: 62692535-62692535 on Build GRCh38
rs746265239 Chr.11: 62692357-62692357 on Build GRCh38
rs896821643 Chr.11: 62692509-62692509 on Build GRCh38
rs998498207 Chr.11: 62692429-62692429 on Build GRCh38
rs766655185 Chr.11: 62692684-62692684 on Build GRCh38
rs756278107 Chr.11: 62692450-62692450 on Build GRCh38
rs745419166 Chr.11: 62692744-62692744 on Build GRCh38
rs911982128 Chr.11: 62692390-62692390 on Build GRCh38
rs112339725 Chr.11: 62692575-62692575 on Build GRCh38
rs1057517660 Chr.11: 62692475-62692475 on Build GRCh38
rs786205072 Chr.11: 62692371-62692371 on Build GRCh38
rs754251691 Chr.11: 62692612-62692612 on Build GRCh38
rs375554369 Chr.11: 62692393-62692393 on Build GRCh38
rs770600493 Chr.11: 62692480-62692480 on Build GRCh38
rs975256313 Chr.11: 62692598-62692598 on Build GRCh38
rs770234112 Chr.11: 62692360-62692360 on Build GRCh38
rs1023577222 Chr.11: 62692501-62692501 on Build GRCh38
rs995016248 Chr.11: 62692741-62692741 on Build GRCh38
rs368353394 Chr.11: 62692729-62692729 on Build GRCh38
rs761806607 Chr.11: 62692664-62692664 on Build GRCh38
rs773290851 Chr.11: 62692675-62692675 on Build GRCh38
rs765752109 Chr.11: 62692691-62692691 on Build GRCh38
rs10776 Chr.11: 62692683-62692683 on Build GRCh38
rs1022725320 Chr.11: 62692709-62692709 on Build GRCh38
rs754043914 Chr.11: 62692685-62692685 on Build GRCh38
rs752527918 Chr.11: 62692548-62692548 on Build GRCh38
rs755254873 Chr.11: 62692615-62692615 on Build GRCh38
rs747297291 Chr.11: 62692690-62692691 on Build GRCh38
rs775371845 Chr.11: 62692488-62692488 on Build GRCh38
rs748711286 Chr.11: 62692639-62692639 on Build GRCh38
rs766453990 Chr.11: 62692523-62692523 on Build GRCh38
rs112962599 Chr.11: 62692375-62692375 on Build GRCh38
rs769219167 Chr.11: 62692466-62692466 on Build GRCh38
rs183210574 Chr.11: 62692647-62692647 on Build GRCh38
rs950978890 Chr.11: 62692768-62692768 on Build GRCh38
rs774312790 Chr.11: 62692512-62692512 on Build GRCh38
rs1013079991 Chr.11: 62692475-62692475 on Build GRCh38
rs964002420 Chr.11: 62692351-62692351 on Build GRCh38
rs919278014 Chr.11: 62692365-62692365 on Build GRCh38
rs766080496 Chr.11: 62692352-62692352 on Build GRCh38
rs201506985 Chr.11: 62692617-62692617 on Build GRCh38
rs757487399 Chr.11: 62692336-62692336 on Build GRCh38
rs762610540 Chr.11: 62692495-62692495 on Build GRCh38
rs965186450 Chr.11: 62692600-62692600 on Build GRCh38
rs750496537 Chr.11: 62692521-62692521 on Build GRCh38
rs2850596 Chr.11: 62692594-62692594 on Build GRCh38
rs139309430 Chr.11: 62692512-62692513 on Build GRCh38
rs370926100 Chr.11: 62692378-62692378 on Build GRCh38
rs879393922 Chr.11: 62692607-62692607 on Build GRCh38
rs758843908 Chr.11: 62692411-62692411 on Build GRCh38
rs370550718 Chr.11: 62692518-62692518 on Build GRCh38
rs151018278 Chr.11: 62692416-62692416 on Build GRCh38
rs185341934 Chr.11: 62692394-62692394 on Build GRCh38
rs764126144 Chr.11: 62692508-62692508 on Build GRCh38
rs759231362 Chr.11: 62692404-62692404 on Build GRCh38
rs888347900 Chr.11: 62692531-62692531 on Build GRCh38
rs780533531 Chr.11: 62692453-62692453 on Build GRCh38
rs746544677 Chr.11: 62692471-62692471 on Build GRCh38
rs760068361 Chr.11: 62692347-62692347 on Build GRCh38
rs543622840 Chr.11: 62692584-62692584 on Build GRCh38
rs140455908 Chr.11: 62692569-62692569 on Build GRCh38
rs939879170 Chr.11: 62692422-62692422 on Build GRCh38
rs777984500 Chr.11: 62692413-62692414 on Build GRCh38
rs774120735 Chr.11: 62692663-62692663 on Build GRCh38
rs902828050 Chr.11: 62692417-62692417 on Build GRCh38
rs754629431 Chr.11: 62692465-62692465 on Build GRCh38
rs899302880 Chr.11: 62692735-62692735 on Build GRCh38
rs763884653 Chr.11: 62692430-62692430 on Build GRCh38
rs758991179 Chr.11: 62692737-62692737 on Build GRCh38
rs781217574 Chr.11: 62692433-62692433 on Build GRCh38
rs74388071 Chr.11: 62692552-62692552 on Build GRCh38
rs771074843 Chr.11: 62692752-62692753 on Build GRCh38
rs142608646 Chr.11: 62692403-62692403 on Build GRCh38
rs200644629 Chr.11: 62692666-62692666 on Build GRCh38
rs950656045 Chr.11: 62692544-62692544 on Build GRCh38
rs79586077 Chr.11: 62692648-62692648 on Build GRCh38
rs367716875 Chr.11: 62692483-62692483 on Build GRCh38
rs961234088 Chr.11: 62692669-62692669 on Build GRCh38
rs372287991 Chr.11: 62692656-62692656 on Build GRCh38
rs750694484 Chr.11: 62692438-62692438 on Build GRCh38
rs137852971 Chr.11: 62692413-62692413 on Build GRCh38
rs779513433 Chr.11: 62692470-62692470 on Build GRCh38
rs751664305 Chr.11: 62692431-62692431 on Build GRCh38
rs749862649 Chr.11: 62692386-62692386 on Build GRCh38
rs1026942330 Chr.11: 62692525-62692525 on Build GRCh38
rs753110889 Chr.11: 62692340-62692341 on Build GRCh38
rs190842600 Chr.11: 62692395-62692395 on Build GRCh38
rs183259035 Chr.11: 62692335-62692335 on Build GRCh38
rs761718623 Chr.11: 62692517-62692517 on Build GRCh38

View Full Product Details