Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 56360558-56361149 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOF
Entrez Gene ID
319
Gene Name
apolipoprotein F
Gene Aliases
Apo-F, LTIP
UniGene
Hs.534302
Gene Chromosome Location
Chr.12: 56360571-56362799 on Build GRCh38
Transcripts
NM_001638.2

SNP Detail

SNP ID Chromosome Location
rs943031716 Chr.12: 56361057-56361057 on Build GRCh38
rs556718323 Chr.12: 56360695-56360695 on Build GRCh38
rs1022053621 Chr.12: 56360874-56360874 on Build GRCh38
rs926507966 Chr.12: 56360586-56360586 on Build GRCh38
rs71715186 Chr.12: 56360588-56360589 on Build GRCh38
rs750604197 Chr.12: 56361059-56361059 on Build GRCh38
rs916306768 Chr.12: 56360977-56360977 on Build GRCh38
rs901493777 Chr.12: 56361109-56361109 on Build GRCh38
rs939115141 Chr.12: 56360678-56360678 on Build GRCh38
rs1043858855 Chr.12: 56360615-56360615 on Build GRCh38
rs984929421 Chr.12: 56360782-56360782 on Build GRCh38
rs560760041 Chr.12: 56360824-56360824 on Build GRCh38
rs977919471 Chr.12: 56360618-56360618 on Build GRCh38
rs4996382 Chr.12: 56360682-56360682 on Build GRCh38
rs926560397 Chr.12: 56360827-56360827 on Build GRCh38
rs569725411 Chr.12: 56360816-56360816 on Build GRCh38
rs894799481 Chr.12: 56360707-56360707 on Build GRCh38
rs1038017208 Chr.12: 56360719-56360719 on Build GRCh38
rs947818358 Chr.12: 56360993-56360993 on Build GRCh38
rs1025771404 Chr.12: 56360750-56360751 on Build GRCh38
rs1026510652 Chr.12: 56360731-56360731 on Build GRCh38
rs77161851 Chr.12: 56360879-56360879 on Build GRCh38
rs898717816 Chr.12: 56360744-56360744 on Build GRCh38
rs1016721133 Chr.12: 56360812-56360812 on Build GRCh38
rs578185279 Chr.12: 56360696-56360696 on Build GRCh38
rs997705196 Chr.12: 56360708-56360708 on Build GRCh38
rs1043488011 Chr.12: 56361009-56361009 on Build GRCh38
rs891326931 Chr.12: 56360763-56360763 on Build GRCh38
rs528033553 Chr.12: 56360853-56360853 on Build GRCh38
rs923774751 Chr.12: 56360626-56360626 on Build GRCh38
rs966256851 Chr.12: 56360702-56360702 on Build GRCh38
rs751640966 Chr.12: 56360596-56360596 on Build GRCh38
rs759037506 Chr.12: 56360626-56360627 on Build GRCh38
rs371263878 Chr.12: 56360589-56360590 on Build GRCh38
rs867024481 Chr.12: 56361080-56361080 on Build GRCh38
rs892276210 Chr.12: 56360606-56360606 on Build GRCh38
rs903557976 Chr.12: 56360627-56360627 on Build GRCh38
rs188464998 Chr.12: 56361117-56361117 on Build GRCh38
rs967954043 Chr.12: 56360896-56360896 on Build GRCh38
rs560971488 Chr.12: 56361042-56361042 on Build GRCh38
rs1044963813 Chr.12: 56360583-56360583 on Build GRCh38
rs1009348332 Chr.12: 56360607-56360607 on Build GRCh38
rs183407583 Chr.12: 56360691-56360691 on Build GRCh38
rs11575219 Chr.12: 56361075-56361075 on Build GRCh38
rs992024986 Chr.12: 56360915-56360915 on Build GRCh38
rs898156694 Chr.12: 56361061-56361061 on Build GRCh38
rs978455411 Chr.12: 56360935-56360935 on Build GRCh38
rs997799612 Chr.12: 56361116-56361116 on Build GRCh38
rs542235212 Chr.12: 56360910-56360910 on Build GRCh38
rs373605036 Chr.12: 56361071-56361071 on Build GRCh38
rs991849856 Chr.12: 56360955-56360955 on Build GRCh38
rs945202199 Chr.12: 56360949-56360949 on Build GRCh38
rs903568091 Chr.12: 56361037-56361037 on Build GRCh38
rs3038412 Chr.12: 56360589-56360589 on Build GRCh38
rs1037966392 Chr.12: 56360996-56360996 on Build GRCh38
rs140395121 Chr.12: 56360748-56360748 on Build GRCh38
rs200023351 Chr.12: 56360587-56360589 on Build GRCh38
rs764299065 Chr.12: 56360588-56360588 on Build GRCh38
rs913769740 Chr.12: 56360945-56360945 on Build GRCh38
rs11575220 Chr.12: 56361072-56361072 on Build GRCh38

View Full Product Details