The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144701.2 | NP_653302.2 | 7 - 8 | 1037 | 79 |
XM_005270516.2 | XP_005270573.1 | 4 - 5 | 683 | 79 | |
XM_011540790.2 | XP_011539092.1 | 7 - 8 | 1505 | 79 | |
XM_011540791.2 | XP_011539093.1 | 7 - 8 | 1022 | 79 | |
GenBank mRNA | AF461422.1 | - | 7 - 8 | 1037 | 79 |
AM990313.1 | - | 4 - 5 | 723 | 79 | |
AM990314.1 | - | 3 - 4 | 353 | 79 | |
AM990315.1 | - | 3 - 4 | 368 | 79 | |
AM990318.1 | - | 4 - 5 | 514 | 79 | |
AM990319.1 | - | 2 - 3 | 207 | 79 | |
AM990323.1 | - | 4 - 5 | 509 | 79 | |
AM990324.1 | - | 4 - 5 | 443 | 79 | |
AM990325.1 | - | 4 - 5 | 514 | 79 | |
AM990328.1 | - | 3 - 4 | 353 | 79 | |
AM990330.1 | - | 2 - 3 | 207 | 79 | |
AM990332.1 | - | 3 - 4 | 220 | 79 | |
AM990337.1 | - | 4 - 5 | 443 | 79 | |
AY937250.1 | - | 5 - 6 | 844 | 79 | |
AY937251.1 | - | 4 - 5 | 683 | 79 | |
AY937252.1 | - | 4 - 5 | 683 | 79 | |
AY937253.1 | - | 5 - 6 | 844 | 79 | |
AY937254.1 | - | 5 - 6 | 844 | 79 | |
AY937255.1 | - | 5 - 6 | 844 | 79 | |
BC016829.1 | - | 2 - 3 | 240 | 79 | |
FM212241.1 | - | 5 - 6 | 673 | 79 |