Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 129278692-129279241 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HMCES
Entrez Gene ID
56941
Gene Name
5-hydroxymethylcytosine (hmC) binding, ES cell-specific
Gene Aliases
C3orf37, DC12, SRAPD1
UniGene
Hs.458320
Gene Chromosome Location
Chr.3: 129278806-129305293 on Build GRCh38
Transcripts
NM_001006109.1 NM_020187.2 XM_005247636.3 XM_005247637.3 XM_017006877.1

SNP Detail

SNP ID Chromosome Location
rs116769111 Chr.3: 129278859-129278859 on Build GRCh38
rs563170047 Chr.3: 129279185-129279185 on Build GRCh38
rs902525372 Chr.3: 129278753-129278753 on Build GRCh38
rs1011299277 Chr.3: 129279123-129279123 on Build GRCh38
rs1032591332 Chr.3: 129278766-129278766 on Build GRCh38
rs1021341289 Chr.3: 129279130-129279130 on Build GRCh38
rs944689600 Chr.3: 129278755-129278755 on Build GRCh38
rs76359518 Chr.3: 129279214-129279214 on Build GRCh38
rs1034099999 Chr.3: 129278813-129278813 on Build GRCh38
rs774465664 Chr.3: 129278739-129278739 on Build GRCh38
rs907448552 Chr.3: 129279031-129279031 on Build GRCh38
rs117737301 Chr.3: 129278765-129278765 on Build GRCh38
rs1033962659 Chr.3: 129279094-129279094 on Build GRCh38
rs558330306 Chr.3: 129278883-129278883 on Build GRCh38
rs977279939 Chr.3: 129279145-129279145 on Build GRCh38
rs541773958 Chr.3: 129279122-129279122 on Build GRCh38
rs989970039 Chr.3: 129278825-129278825 on Build GRCh38
rs987927923 Chr.3: 129278774-129278774 on Build GRCh38
rs912441939 Chr.3: 129278856-129278856 on Build GRCh38
rs977750501 Chr.3: 129278855-129278855 on Build GRCh38
rs199845051 Chr.3: 129278866-129278868 on Build GRCh38
rs528587945 Chr.3: 129278734-129278734 on Build GRCh38
rs888062003 Chr.3: 129278778-129278778 on Build GRCh38
rs867263791 Chr.3: 129278887-129278887 on Build GRCh38
rs958439360 Chr.3: 129278824-129278824 on Build GRCh38
rs753719430 Chr.3: 129279055-129279055 on Build GRCh38
rs1005565997 Chr.3: 129278781-129278781 on Build GRCh38
rs1037022663 Chr.3: 129279022-129279022 on Build GRCh38
rs1042414240 Chr.3: 129278743-129278743 on Build GRCh38
rs1049708808 Chr.3: 129278721-129278721 on Build GRCh38
rs909258061 Chr.3: 129279223-129279223 on Build GRCh38
rs967618851 Chr.3: 129279132-129279132 on Build GRCh38
rs900586208 Chr.3: 129278764-129278764 on Build GRCh38
rs534515559 Chr.3: 129278986-129278986 on Build GRCh38
rs1040485590 Chr.3: 129278758-129278758 on Build GRCh38
rs574467755 Chr.3: 129279058-129279058 on Build GRCh38
rs564697289 Chr.3: 129278719-129278719 on Build GRCh38
rs769442721 Chr.3: 129278733-129278733 on Build GRCh38
rs976106606 Chr.3: 129279183-129279183 on Build GRCh38
rs1024753300 Chr.3: 129278942-129278943 on Build GRCh38
rs576665113 Chr.3: 129278967-129278967 on Build GRCh38
rs992425192 Chr.3: 129278865-129278865 on Build GRCh38
rs909488817 Chr.3: 129279035-129279035 on Build GRCh38
rs1020365176 Chr.3: 129278780-129278780 on Build GRCh38
rs912504836 Chr.3: 129279152-129279152 on Build GRCh38
rs965313187 Chr.3: 129279165-129279165 on Build GRCh38
rs960566998 Chr.3: 129278831-129278831 on Build GRCh38
rs112795705 Chr.3: 129278848-129278848 on Build GRCh38
rs111584925 Chr.3: 129278803-129278803 on Build GRCh38
rs371629346 Chr.3: 129278730-129278730 on Build GRCh38
rs536980174 Chr.3: 129278864-129278864 on Build GRCh38
rs914388179 Chr.3: 129278839-129278839 on Build GRCh38
rs931943447 Chr.3: 129279192-129279192 on Build GRCh38
rs887959294 Chr.3: 129278990-129278990 on Build GRCh38
rs923410605 Chr.3: 129279076-129279077 on Build GRCh38
rs1015145448 Chr.3: 129278792-129278792 on Build GRCh38
rs183418666 Chr.3: 129278810-129278810 on Build GRCh38
rs936389755 Chr.3: 129279159-129279159 on Build GRCh38
rs971436036 Chr.3: 129278782-129278782 on Build GRCh38
rs985191667 Chr.3: 129279033-129279033 on Build GRCh38
rs945940087 Chr.3: 129278742-129278742 on Build GRCh38
rs892376213 Chr.3: 129278900-129278904 on Build GRCh38
rs527729970 Chr.3: 129278975-129278975 on Build GRCh38
rs940907439 Chr.3: 129279055-129279056 on Build GRCh38
rs927270296 Chr.3: 129278797-129278797 on Build GRCh38
rs922041869 Chr.3: 129278861-129278861 on Build GRCh38
rs569601045 Chr.3: 129278860-129278860 on Build GRCh38
rs552983870 Chr.3: 129278998-129278998 on Build GRCh38
rs1005015400 Chr.3: 129279006-129279006 on Build GRCh38
rs958471804 Chr.3: 129279099-129279099 on Build GRCh38
rs866992690 Chr.3: 129278772-129278772 on Build GRCh38
rs971476149 Chr.3: 129278793-129278793 on Build GRCh38
rs1036145713 Chr.3: 129278740-129278740 on Build GRCh38
rs933059413 Chr.3: 129279097-129279097 on Build GRCh38
rs1009379081 Chr.3: 129278913-129278913 on Build GRCh38

View Full Product Details