Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 186616301-186616700 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AHSG
Entrez Gene ID
197
Gene Name
alpha 2-HS glycoprotein
Gene Aliases
A2HS, AHS, FETUA, HSGA
UniGene
Hs.324746
Gene Chromosome Location
Chr.3: 186612928-186621318 on Build GRCh38
Transcripts
NM_001622.2 XM_017005840.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4657498 c.348C>T p.F116F Chr.3: 186616466-186616466 on Build GRCh38
4799107 c.358A>G p.K120E Chr.3: 186616476-186616476 on Build GRCh38
1182307 c.383T>C p.V128A Chr.3: 186616501-186616501 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs370042922 Chr.3: 186616481-186616481 on Build GRCh38
rs759854890 Chr.3: 186616387-186616387 on Build GRCh38
rs746426747 Chr.3: 186616429-186616429 on Build GRCh38
rs551537344 Chr.3: 186616534-186616534 on Build GRCh38
rs1013003765 Chr.3: 186616372-186616372 on Build GRCh38
rs545113114 Chr.3: 186616673-186616673 on Build GRCh38
rs759889971 Chr.3: 186616396-186616396 on Build GRCh38
rs149190153 Chr.3: 186616369-186616369 on Build GRCh38
rs929104860 Chr.3: 186616329-186616329 on Build GRCh38
rs776029982 Chr.3: 186616446-186616446 on Build GRCh38
rs750452843 Chr.3: 186616393-186616393 on Build GRCh38
rs761554634 Chr.3: 186616505-186616505 on Build GRCh38
rs764228063 Chr.3: 186616562-186616562 on Build GRCh38
rs776787557 Chr.3: 186616345-186616345 on Build GRCh38
rs956328700 Chr.3: 186616405-186616405 on Build GRCh38
rs538131134 Chr.3: 186616596-186616596 on Build GRCh38
rs773162375 Chr.3: 186616513-186616513 on Build GRCh38
rs756150821 Chr.3: 186616410-186616410 on Build GRCh38
rs757682407 Chr.3: 186616425-186616425 on Build GRCh38
rs749508333 Chr.3: 186616422-186616422 on Build GRCh38
rs766370586 Chr.3: 186616400-186616400 on Build GRCh38
rs779215090 Chr.3: 186616426-186616426 on Build GRCh38
rs567591601 Chr.3: 186616570-186616570 on Build GRCh38
rs780576259 Chr.3: 186616400-186616400 on Build GRCh38
rs550608087 Chr.3: 186616343-186616343 on Build GRCh38
rs756960189 Chr.3: 186616506-186616506 on Build GRCh38
rs1012270293 Chr.3: 186616424-186616424 on Build GRCh38
rs556136544 Chr.3: 186616628-186616628 on Build GRCh38
rs555583609 Chr.3: 186616567-186616567 on Build GRCh38
rs777869071 Chr.3: 186616413-186616413 on Build GRCh38
rs751453434 Chr.3: 186616552-186616552 on Build GRCh38
rs145351431 Chr.3: 186616326-186616326 on Build GRCh38
rs769731773 Chr.3: 186616431-186616431 on Build GRCh38
rs564642361 Chr.3: 186616595-186616595 on Build GRCh38
rs776204909 Chr.3: 186616497-186616497 on Build GRCh38
rs887633768 Chr.3: 186616359-186616359 on Build GRCh38
rs1018304137 Chr.3: 186616569-186616569 on Build GRCh38
rs948543782 Chr.3: 186616580-186616580 on Build GRCh38
rs566679913 Chr.3: 186616551-186616551 on Build GRCh38
rs1801111 Chr.3: 186616492-186616492 on Build GRCh38
rs376930727 Chr.3: 186616449-186616449 on Build GRCh38
rs112363978 Chr.3: 186616442-186616442 on Build GRCh38
rs747748363 Chr.3: 186616552-186616553 on Build GRCh38
rs907303941 Chr.3: 186616599-186616599 on Build GRCh38
rs367903461 Chr.3: 186616392-186616392 on Build GRCh38
rs528640304 Chr.3: 186616371-186616371 on Build GRCh38
rs945511983 Chr.3: 186616660-186616660 on Build GRCh38
rs1032316015 Chr.3: 186616397-186616397 on Build GRCh38
rs770748133 Chr.3: 186616344-186616344 on Build GRCh38
rs142477382 Chr.3: 186616670-186616670 on Build GRCh38
rs754130437 Chr.3: 186616403-186616403 on Build GRCh38
rs371601256 Chr.3: 186616394-186616394 on Build GRCh38
rs79747711 Chr.3: 186616554-186616554 on Build GRCh38
rs374387917 Chr.3: 186616420-186616420 on Build GRCh38
rs914076719 Chr.3: 186616609-186616609 on Build GRCh38

View Full Product Details