Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 99853124-99853525 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAPP1
Entrez Gene ID
27071
Gene Name
dual adaptor of phosphotyrosine and 3-phosphoinositides 1
Gene Aliases
BAM32
UniGene
Hs.436271
Gene Chromosome Location
Chr.4: 99816824-99872290 on Build GRCh38
Transcripts
NM_001306151.1 NM_014395.2 XM_011531840.2 XM_011531842.1 XM_011531843.2 XM_017008023.1 XM_017008024.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1049459 c.403G>A p.E135K Chr.4: 99853262-99853262 on Build GRCh38
1049457 c.364C>A p.L122M Chr.4: 99853223-99853223 on Build GRCh38
5529568 c.452G>A p.G151E Chr.4: 99853311-99853311 on Build GRCh38
6290525 c.366G>A p.L122L Chr.4: 99853225-99853225 on Build GRCh38
7363691 c.460G>T p.E154* Chr.4: 99853319-99853319 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1038860181 Chr.4: 99853373-99853373 on Build GRCh38
rs1010236106 Chr.4: 99853431-99853431 on Build GRCh38
rs779751999 Chr.4: 99853192-99853192 on Build GRCh38
rs761728546 Chr.4: 99853168-99853168 on Build GRCh38
rs778388533 Chr.4: 99853193-99853194 on Build GRCh38
rs778276587 Chr.4: 99853198-99853198 on Build GRCh38
rs938148755 Chr.4: 99853386-99853386 on Build GRCh38
rs753290740 Chr.4: 99853193-99853193 on Build GRCh38
rs908547761 Chr.4: 99853281-99853281 on Build GRCh38
rs534926032 Chr.4: 99853365-99853365 on Build GRCh38
rs977857239 Chr.4: 99853494-99853494 on Build GRCh38
rs756706401 Chr.4: 99853192-99853193 on Build GRCh38
rs747477341 Chr.4: 99853201-99853201 on Build GRCh38
rs1056722518 Chr.4: 99853182-99853182 on Build GRCh38
rs1014853943 Chr.4: 99853326-99853326 on Build GRCh38
rs750309314 Chr.4: 99853256-99853256 on Build GRCh38
rs999902893 Chr.4: 99853463-99853463 on Build GRCh38
rs901853231 Chr.4: 99853327-99853327 on Build GRCh38
rs753481523 Chr.4: 99853283-99853283 on Build GRCh38
rs780166482 Chr.4: 99853376-99853376 on Build GRCh38
rs896717304 Chr.4: 99853425-99853425 on Build GRCh38
rs17029601 Chr.4: 99853481-99853481 on Build GRCh38
rs774536733 Chr.4: 99853246-99853246 on Build GRCh38
rs752181928 Chr.4: 99853298-99853298 on Build GRCh38
rs767547987 Chr.4: 99853250-99853250 on Build GRCh38
rs749423368 Chr.4: 99853208-99853208 on Build GRCh38
rs775717540 Chr.4: 99853237-99853237 on Build GRCh38
rs902838960 Chr.4: 99853453-99853453 on Build GRCh38
rs747244412 Chr.4: 99853493-99853493 on Build GRCh38
rs372503686 Chr.4: 99853190-99853190 on Build GRCh38
rs376041630 Chr.4: 99853287-99853287 on Build GRCh38
rs763713752 Chr.4: 99853452-99853452 on Build GRCh38
rs373393300 Chr.4: 99853395-99853395 on Build GRCh38
rs369201523 Chr.4: 99853364-99853364 on Build GRCh38
rs755780195 Chr.4: 99853191-99853191 on Build GRCh38
rs768738803 Chr.4: 99853243-99853243 on Build GRCh38
rs781588588 Chr.4: 99853216-99853216 on Build GRCh38
rs985331890 Chr.4: 99853472-99853472 on Build GRCh38
rs745979526 Chr.4: 99853440-99853440 on Build GRCh38
rs769979490 Chr.4: 99853225-99853225 on Build GRCh38
rs767456253 Chr.4: 99853178-99853178 on Build GRCh38
rs760451809 Chr.4: 99853259-99853259 on Build GRCh38
rs954009935 Chr.4: 99853457-99853457 on Build GRCh38
rs746234384 Chr.4: 99853218-99853218 on Build GRCh38
rs749561215 Chr.4: 99853379-99853379 on Build GRCh38
rs757821465 Chr.4: 99853363-99853363 on Build GRCh38
rs144829888 Chr.4: 99853376-99853377 on Build GRCh38
rs1049610116 Chr.4: 99853202-99853202 on Build GRCh38
rs1032703233 Chr.4: 99853475-99853475 on Build GRCh38
rs768974945 Chr.4: 99853389-99853389 on Build GRCh38
rs771397827 Chr.4: 99853204-99853204 on Build GRCh38
rs933920555 Chr.4: 99853219-99853219 on Build GRCh38
rs762962153 Chr.4: 99853240-99853240 on Build GRCh38
rs756447109 Chr.4: 99853370-99853370 on Build GRCh38
rs967763295 Chr.4: 99853490-99853490 on Build GRCh38
rs140890472 Chr.4: 99853420-99853420 on Build GRCh38
rs374882337 Chr.4: 99853273-99853273 on Build GRCh38
rs548601611 Chr.4: 99853286-99853286 on Build GRCh38
rs761887882 Chr.4: 99853247-99853247 on Build GRCh38
rs754452099 Chr.4: 99853194-99853194 on Build GRCh38

View Full Product Details