Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 119339208-119339701 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1QTNF5
Entrez Gene ID
114902
Gene Name
C1q and tumor necrosis factor related protein 5
Gene Aliases
CTRP5
UniGene
Hs.632102
Gene Chromosome Location
Chr.11: 119338934-119346673 on Build GRCh38
Transcripts
NM_001278431.1 NM_015645.4 NM_031433.3

Gene Symbol
MFRP
Entrez Gene ID
83552
Gene Name
membrane frizzled-related protein
Gene Aliases
MCOP5, NNO2, RD6
UniGene
Hs.632102
Gene Chromosome Location
Chr.11: 119338934-119346673 on Build GRCh38
Transcripts
NM_001278431.1 NM_015645.4 NM_031433.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5882024 c.584_585insG p.M196fs*7 Chr.11: 119339478-119339479 on Build GRCh38
5371808 c.585C>T p.A195A Chr.11: 119339478-119339478 on Build GRCh38
6044779 c.582_583insG p.A195fs*8 Chr.11: 119339480-119339481 on Build GRCh38
5999639 c.575C>T p.S192L Chr.11: 119339488-119339488 on Build GRCh38
5965387 c.380G>A p.R127H Chr.11: 119339683-119339683 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749820882 Chr.11: 119339568-119339568 on Build GRCh38
rs1019399817 Chr.11: 119339259-119339259 on Build GRCh38
rs777149267 Chr.11: 119339313-119339313 on Build GRCh38
rs745716719 Chr.11: 119339400-119339400 on Build GRCh38
rs983080084 Chr.11: 119339295-119339295 on Build GRCh38
rs766867289 Chr.11: 119339479-119339479 on Build GRCh38
rs369261260 Chr.11: 119339323-119339323 on Build GRCh38
rs371634525 Chr.11: 119339496-119339496 on Build GRCh38
rs757188433 Chr.11: 119339491-119339491 on Build GRCh38
rs921543763 Chr.11: 119339349-119339349 on Build GRCh38
rs768085743 Chr.11: 119339429-119339429 on Build GRCh38
rs751663826 Chr.11: 119339346-119339346 on Build GRCh38
rs773466129 Chr.11: 119339328-119339328 on Build GRCh38
rs746520820 Chr.11: 119339481-119339482 on Build GRCh38
rs770249529 Chr.11: 119339438-119339438 on Build GRCh38
rs141453967 Chr.11: 119339514-119339514 on Build GRCh38
rs906525288 Chr.11: 119339494-119339494 on Build GRCh38
rs758129925 Chr.11: 119339505-119339505 on Build GRCh38
rs747647680 Chr.11: 119339523-119339523 on Build GRCh38
rs747980638 Chr.11: 119339565-119339577 on Build GRCh38
rs200181343 Chr.11: 119339297-119339297 on Build GRCh38
rs746563698 Chr.11: 119339290-119339290 on Build GRCh38
rs945456363 Chr.11: 119339482-119339482 on Build GRCh38
rs557404721 Chr.11: 119339515-119339515 on Build GRCh38
rs749758726 Chr.11: 119339389-119339392 on Build GRCh38
rs752412062 Chr.11: 119339673-119339673 on Build GRCh38
rs765574187 Chr.11: 119339325-119339325 on Build GRCh38
rs1004893867 Chr.11: 119339249-119339249 on Build GRCh38
rs9640 Chr.11: 119339269-119339269 on Build GRCh38
rs142208418 Chr.11: 119339330-119339330 on Build GRCh38
rs35448656 Chr.11: 119339662-119339662 on Build GRCh38
rs567782248 Chr.11: 119339240-119339240 on Build GRCh38
rs146971652 Chr.11: 119339670-119339670 on Build GRCh38
rs779732274 Chr.11: 119339419-119339419 on Build GRCh38
rs757154342 Chr.11: 119339379-119339379 on Build GRCh38
rs771385078 Chr.11: 119339579-119339579 on Build GRCh38
rs575398894 Chr.11: 119339647-119339647 on Build GRCh38
rs373753331 Chr.11: 119339329-119339329 on Build GRCh38
rs759871657 Chr.11: 119339481-119339481 on Build GRCh38
rs878943319 Chr.11: 119339663-119339663 on Build GRCh38
rs766109898 Chr.11: 119339660-119339660 on Build GRCh38
rs915293809 Chr.11: 119339478-119339478 on Build GRCh38
rs376692123 Chr.11: 119339391-119339391 on Build GRCh38
rs748636086 Chr.11: 119339436-119339436 on Build GRCh38
rs755057211 Chr.11: 119339361-119339361 on Build GRCh38
rs768357759 Chr.11: 119339510-119339510 on Build GRCh38
rs759811129 Chr.11: 119339615-119339615 on Build GRCh38
rs774880041 Chr.11: 119339582-119339582 on Build GRCh38
rs755515253 Chr.11: 119339676-119339676 on Build GRCh38
rs35341101 Chr.11: 119339681-119339681 on Build GRCh38
rs776247392 Chr.11: 119339510-119339511 on Build GRCh38
rs151230738 Chr.11: 119339487-119339487 on Build GRCh38
rs779555274 Chr.11: 119339480-119339480 on Build GRCh38
rs199537571 Chr.11: 119339511-119339511 on Build GRCh38
rs746676244 Chr.11: 119339421-119339421 on Build GRCh38
rs774976658 Chr.11: 119339280-119339280 on Build GRCh38
rs775635534 Chr.11: 119339625-119339625 on Build GRCh38
rs376231714 Chr.11: 119339631-119339631 on Build GRCh38
rs763473287 Chr.11: 119339465-119339465 on Build GRCh38
rs772224131 Chr.11: 119339621-119339621 on Build GRCh38
rs773626011 Chr.11: 119339462-119339462 on Build GRCh38
rs751401877 Chr.11: 119339669-119339669 on Build GRCh38
rs758015507 Chr.11: 119339410-119339410 on Build GRCh38
rs956797251 Chr.11: 119339406-119339406 on Build GRCh38
rs923859666 Chr.11: 119339352-119339352 on Build GRCh38
rs932791882 Chr.11: 119339386-119339386 on Build GRCh38
rs111033578 Chr.11: 119339574-119339574 on Build GRCh38
rs779003678 Chr.11: 119339547-119339547 on Build GRCh38
rs369839371 Chr.11: 119339479-119339480 on Build GRCh38
rs770325735 Chr.11: 119339540-119339540 on Build GRCh38
rs778087130 Chr.11: 119339565-119339565 on Build GRCh38
rs750204794 Chr.11: 119339497-119339497 on Build GRCh38
rs753740836 Chr.11: 119339377-119339377 on Build GRCh38
rs764246519 Chr.11: 119339638-119339638 on Build GRCh38
rs537084889 Chr.11: 119339324-119339324 on Build GRCh38
rs767522695 Chr.11: 119339362-119339362 on Build GRCh38
rs776032913 Chr.11: 119339433-119339433 on Build GRCh38
rs774651014 Chr.11: 119339480-119339480 on Build GRCh38
rs368962202 Chr.11: 119339648-119339648 on Build GRCh38

View Full Product Details