Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 40251339-40251874 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRK2
Entrez Gene ID
120892
Gene Name
leucine rich repeat kinase 2
Gene Aliases
AURA17, DARDARIN, PARK8, RIPK7, ROCO2
UniGene
Hs.187636
Gene Chromosome Location
Chr.12: 40224895-40369285 on Build GRCh38
Transcripts
NM_198578.3 XM_005268629.3 XM_011537877.2 XM_011537881.2 XM_011537882.2 XM_017018786.1 XM_017018789.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1322535 c.1095C>G p.H365Q Chr.12: 40251368-40251368 on Build GRCh38
1256908 c.1161G>T p.K387N Chr.12: 40251524-40251524 on Build GRCh38
6351848 c.1102-10C>A p.? Chr.12: 40251455-40251455 on Build GRCh38
1676854 c.1113C>A p.C371* Chr.12: 40251476-40251476 on Build GRCh38
5031833 c.1108G>A p.A370T Chr.12: 40251471-40251471 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs959469163 Chr.12: 40251775-40251775 on Build GRCh38
rs1051726518 Chr.12: 40251686-40251686 on Build GRCh38
rs777766351 Chr.12: 40251632-40251632 on Build GRCh38
rs916247113 Chr.12: 40251391-40251391 on Build GRCh38
rs780642151 Chr.12: 40251580-40251580 on Build GRCh38
rs1006803142 Chr.12: 40251569-40251569 on Build GRCh38
rs771950219 Chr.12: 40251420-40251421 on Build GRCh38
rs916785418 Chr.12: 40251707-40251707 on Build GRCh38
rs200189771 Chr.12: 40251471-40251471 on Build GRCh38
rs778567252 Chr.12: 40251423-40251423 on Build GRCh38
rs201929195 Chr.12: 40251517-40251517 on Build GRCh38
rs746474801 Chr.12: 40251432-40251432 on Build GRCh38
rs190626994 Chr.12: 40251618-40251618 on Build GRCh38
rs1000833828 Chr.12: 40251770-40251770 on Build GRCh38
rs777498096 Chr.12: 40251515-40251515 on Build GRCh38
rs764353764 Chr.12: 40251382-40251382 on Build GRCh38
rs199611108 Chr.12: 40251462-40251462 on Build GRCh38
rs999593662 Chr.12: 40251649-40251649 on Build GRCh38
rs1033614413 Chr.12: 40251771-40251771 on Build GRCh38
rs144975499 Chr.12: 40251368-40251368 on Build GRCh38
rs760414206 Chr.12: 40251486-40251488 on Build GRCh38
rs962997402 Chr.12: 40251772-40251772 on Build GRCh38
rs7955902 Chr.12: 40251455-40251455 on Build GRCh38
rs899598240 Chr.12: 40251623-40251623 on Build GRCh38
rs908246228 Chr.12: 40251564-40251564 on Build GRCh38
rs752576004 Chr.12: 40251424-40251424 on Build GRCh38
rs982984779 Chr.12: 40251800-40251800 on Build GRCh38
rs367729913 Chr.12: 40251463-40251463 on Build GRCh38
rs941058459 Chr.12: 40251434-40251434 on Build GRCh38
rs757025267 Chr.12: 40251529-40251529 on Build GRCh38
rs113065049 Chr.12: 40251369-40251369 on Build GRCh38
rs751266778 Chr.12: 40251526-40251526 on Build GRCh38
rs201171722 Chr.12: 40251498-40251498 on Build GRCh38
rs746359858 Chr.12: 40251456-40251456 on Build GRCh38
rs780820960 Chr.12: 40251539-40251539 on Build GRCh38
rs755047938 Chr.12: 40251697-40251697 on Build GRCh38
rs541749121 Chr.12: 40251435-40251435 on Build GRCh38
rs202170315 Chr.12: 40251480-40251480 on Build GRCh38
rs758295804 Chr.12: 40251510-40251510 on Build GRCh38
rs890975722 Chr.12: 40251516-40251516 on Build GRCh38
rs547818910 Chr.12: 40251792-40251792 on Build GRCh38
rs751590550 Chr.12: 40251383-40251383 on Build GRCh38
rs533365997 Chr.12: 40251803-40251803 on Build GRCh38
rs757360567 Chr.12: 40251386-40251386 on Build GRCh38
rs932367002 Chr.12: 40251650-40251650 on Build GRCh38
rs979362047 Chr.12: 40251412-40251412 on Build GRCh38
rs757965270 Chr.12: 40251426-40251426 on Build GRCh38
rs1010272196 Chr.12: 40251718-40251718 on Build GRCh38
rs763402253 Chr.12: 40251464-40251464 on Build GRCh38
rs759359344 Chr.12: 40251486-40251486 on Build GRCh38
rs142060367 Chr.12: 40251619-40251619 on Build GRCh38
rs1021208670 Chr.12: 40251720-40251720 on Build GRCh38
rs991080591 Chr.12: 40251673-40251673 on Build GRCh38
rs766239042 Chr.12: 40251398-40251398 on Build GRCh38
rs767573954 Chr.12: 40251473-40251473 on Build GRCh38
rs961202625 Chr.12: 40251672-40251672 on Build GRCh38
rs1035133181 Chr.12: 40251666-40251666 on Build GRCh38
rs770155733 Chr.12: 40251579-40251579 on Build GRCh38
rs753788515 Chr.12: 40251405-40251405 on Build GRCh38
rs201712413 Chr.12: 40251419-40251419 on Build GRCh38
rs777586316 Chr.12: 40251429-40251429 on Build GRCh38
rs149913043 Chr.12: 40251827-40251827 on Build GRCh38
rs764305897 Chr.12: 40251553-40251559 on Build GRCh38
rs564424994 Chr.12: 40251604-40251604 on Build GRCh38
rs199565026 Chr.12: 40251413-40251413 on Build GRCh38
rs73106357 Chr.12: 40251788-40251788 on Build GRCh38
rs200462425 Chr.12: 40251365-40251365 on Build GRCh38
rs113116409 Chr.12: 40251557-40251557 on Build GRCh38
rs562095384 Chr.12: 40251641-40251641 on Build GRCh38
rs765509204 Chr.12: 40251366-40251366 on Build GRCh38
rs775302781 Chr.12: 40251424-40251426 on Build GRCh38
rs745435795 Chr.12: 40251572-40251572 on Build GRCh38
rs1491939 Chr.12: 40251794-40251794 on Build GRCh38
rs1491938 Chr.12: 40251828-40251828 on Build GRCh38
rs200870486 Chr.12: 40251495-40251495 on Build GRCh38
rs867566547 Chr.12: 40251743-40251743 on Build GRCh38
rs200440892 Chr.12: 40251503-40251503 on Build GRCh38
rs201261152 Chr.12: 40251470-40251470 on Build GRCh38

View Full Product Details