Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 122753579-122754085 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DENR
Entrez Gene ID
8562
Gene Name
density regulated re-initiation and release factor
Gene Aliases
DRP, DRP1, SMAP-3
UniGene
Hs.22393
Gene Chromosome Location
Chr.12: 122752821-122771064 on Build GRCh38
Transcripts
NM_003677.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1197051 c.65A>C p.K22T Chr.12: 122753766-122753766 on Build GRCh38
3457385 c.67T>C p.L23L Chr.12: 122753768-122753768 on Build GRCh38
3416572 c.88C>T p.R30* Chr.12: 122753789-122753789 on Build GRCh38
5653602 c.106+2C>A p.? Chr.12: 122753809-122753809 on Build GRCh38
7153071 c.26G>T p.S9I Chr.12: 122753727-122753727 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs888991684 Chr.12: 122754008-122754008 on Build GRCh38
rs1030177328 Chr.12: 122753682-122753682 on Build GRCh38
rs771012466 Chr.12: 122753918-122753933 on Build GRCh38
rs376132191 Chr.12: 122753796-122753796 on Build GRCh38
rs551308733 Chr.12: 122753814-122753814 on Build GRCh38
rs749337271 Chr.12: 122753776-122753776 on Build GRCh38
rs757045169 Chr.12: 122753750-122753750 on Build GRCh38
rs766807569 Chr.12: 122753818-122753818 on Build GRCh38
rs747927789 Chr.12: 122753964-122753965 on Build GRCh38
rs749388565 Chr.12: 122753684-122753684 on Build GRCh38
rs773681727 Chr.12: 122753718-122753718 on Build GRCh38
rs780915320 Chr.12: 122753763-122753763 on Build GRCh38
rs548355884 Chr.12: 122753643-122753643 on Build GRCh38
rs781165606 Chr.12: 122753668-122753668 on Build GRCh38
rs569567664 Chr.12: 122753826-122753826 on Build GRCh38
rs757023572 Chr.12: 122753658-122753658 on Build GRCh38
rs1052384925 Chr.12: 122753646-122753646 on Build GRCh38
rs60769772 Chr.12: 122753966-122753967 on Build GRCh38
rs780087179 Chr.12: 122753848-122753848 on Build GRCh38
rs764714925 Chr.12: 122753725-122753725 on Build GRCh38
rs762192574 Chr.12: 122753713-122753713 on Build GRCh38
rs756002032 Chr.12: 122753847-122753847 on Build GRCh38
rs1051724229 Chr.12: 122753973-122753973 on Build GRCh38
rs1010120695 Chr.12: 122754000-122754000 on Build GRCh38
rs746753397 Chr.12: 122753657-122753657 on Build GRCh38
rs1016378431 Chr.12: 122754048-122754048 on Build GRCh38
rs995998449 Chr.12: 122753952-122753952 on Build GRCh38
rs968330274 Chr.12: 122754015-122754015 on Build GRCh38
rs1048197691 Chr.12: 122753900-122753900 on Build GRCh38
rs757909355 Chr.12: 122753729-122753729 on Build GRCh38
rs748363220 Chr.12: 122753780-122753780 on Build GRCh38
rs778931158 Chr.12: 122753856-122753856 on Build GRCh38
rs764548752 Chr.12: 122753868-122753868 on Build GRCh38
rs997443330 Chr.12: 122753942-122753942 on Build GRCh38
rs571267825 Chr.12: 122754019-122754019 on Build GRCh38
rs750172400 Chr.12: 122753843-122753843 on Build GRCh38
rs376973866 Chr.12: 122753748-122753748 on Build GRCh38
rs1016114761 Chr.12: 122753629-122753629 on Build GRCh38
rs763752003 Chr.12: 122753736-122753736 on Build GRCh38
rs539929361 Chr.12: 122753949-122753949 on Build GRCh38
rs147566100 Chr.12: 122753842-122753842 on Build GRCh38
rs745783273 Chr.12: 122753764-122753764 on Build GRCh38
rs200474298 Chr.12: 122753766-122753766 on Build GRCh38
rs373035127 Chr.12: 122753851-122753851 on Build GRCh38
rs371824659 Chr.12: 122753777-122753777 on Build GRCh38
rs936407487 Chr.12: 122753916-122753916 on Build GRCh38
rs900308182 Chr.12: 122753630-122753630 on Build GRCh38
rs193015459 Chr.12: 122754037-122754037 on Build GRCh38
rs754682328 Chr.12: 122753819-122753819 on Build GRCh38
rs745840391 Chr.12: 122753669-122753669 on Build GRCh38
rs372438961 Chr.12: 122753716-122753716 on Build GRCh38
rs533787605 Chr.12: 122753962-122753963 on Build GRCh38
rs759327726 Chr.12: 122753669-122753670 on Build GRCh38
rs1052550893 Chr.12: 122753926-122753926 on Build GRCh38
rs763550154 Chr.12: 122753837-122753837 on Build GRCh38
rs948879843 Chr.12: 122753858-122753858 on Build GRCh38
rs991114837 Chr.12: 122753863-122753863 on Build GRCh38
rs773626223 Chr.12: 122753799-122753799 on Build GRCh38
rs997404221 Chr.12: 122753676-122753676 on Build GRCh38
rs751220390 Chr.12: 122753839-122753839 on Build GRCh38
rs752145058 Chr.12: 122753728-122753728 on Build GRCh38
rs368485489 Chr.12: 122753700-122753700 on Build GRCh38
rs761240519 Chr.12: 122753721-122753721 on Build GRCh38
rs188809598 Chr.12: 122753938-122753938 on Build GRCh38
rs775767376 Chr.12: 122753677-122753677 on Build GRCh38
rs79350782 Chr.12: 122753869-122753869 on Build GRCh38
rs762501397 Chr.12: 122753836-122753836 on Build GRCh38
rs768853390 Chr.12: 122753693-122753693 on Build GRCh38
rs942106948 Chr.12: 122753945-122753945 on Build GRCh38
rs767823102 Chr.12: 122753714-122753714 on Build GRCh38
rs58901651 Chr.12: 122753604-122753604 on Build GRCh38
rs900361923 Chr.12: 122753950-122753950 on Build GRCh38
rs566919026 Chr.12: 122753924-122753924 on Build GRCh38
rs890370602 Chr.12: 122753999-122753999 on Build GRCh38
rs907837452 Chr.12: 122753935-122753935 on Build GRCh38
rs78403365 Chr.12: 122754014-122754014 on Build GRCh38
rs780142224 Chr.12: 122753768-122753768 on Build GRCh38
rs1006273352 Chr.12: 122753625-122753625 on Build GRCh38

View Full Product Details