Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 122770421-122770928 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DENR
Entrez Gene ID
8562
Gene Name
density regulated re-initiation and release factor
Gene Aliases
DRP, DRP1, SMAP-3
UniGene
Hs.22393
Gene Chromosome Location
Chr.12: 122752821-122771064 on Build GRCh38
Transcripts
NM_003677.4

SNP Detail

SNP ID Chromosome Location
rs150447436 Chr.12: 122770511-122770511 on Build GRCh38
rs568465621 Chr.12: 122770627-122770627 on Build GRCh38
rs770229659 Chr.12: 122770871-122770871 on Build GRCh38
rs879088546 Chr.12: 122770585-122770585 on Build GRCh38
rs921521941 Chr.12: 122770486-122770486 on Build GRCh38
rs149206665 Chr.12: 122770764-122770764 on Build GRCh38
rs1002543368 Chr.12: 122770670-122770670 on Build GRCh38
rs878913326 Chr.12: 122770894-122770894 on Build GRCh38
rs778217539 Chr.12: 122770819-122770819 on Build GRCh38
rs548395542 Chr.12: 122770780-122770780 on Build GRCh38
rs968535597 Chr.12: 122770888-122770888 on Build GRCh38
rs145365644 Chr.12: 122770601-122770601 on Build GRCh38
rs770501905 Chr.12: 122770684-122770684 on Build GRCh38
rs894422312 Chr.12: 122770712-122770712 on Build GRCh38
rs748839266 Chr.12: 122770635-122770635 on Build GRCh38
rs977198932 Chr.12: 122770889-122770889 on Build GRCh38
rs1006006545 Chr.12: 122770561-122770561 on Build GRCh38
rs1049079767 Chr.12: 122770519-122770519 on Build GRCh38
rs924669621 Chr.12: 122770465-122770465 on Build GRCh38
rs566720002 Chr.12: 122770860-122770860 on Build GRCh38
rs534061291 Chr.12: 122770872-122770872 on Build GRCh38
rs568614696 Chr.12: 122770494-122770494 on Build GRCh38
rs748190836 Chr.12: 122770522-122770523 on Build GRCh38
rs1040981654 Chr.12: 122770479-122770479 on Build GRCh38
rs35460169 Chr.12: 122770816-122770817 on Build GRCh38

View Full Product Details