Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 20457307-20457816 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APEX1
Entrez Gene ID
328
Gene Name
apurinic/apyrimidinic endodeoxyribonuclease 1
Gene Aliases
APE, APE1, APEN, APEX, APX, HAP1, REF1
UniGene
Hs.73722
Gene Chromosome Location
Chr.14: 20455131-20457772 on Build GRCh38
Transcripts
NM_001244249.1 NM_001641.3 NM_080648.2 NM_080649.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4955489 c.839G>C p.W280S Chr.14: 20457390-20457390 on Build GRCh38
4830097 c.768C>G p.L256L Chr.14: 20457319-20457319 on Build GRCh38
7140099 c.820C>A p.R274R Chr.14: 20457371-20457371 on Build GRCh38
2029143 c.804T>G p.T268T Chr.14: 20457355-20457355 on Build GRCh38
95762 c.828G>A p.K276K Chr.14: 20457379-20457379 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs904741245 Chr.14: 20457521-20457521 on Build GRCh38
rs539695269 Chr.14: 20457444-20457444 on Build GRCh38
rs775971709 Chr.14: 20457342-20457343 on Build GRCh38
rs762306905 Chr.14: 20457507-20457507 on Build GRCh38
rs772111581 Chr.14: 20457359-20457359 on Build GRCh38
rs566098919 Chr.14: 20457466-20457466 on Build GRCh38
rs1065749 Chr.14: 20457358-20457358 on Build GRCh38
rs767141318 Chr.14: 20457527-20457527 on Build GRCh38
rs764007376 Chr.14: 20457549-20457549 on Build GRCh38
rs749097921 Chr.14: 20457452-20457452 on Build GRCh38
rs1016848185 Chr.14: 20457352-20457352 on Build GRCh38
rs17112002 Chr.14: 20457510-20457510 on Build GRCh38
rs765357238 Chr.14: 20457545-20457546 on Build GRCh38
rs759225207 Chr.14: 20457526-20457526 on Build GRCh38
rs3189662 Chr.14: 20457694-20457694 on Build GRCh38
rs565835054 Chr.14: 20457418-20457418 on Build GRCh38
rs1034713911 Chr.14: 20457693-20457694 on Build GRCh38
rs1803119 Chr.14: 20457687-20457687 on Build GRCh38
rs967960316 Chr.14: 20457760-20457760 on Build GRCh38
rs771085264 Chr.14: 20457351-20457351 on Build GRCh38
rs777128613 Chr.14: 20457451-20457451 on Build GRCh38
rs1803118 Chr.14: 20457501-20457501 on Build GRCh38
rs770711224 Chr.14: 20457453-20457453 on Build GRCh38
rs987876913 Chr.14: 20457486-20457486 on Build GRCh38
rs754583946 Chr.14: 20457414-20457414 on Build GRCh38
rs576207886 Chr.14: 20457773-20457773 on Build GRCh38
rs777188321 Chr.14: 20457500-20457500 on Build GRCh38
rs756927677 Chr.14: 20457665-20457665 on Build GRCh38
rs1047115775 Chr.14: 20457598-20457598 on Build GRCh38
rs942701110 Chr.14: 20457523-20457523 on Build GRCh38
rs749531006 Chr.14: 20457343-20457343 on Build GRCh38
rs573714948 Chr.14: 20457634-20457634 on Build GRCh38
rs774545253 Chr.14: 20457357-20457357 on Build GRCh38
rs780926903 Chr.14: 20457746-20457746 on Build GRCh38
rs1043739629 Chr.14: 20457514-20457514 on Build GRCh38
rs1022831389 Chr.14: 20457635-20457635 on Build GRCh38
rs773464873 Chr.14: 20457555-20457555 on Build GRCh38
rs755938382 Chr.14: 20457420-20457420 on Build GRCh38
rs61752835 Chr.14: 20457496-20457496 on Build GRCh38
rs754090827 Chr.14: 20457371-20457371 on Build GRCh38
rs751223947 Chr.14: 20457392-20457392 on Build GRCh38
rs776029898 Chr.14: 20457361-20457361 on Build GRCh38
rs746072642 Chr.14: 20457467-20457467 on Build GRCh38
rs376939375 Chr.14: 20457661-20457661 on Build GRCh38
rs761809761 Chr.14: 20457523-20457524 on Build GRCh38
rs1005375385 Chr.14: 20457642-20457642 on Build GRCh38
rs761043096 Chr.14: 20457364-20457364 on Build GRCh38
rs898817536 Chr.14: 20457451-20457451 on Build GRCh38
rs959487488 Chr.14: 20457593-20457593 on Build GRCh38
rs775302420 Chr.14: 20457422-20457423 on Build GRCh38
rs747329195 Chr.14: 20457418-20457419 on Build GRCh38
rs540658996 Chr.14: 20457659-20457659 on Build GRCh38
rs761255137 Chr.14: 20457489-20457489 on Build GRCh38
rs113056798 Chr.14: 20457488-20457488 on Build GRCh38
rs372749250 Chr.14: 20457360-20457360 on Build GRCh38
rs764520459 Chr.14: 20457367-20457367 on Build GRCh38
rs752091024 Chr.14: 20457535-20457535 on Build GRCh38
rs774168347 Chr.14: 20457511-20457511 on Build GRCh38
rs185469522 Chr.14: 20457709-20457709 on Build GRCh38
rs1049498 Chr.14: 20457693-20457693 on Build GRCh38
rs1001883961 Chr.14: 20457681-20457684 on Build GRCh38
rs913647663 Chr.14: 20457731-20457731 on Build GRCh38
rs4748 Chr.14: 20457407-20457407 on Build GRCh38
rs374977590 Chr.14: 20457372-20457372 on Build GRCh38
rs193287013 Chr.14: 20457557-20457557 on Build GRCh38
rs901275905 Chr.14: 20457590-20457590 on Build GRCh38
rs546055483 Chr.14: 20457724-20457724 on Build GRCh38
rs780395464 Chr.14: 20457769-20457769 on Build GRCh38
rs772166431 Chr.14: 20457480-20457480 on Build GRCh38
rs12590363 Chr.14: 20457517-20457517 on Build GRCh38
rs564462170 Chr.14: 20457743-20457743 on Build GRCh38
rs923950459 Chr.14: 20457674-20457674 on Build GRCh38
rs943109400 Chr.14: 20457442-20457442 on Build GRCh38
rs561523887 Chr.14: 20457718-20457720 on Build GRCh38
rs755638723 Chr.14: 20457540-20457540 on Build GRCh38
rs1803120 Chr.14: 20457482-20457482 on Build GRCh38
rs987929416 Chr.14: 20457703-20457703 on Build GRCh38
rs529437043 Chr.14: 20457791-20457791 on Build GRCh38
rs955582331 Chr.14: 20457692-20457692 on Build GRCh38
rs139391369 Chr.14: 20457785-20457785 on Build GRCh38
rs370190429 Chr.14: 20457381-20457381 on Build GRCh38
rs753786698 Chr.14: 20457556-20457556 on Build GRCh38
rs553873319 Chr.14: 20457781-20457781 on Build GRCh38
rs753152822 Chr.14: 20457424-20457424 on Build GRCh38
rs780800339 Chr.14: 20457415-20457415 on Build GRCh38

View Full Product Details