Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 16198023-16198525 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF19
Entrez Gene ID
128272
Gene Name
Rho guanine nucleotide exchange factor 19
Gene Aliases
WGEF
UniGene
Hs.591532
Gene Chromosome Location
Chr.1: 16197856-16212699 on Build GRCh38
Transcripts
NM_153213.3 XM_011540706.2

SNP Detail

SNP ID Chromosome Location
rs546469339 Chr.1: 16198179-16198179 on Build GRCh38
rs1013811762 Chr.1: 16198456-16198456 on Build GRCh38
rs1002107504 Chr.1: 16198071-16198071 on Build GRCh38
rs747296303 Chr.1: 16198327-16198328 on Build GRCh38
rs573808520 Chr.1: 16198483-16198483 on Build GRCh38
rs558511896 Chr.1: 16198453-16198453 on Build GRCh38
rs895435926 Chr.1: 16198118-16198118 on Build GRCh38
rs958343974 Chr.1: 16198212-16198212 on Build GRCh38
rs1025099159 Chr.1: 16198164-16198164 on Build GRCh38
rs987214627 Chr.1: 16198178-16198178 on Build GRCh38
rs567128145 Chr.1: 16198232-16198232 on Build GRCh38
rs1018686662 Chr.1: 16198452-16198452 on Build GRCh38
rs937320600 Chr.1: 16198368-16198368 on Build GRCh38
rs570332272 Chr.1: 16198484-16198484 on Build GRCh38
rs183438458 Chr.1: 16198433-16198433 on Build GRCh38
rs1045781662 Chr.1: 16198356-16198356 on Build GRCh38
rs190306313 Chr.1: 16198283-16198283 on Build GRCh38
rs950476960 Chr.1: 16198173-16198173 on Build GRCh38
rs899042867 Chr.1: 16198393-16198393 on Build GRCh38
rs221031 Chr.1: 16198374-16198374 on Build GRCh38
rs1032416347 Chr.1: 16198208-16198208 on Build GRCh38
rs1026070427 Chr.1: 16198169-16198169 on Build GRCh38
rs1033269456 Chr.1: 16198489-16198489 on Build GRCh38
rs560286239 Chr.1: 16198482-16198482 on Build GRCh38
rs868132516 Chr.1: 16198055-16198055 on Build GRCh38
rs937077363 Chr.1: 16198432-16198432 on Build GRCh38
rs112493299 Chr.1: 16198363-16198363 on Build GRCh38
rs994680873 Chr.1: 16198401-16198401 on Build GRCh38
rs548995683 Chr.1: 16198224-16198224 on Build GRCh38
rs933259972 Chr.1: 16198270-16198270 on Build GRCh38
rs149825220 Chr.1: 16198095-16198095 on Build GRCh38
rs546366714 Chr.1: 16198312-16198312 on Build GRCh38
rs539507586 Chr.1: 16198157-16198157 on Build GRCh38
rs974810112 Chr.1: 16198233-16198233 on Build GRCh38
rs886263091 Chr.1: 16198417-16198417 on Build GRCh38
rs140737590 Chr.1: 16198316-16198316 on Build GRCh38
rs978158778 Chr.1: 16198192-16198192 on Build GRCh38
rs913398360 Chr.1: 16198308-16198308 on Build GRCh38
rs12087967 Chr.1: 16198413-16198413 on Build GRCh38
rs221030 Chr.1: 16198158-16198158 on Build GRCh38
rs145849336 Chr.1: 16198242-16198242 on Build GRCh38
rs907722886 Chr.1: 16198302-16198302 on Build GRCh38

View Full Product Details