Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 16201686-16202179 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF19
Entrez Gene ID
128272
Gene Name
Rho guanine nucleotide exchange factor 19
Gene Aliases
WGEF
UniGene
Hs.591532
Gene Chromosome Location
Chr.1: 16197856-16212699 on Build GRCh38
Transcripts
NM_153213.3 XM_011540706.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5844354 c.2146G>A p.D716N Chr.1: 16201782-16201782 on Build GRCh38
3477616 c.2110C>T p.P704S Chr.1: 16201818-16201818 on Build GRCh38
3863392 c.2130C>T p.V710V Chr.1: 16201798-16201798 on Build GRCh38
4663096 c.2143delG p.E715fs*20 Chr.1: 16201785-16201785 on Build GRCh38
6459565 c.2114A>C p.Q705P Chr.1: 16201814-16201814 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376974048 Chr.1: 16201776-16201776 on Build GRCh38
rs997932068 Chr.1: 16201998-16201998 on Build GRCh38
rs35799309 Chr.1: 16202035-16202036 on Build GRCh38
rs202195679 Chr.1: 16201909-16201909 on Build GRCh38
rs113595914 Chr.1: 16201789-16201789 on Build GRCh38
rs767683114 Chr.1: 16201800-16201800 on Build GRCh38
rs761842509 Chr.1: 16201797-16201798 on Build GRCh38
rs58225172 Chr.1: 16201975-16201976 on Build GRCh38
rs906156855 Chr.1: 16202026-16202026 on Build GRCh38
rs960814486 Chr.1: 16202106-16202106 on Build GRCh38
rs758891803 Chr.1: 16201754-16201754 on Build GRCh38
rs1022517200 Chr.1: 16202045-16202045 on Build GRCh38
rs878958382 Chr.1: 16201746-16201746 on Build GRCh38
rs758908164 Chr.1: 16201865-16201865 on Build GRCh38
rs563865015 Chr.1: 16201747-16201747 on Build GRCh38
rs776860906 Chr.1: 16201779-16201779 on Build GRCh38
rs755610018 Chr.1: 16201741-16201741 on Build GRCh38
rs181467047 Chr.1: 16202043-16202043 on Build GRCh38
rs369851801 Chr.1: 16201810-16201810 on Build GRCh38
rs879350436 Chr.1: 16201729-16201730 on Build GRCh38
rs765148468 Chr.1: 16201978-16201991 on Build GRCh38
rs1003047893 Chr.1: 16202039-16202039 on Build GRCh38
rs770138584 Chr.1: 16201900-16201900 on Build GRCh38
rs778174274 Chr.1: 16201771-16201771 on Build GRCh38
rs992192237 Chr.1: 16202140-16202140 on Build GRCh38
rs6676143 Chr.1: 16201723-16201723 on Build GRCh38
rs539628331 Chr.1: 16202079-16202079 on Build GRCh38
rs757683816 Chr.1: 16201873-16201873 on Build GRCh38
rs781392668 Chr.1: 16201883-16201883 on Build GRCh38
rs377273211 Chr.1: 16201729-16201729 on Build GRCh38
rs906104030 Chr.1: 16201715-16201715 on Build GRCh38
rs751888521 Chr.1: 16201872-16201872 on Build GRCh38
rs146123988 Chr.1: 16201782-16201782 on Build GRCh38
rs968195449 Chr.1: 16202072-16202072 on Build GRCh38
rs772964344 Chr.1: 16201937-16201937 on Build GRCh38
rs200992860 Chr.1: 16201989-16201989 on Build GRCh38
rs558074200 Chr.1: 16201983-16201989 on Build GRCh38
rs751397392 Chr.1: 16201733-16201733 on Build GRCh38
rs557219359 Chr.1: 16202040-16202040 on Build GRCh38
rs767107706 Chr.1: 16201842-16201842 on Build GRCh38
rs221037 Chr.1: 16201738-16201738 on Build GRCh38
rs1044303850 Chr.1: 16201849-16201849 on Build GRCh38
rs200360405 Chr.1: 16201820-16201820 on Build GRCh38
rs879637432 Chr.1: 16201726-16201727 on Build GRCh38
rs1010335052 Chr.1: 16202142-16202142 on Build GRCh38
rs768735917 Chr.1: 16201903-16201903 on Build GRCh38
rs747644797 Chr.1: 16201734-16201734 on Build GRCh38
rs751768949 Chr.1: 16201751-16201752 on Build GRCh38
rs200243292 Chr.1: 16201978-16201989 on Build GRCh38
rs761620921 Chr.1: 16201839-16201839 on Build GRCh38
rs978193033 Chr.1: 16202083-16202088 on Build GRCh38
rs202094944 Chr.1: 16201737-16201737 on Build GRCh38
rs765929754 Chr.1: 16201848-16201848 on Build GRCh38
rs768649657 Chr.1: 16201799-16201799 on Build GRCh38
rs201439499 Chr.1: 16201977-16201990 on Build GRCh38
rs3818162 Chr.1: 16202105-16202105 on Build GRCh38
rs748701756 Chr.1: 16201752-16201752 on Build GRCh38
rs575450111 Chr.1: 16201975-16201981 on Build GRCh38
rs377601379 Chr.1: 16201774-16201774 on Build GRCh38
rs749324899 Chr.1: 16201902-16201902 on Build GRCh38
rs764041675 Chr.1: 16201732-16201732 on Build GRCh38
rs916594176 Chr.1: 16202142-16202142 on Build GRCh38
rs572545970 Chr.1: 16202126-16202126 on Build GRCh38
rs770052418 Chr.1: 16201788-16201788 on Build GRCh38
rs774305149 Chr.1: 16201908-16201908 on Build GRCh38
rs377484946 Chr.1: 16201718-16201718 on Build GRCh38
rs750130078 Chr.1: 16201843-16201843 on Build GRCh38
rs568051462 Chr.1: 16201955-16201955 on Build GRCh38
rs151266607 Chr.1: 16201975-16201988 on Build GRCh38
rs370364259 Chr.1: 16202069-16202069 on Build GRCh38
rs536907559 Chr.1: 16202011-16202011 on Build GRCh38
rs534426729 Chr.1: 16201847-16201847 on Build GRCh38
rs746162562 Chr.1: 16201896-16201896 on Build GRCh38
rs750444474 Chr.1: 16201847-16201848 on Build GRCh38
rs1029775045 Chr.1: 16201999-16201999 on Build GRCh38
rs77969172 Chr.1: 16201975-16201975 on Build GRCh38
rs386628900 Chr.1: 16201975-16201989 on Build GRCh38
rs760611549 Chr.1: 16201979-16201990 on Build GRCh38
rs753339154 Chr.1: 16201860-16201860 on Build GRCh38

View Full Product Details