Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 18871682-18872174 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH4A1
Entrez Gene ID
8659
Gene Name
aldehyde dehydrogenase 4 family member A1
Gene Aliases
ALDH4, P5CD, P5CDh
UniGene
Hs.77448
Gene Chromosome Location
Chr.1: 18871430-18902799 on Build GRCh38
Transcripts
NM_001161504.1 NM_001319218.1 NM_003748.3 NM_170726.2

SNP Detail

SNP ID Chromosome Location
rs1038642381 Chr.1: 18871824-18871824 on Build GRCh38
rs886045704 Chr.1: 18872155-18872155 on Build GRCh38
rs971807996 Chr.1: 18871998-18871998 on Build GRCh38
rs148846466 Chr.1: 18871942-18871942 on Build GRCh38
rs930272413 Chr.1: 18872116-18872116 on Build GRCh38
rs956558603 Chr.1: 18872142-18872142 on Build GRCh38
rs554220453 Chr.1: 18872073-18872073 on Build GRCh38
rs3202002 Chr.1: 18872143-18872143 on Build GRCh38
rs956599027 Chr.1: 18871919-18871919 on Build GRCh38
rs1138269 Chr.1: 18871924-18871924 on Build GRCh38
rs1025836482 Chr.1: 18871860-18871860 on Build GRCh38
rs564620605 Chr.1: 18872107-18872107 on Build GRCh38
rs145580320 Chr.1: 18871796-18871796 on Build GRCh38
rs534240960 Chr.1: 18871868-18871868 on Build GRCh38
rs755604885 Chr.1: 18871907-18871907 on Build GRCh38
rs980320208 Chr.1: 18871727-18871727 on Build GRCh38
rs1034579513 Chr.1: 18872069-18872069 on Build GRCh38
rs1140477 Chr.1: 18871920-18871920 on Build GRCh38
rs898482069 Chr.1: 18872108-18872108 on Build GRCh38
rs778905588 Chr.1: 18872126-18872126 on Build GRCh38
rs898723367 Chr.1: 18871825-18871825 on Build GRCh38
rs566967465 Chr.1: 18871823-18871823 on Build GRCh38
rs766595619 Chr.1: 18872117-18872117 on Build GRCh38
rs181122436 Chr.1: 18871790-18871790 on Build GRCh38
rs1032234050 Chr.1: 18871913-18871913 on Build GRCh38
rs1008772655 Chr.1: 18872121-18872121 on Build GRCh38
rs1008012759 Chr.1: 18871873-18871873 on Build GRCh38
rs886045703 Chr.1: 18871901-18871901 on Build GRCh38
rs891909298 Chr.1: 18871928-18871928 on Build GRCh38
rs962911421 Chr.1: 18872074-18872074 on Build GRCh38
rs767048232 Chr.1: 18871862-18871862 on Build GRCh38
rs1025307025 Chr.1: 18872155-18872155 on Build GRCh38
rs539978283 Chr.1: 18872087-18872087 on Build GRCh38
rs34467064 Chr.1: 18871928-18871928 on Build GRCh38
rs14311 Chr.1: 18872133-18872133 on Build GRCh38
rs538426496 Chr.1: 18871923-18871923 on Build GRCh38
rs927330151 Chr.1: 18872039-18872039 on Build GRCh38
rs1138292 Chr.1: 18871711-18871711 on Build GRCh38
rs796989099 Chr.1: 18871945-18871945 on Build GRCh38
rs927510349 Chr.1: 18871749-18871749 on Build GRCh38
rs899563315 Chr.1: 18872131-18872131 on Build GRCh38
rs542623842 Chr.1: 18872005-18872005 on Build GRCh38
rs543972561 Chr.1: 18872134-18872134 on Build GRCh38
rs994764530 Chr.1: 18871849-18871849 on Build GRCh38
rs1018438743 Chr.1: 18871901-18871901 on Build GRCh38
rs1001457621 Chr.1: 18872047-18872047 on Build GRCh38
rs911538510 Chr.1: 18871880-18871882 on Build GRCh38
rs1138267 Chr.1: 18872082-18872082 on Build GRCh38
rs559093113 Chr.1: 18871888-18871888 on Build GRCh38
rs750054601 Chr.1: 18871863-18871863 on Build GRCh38
rs774495567 Chr.1: 18871912-18871912 on Build GRCh38
rs891334539 Chr.1: 18872120-18872120 on Build GRCh38
rs76525996 Chr.1: 18871805-18871805 on Build GRCh38
rs918736652 Chr.1: 18872090-18872090 on Build GRCh38
rs981553530 Chr.1: 18871966-18871966 on Build GRCh38
rs386629171 Chr.1: 18872133-18872134 on Build GRCh38
rs963683374 Chr.1: 18871906-18871906 on Build GRCh38

View Full Product Details