Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 18874378-18874871 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH4A1
Entrez Gene ID
8659
Gene Name
aldehyde dehydrogenase 4 family member A1
Gene Aliases
ALDH4, P5CD, P5CDh
UniGene
Hs.77448
Gene Chromosome Location
Chr.1: 18871430-18902799 on Build GRCh38
Transcripts
NM_001161504.1 NM_001319218.1 NM_003748.3 NM_170726.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7197499 c.1529C>G p.S510C Chr.1: 18874513-18874513 on Build GRCh38
7147171 c.1518C>G p.I506M Chr.1: 18874524-18874524 on Build GRCh38
4227840 c.1538C>T p.S513L Chr.1: 18874504-18874504 on Build GRCh38
3480530 c.1574C>G p.A525G Chr.1: 18874468-18874468 on Build GRCh38
7335948 c.1551G>T p.Q517H Chr.1: 18874491-18874491 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200711248 Chr.1: 18874480-18874480 on Build GRCh38
rs567423376 Chr.1: 18874798-18874798 on Build GRCh38
rs190551154 Chr.1: 18874630-18874630 on Build GRCh38
rs757032042 Chr.1: 18874471-18874471 on Build GRCh38
rs909198878 Chr.1: 18874640-18874640 on Build GRCh38
rs767145294 Chr.1: 18874503-18874503 on Build GRCh38
rs28758607 Chr.1: 18874652-18874652 on Build GRCh38
rs985102930 Chr.1: 18874815-18874815 on Build GRCh38
rs114610695 Chr.1: 18874850-18874850 on Build GRCh38
rs6665917 Chr.1: 18874690-18874690 on Build GRCh38
rs1012169317 Chr.1: 18874752-18874752 on Build GRCh38
rs78511665 Chr.1: 18874582-18874582 on Build GRCh38
rs1016713276 Chr.1: 18874842-18874842 on Build GRCh38
rs757085430 Chr.1: 18874521-18874521 on Build GRCh38
rs1012453169 Chr.1: 18874527-18874527 on Build GRCh38
rs946531037 Chr.1: 18874456-18874456 on Build GRCh38
rs397702649 Chr.1: 18874718-18874719 on Build GRCh38
rs377587520 Chr.1: 18874575-18874575 on Build GRCh38
rs200337577 Chr.1: 18874467-18874467 on Build GRCh38
rs74056665 Chr.1: 18874789-18874789 on Build GRCh38
rs749103970 Chr.1: 18874469-18874469 on Build GRCh38
rs776912031 Chr.1: 18874485-18874485 on Build GRCh38
rs28529092 Chr.1: 18874414-18874414 on Build GRCh38
rs1036609862 Chr.1: 18874681-18874681 on Build GRCh38
rs35163726 Chr.1: 18874717-18874718 on Build GRCh38
rs369624696 Chr.1: 18874546-18874546 on Build GRCh38
rs61749348 Chr.1: 18874494-18874494 on Build GRCh38
rs753521283 Chr.1: 18874613-18874613 on Build GRCh38
rs902834863 Chr.1: 18874755-18874755 on Build GRCh38
rs536840468 Chr.1: 18874795-18874795 on Build GRCh38
rs750955077 Chr.1: 18874423-18874423 on Build GRCh38
rs35657817 Chr.1: 18874401-18874401 on Build GRCh38
rs34319466 Chr.1: 18874445-18874446 on Build GRCh38
rs150916561 Chr.1: 18874577-18874577 on Build GRCh38
rs754573654 Chr.1: 18874433-18874433 on Build GRCh38
rs751355712 Chr.1: 18874595-18874595 on Build GRCh38
rs367612554 Chr.1: 18874452-18874452 on Build GRCh38
rs772454798 Chr.1: 18874782-18874782 on Build GRCh38
rs563859389 Chr.1: 18874571-18874571 on Build GRCh38
rs750153450 Chr.1: 18874632-18874632 on Build GRCh38
rs779950402 Chr.1: 18874482-18874482 on Build GRCh38
rs972418738 Chr.1: 18874606-18874606 on Build GRCh38
rs1050139862 Chr.1: 18874696-18874696 on Build GRCh38
rs201961228 Chr.1: 18874607-18874607 on Build GRCh38
rs756504063 Chr.1: 18874454-18874454 on Build GRCh38
rs778642174 Chr.1: 18874530-18874530 on Build GRCh38
rs947091135 Chr.1: 18874726-18874726 on Build GRCh38
rs764572959 Chr.1: 18874470-18874471 on Build GRCh38
rs760150335 Chr.1: 18874591-18874591 on Build GRCh38
rs545207632 Chr.1: 18874504-18874504 on Build GRCh38
rs111526409 Chr.1: 18874683-18874683 on Build GRCh38
rs940889890 Chr.1: 18874402-18874402 on Build GRCh38
rs768771214 Chr.1: 18874565-18874565 on Build GRCh38
rs777628556 Chr.1: 18874468-18874468 on Build GRCh38
rs370369233 Chr.1: 18874839-18874839 on Build GRCh38
rs984881827 Chr.1: 18874819-18874819 on Build GRCh38
rs773884295 Chr.1: 18874495-18874495 on Build GRCh38
rs1043139008 Chr.1: 18874732-18874732 on Build GRCh38
rs969902800 Chr.1: 18874818-18874818 on Build GRCh38
rs34857453 Chr.1: 18874780-18874781 on Build GRCh38
rs766982473 Chr.1: 18874538-18874538 on Build GRCh38
rs757840022 Chr.1: 18874475-18874475 on Build GRCh38
rs75528896 Chr.1: 18874797-18874797 on Build GRCh38
rs376695697 Chr.1: 18874539-18874539 on Build GRCh38
rs755659899 Chr.1: 18874454-18874454 on Build GRCh38
rs888782800 Chr.1: 18874475-18874475 on Build GRCh38
rs760227262 Chr.1: 18874513-18874513 on Build GRCh38
rs371499176 Chr.1: 18874520-18874520 on Build GRCh38
rs369003574 Chr.1: 18874578-18874578 on Build GRCh38
rs762329320 Chr.1: 18874492-18874492 on Build GRCh38
rs747269131 Chr.1: 18874483-18874483 on Build GRCh38
rs779536510 Chr.1: 18874481-18874482 on Build GRCh38
rs199789740 Chr.1: 18874472-18874472 on Build GRCh38
rs79648107 Chr.1: 18874705-18874705 on Build GRCh38
rs1035551031 Chr.1: 18874536-18874536 on Build GRCh38
rs374982673 Chr.1: 18874443-18874443 on Build GRCh38
rs763710453 Chr.1: 18874517-18874517 on Build GRCh38
rs773917228 Chr.1: 18874784-18874784 on Build GRCh38
rs6666036 Chr.1: 18874803-18874803 on Build GRCh38
rs199624169 Chr.1: 18874574-18874574 on Build GRCh38
rs747128730 Chr.1: 18874559-18874559 on Build GRCh38
rs959859902 Chr.1: 18874537-18874537 on Build GRCh38
rs780190351 Chr.1: 18874554-18874554 on Build GRCh38
rs767092438 Chr.1: 18874438-18874438 on Build GRCh38
rs754334877 Chr.1: 18874474-18874475 on Build GRCh38
rs985377082 Chr.1: 18874623-18874623 on Build GRCh38
rs74056664 Chr.1: 18874697-18874697 on Build GRCh38
rs768698409 Chr.1: 18874484-18874484 on Build GRCh38

View Full Product Details