Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 18877237-18877727 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH4A1
Entrez Gene ID
8659
Gene Name
aldehyde dehydrogenase 4 family member A1
Gene Aliases
ALDH4, P5CD, P5CDh
UniGene
Hs.77448
Gene Chromosome Location
Chr.1: 18871430-18902799 on Build GRCh38
Transcripts
NM_001161504.1 NM_001319218.1 NM_003748.3 NM_170726.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5779797 c.1085C>T p.P362L Chr.1: 18877468-18877468 on Build GRCh38
1146107 c.1073A>G p.H358R Chr.1: 18877480-18877480 on Build GRCh38
424869 c.1050G>C p.A350A Chr.1: 18877503-18877503 on Build GRCh38
5625711 c.1068G>A p.V356V Chr.1: 18877485-18877485 on Build GRCh38
5779796 c.1085C>T p.P362L Chr.1: 18877468-18877468 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768124510 Chr.1: 18877515-18877515 on Build GRCh38
rs41310410 Chr.1: 18877467-18877467 on Build GRCh38
rs578065275 Chr.1: 18877704-18877704 on Build GRCh38
rs757671701 Chr.1: 18877640-18877640 on Build GRCh38
rs974695765 Chr.1: 18877394-18877394 on Build GRCh38
rs751732137 Chr.1: 18877497-18877497 on Build GRCh38
rs1020074166 Chr.1: 18877539-18877539 on Build GRCh38
rs767652523 Chr.1: 18877559-18877559 on Build GRCh38
rs751507625 Chr.1: 18877548-18877548 on Build GRCh38
rs1028530044 Chr.1: 18877693-18877693 on Build GRCh38
rs1019789439 Chr.1: 18877472-18877472 on Build GRCh38
rs769168827 Chr.1: 18877300-18877300 on Build GRCh38
rs765634333 Chr.1: 18877490-18877491 on Build GRCh38
rs41306567 Chr.1: 18877457-18877457 on Build GRCh38
rs146767266 Chr.1: 18877476-18877476 on Build GRCh38
rs574681452 Chr.1: 18877275-18877275 on Build GRCh38
rs1046360448 Chr.1: 18877508-18877508 on Build GRCh38
rs754246627 Chr.1: 18877376-18877376 on Build GRCh38
rs150927009 Chr.1: 18877587-18877587 on Build GRCh38
rs753784956 Chr.1: 18877268-18877268 on Build GRCh38
rs750978384 Chr.1: 18877360-18877360 on Build GRCh38
rs375007623 Chr.1: 18877380-18877380 on Build GRCh38
rs769168580 Chr.1: 18877659-18877659 on Build GRCh38
rs370298372 Chr.1: 18877273-18877273 on Build GRCh38
rs760173262 Chr.1: 18877468-18877468 on Build GRCh38
rs755188126 Chr.1: 18877433-18877433 on Build GRCh38
rs763881035 Chr.1: 18877390-18877390 on Build GRCh38
rs576948956 Chr.1: 18877524-18877524 on Build GRCh38
rs762419163 Chr.1: 18877376-18877376 on Build GRCh38
rs137852937 Chr.1: 18877498-18877498 on Build GRCh38
rs372020200 Chr.1: 18877407-18877407 on Build GRCh38
rs757034039 Chr.1: 18877403-18877403 on Build GRCh38
rs145243354 Chr.1: 18877480-18877480 on Build GRCh38
rs757752207 Chr.1: 18877302-18877302 on Build GRCh38
rs758441851 Chr.1: 18877413-18877413 on Build GRCh38
rs920216260 Chr.1: 18877626-18877626 on Build GRCh38
rs201696429 Chr.1: 18877305-18877305 on Build GRCh38
rs1056413751 Chr.1: 18877354-18877354 on Build GRCh38
rs951663150 Chr.1: 18877371-18877371 on Build GRCh38
rs766401941 Chr.1: 18877545-18877545 on Build GRCh38
rs144311914 Chr.1: 18877568-18877568 on Build GRCh38
rs763793413 Chr.1: 18877471-18877471 on Build GRCh38
rs6426814 Chr.1: 18877679-18877679 on Build GRCh38
rs1029297447 Chr.1: 18877402-18877402 on Build GRCh38
rs958408213 Chr.1: 18877697-18877697 on Build GRCh38
rs758300845 Chr.1: 18877483-18877483 on Build GRCh38
rs368174095 Chr.1: 18877463-18877463 on Build GRCh38
rs937303527 Chr.1: 18877452-18877452 on Build GRCh38
rs867081902 Chr.1: 18877432-18877432 on Build GRCh38
rs756567062 Chr.1: 18877504-18877504 on Build GRCh38
rs957840703 Chr.1: 18877495-18877495 on Build GRCh38
rs568298612 Chr.1: 18877703-18877703 on Build GRCh38
rs552112789 Chr.1: 18877594-18877594 on Build GRCh38
rs1026927682 Chr.1: 18877370-18877370 on Build GRCh38
rs981449286 Chr.1: 18877441-18877441 on Build GRCh38
rs376209051 Chr.1: 18877381-18877381 on Build GRCh38
rs764424547 Chr.1: 18877630-18877630 on Build GRCh38
rs377590498 Chr.1: 18877259-18877259 on Build GRCh38
rs762655314 Chr.1: 18877541-18877541 on Build GRCh38
rs913349768 Chr.1: 18877420-18877420 on Build GRCh38
rs768179480 Chr.1: 18877580-18877580 on Build GRCh38
rs770756077 Chr.1: 18877460-18877460 on Build GRCh38
rs757748374 Chr.1: 18877488-18877489 on Build GRCh38
rs555976300 Chr.1: 18877686-18877686 on Build GRCh38
rs746571941 Chr.1: 18877513-18877513 on Build GRCh38
rs757119976 Chr.1: 18877271-18877271 on Build GRCh38
rs774448725 Chr.1: 18877608-18877608 on Build GRCh38
rs1011452656 Chr.1: 18877708-18877708 on Build GRCh38
rs779778218 Chr.1: 18877338-18877338 on Build GRCh38
rs750973043 Chr.1: 18877495-18877496 on Build GRCh38
rs376049478 Chr.1: 18877529-18877529 on Build GRCh38
rs748839454 Chr.1: 18877661-18877661 on Build GRCh38
rs1053479092 Chr.1: 18877396-18877396 on Build GRCh38
rs764483099 Chr.1: 18877375-18877376 on Build GRCh38
rs147471420 Chr.1: 18877455-18877455 on Build GRCh38
rs759485232 Chr.1: 18877616-18877616 on Build GRCh38
rs906372772 Chr.1: 18877425-18877425 on Build GRCh38
rs747212428 Chr.1: 18877419-18877419 on Build GRCh38
rs887115410 Chr.1: 18877618-18877618 on Build GRCh38
rs139430775 Chr.1: 18877576-18877576 on Build GRCh38
rs140704780 Chr.1: 18877416-18877416 on Build GRCh38
rs1054483735 Chr.1: 18877314-18877314 on Build GRCh38
rs779514434 Chr.1: 18877575-18877575 on Build GRCh38
rs780624320 Chr.1: 18877586-18877586 on Build GRCh38
rs146741343 Chr.1: 18877569-18877569 on Build GRCh38
rs766376495 Chr.1: 18877493-18877493 on Build GRCh38
rs754399496 Chr.1: 18877501-18877501 on Build GRCh38
rs928657634 Chr.1: 18877449-18877449 on Build GRCh38
rs562344368 Chr.1: 18877454-18877454 on Build GRCh38
rs758838564 Chr.1: 18877653-18877653 on Build GRCh38
rs190991471 Chr.1: 18877389-18877389 on Build GRCh38
rs777494577 Chr.1: 18877263-18877263 on Build GRCh38
rs572502588 Chr.1: 18877316-18877316 on Build GRCh38
rs753976040 Chr.1: 18877637-18877637 on Build GRCh38
rs142969512 Chr.1: 18877505-18877505 on Build GRCh38
rs755759068 Chr.1: 18877663-18877664 on Build GRCh38
rs765042785 Chr.1: 18877479-18877479 on Build GRCh38
rs901571593 Chr.1: 18877673-18877674 on Build GRCh38
rs752281738 Chr.1: 18877589-18877593 on Build GRCh38
rs781564812 Chr.1: 18877453-18877453 on Build GRCh38
rs372376622 Chr.1: 18877560-18877560 on Build GRCh38
rs769491686 Chr.1: 18877530-18877530 on Build GRCh38
rs1019841480 Chr.1: 18877648-18877648 on Build GRCh38
rs747718196 Chr.1: 18877656-18877656 on Build GRCh38
rs901522077 Chr.1: 18877491-18877491 on Build GRCh38
rs755890443 Chr.1: 18877261-18877261 on Build GRCh38
rs368635653 Chr.1: 18877537-18877537 on Build GRCh38
rs760657167 Chr.1: 18877624-18877624 on Build GRCh38
rs921414727 Chr.1: 18877488-18877488 on Build GRCh38
rs995497580 Chr.1: 18877353-18877353 on Build GRCh38
rs780538072 Chr.1: 18877655-18877655 on Build GRCh38
rs542189193 Chr.1: 18877284-18877284 on Build GRCh38
rs368218270 Chr.1: 18877393-18877393 on Build GRCh38
rs1040143488 Chr.1: 18877671-18877671 on Build GRCh38
rs927843333 Chr.1: 18877523-18877523 on Build GRCh38
rs770587213 Chr.1: 18877605-18877605 on Build GRCh38
rs931544592 Chr.1: 18877628-18877628 on Build GRCh38
rs749404523 Chr.1: 18877404-18877404 on Build GRCh38
rs759716094 Chr.1: 18877550-18877550 on Build GRCh38
rs760988407 Chr.1: 18877266-18877267 on Build GRCh38
rs771533896 Chr.1: 18877511-18877511 on Build GRCh38
rs371375350 Chr.1: 18877382-18877382 on Build GRCh38
rs373944705 Chr.1: 18877427-18877427 on Build GRCh38
rs546153073 Chr.1: 18877348-18877348 on Build GRCh38
rs752889674 Chr.1: 18877621-18877621 on Build GRCh38
rs772730217 Chr.1: 18877592-18877592 on Build GRCh38
rs747216007 Chr.1: 18877292-18877292 on Build GRCh38
rs564431083 Chr.1: 18877379-18877379 on Build GRCh38
rs767707551 Chr.1: 18877619-18877619 on Build GRCh38
rs779200944 Chr.1: 18877641-18877641 on Build GRCh38
rs780768733 Chr.1: 18877506-18877506 on Build GRCh38
rs753700338 Chr.1: 18877397-18877397 on Build GRCh38
rs139272675 Chr.1: 18877506-18877506 on Build GRCh38
rs374282940 Chr.1: 18877642-18877642 on Build GRCh38
rs958973427 Chr.1: 18877325-18877325 on Build GRCh38
rs6426813 Chr.1: 18877660-18877660 on Build GRCh38
rs774707581 Chr.1: 18877512-18877512 on Build GRCh38
rs2230705 Chr.1: 18877503-18877503 on Build GRCh38

View Full Product Details