Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 19283904-19284402 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKR7A3
Entrez Gene ID
22977
Gene Name
aldo-keto reductase family 7 member A3
Gene Aliases
AFAR2
UniGene
Hs.6980
Gene Chromosome Location
Chr.1: 19274309-19289362 on Build GRCh38
Transcripts
NM_012067.2 XM_011541046.2 XM_017000714.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1687117 c.782C>T p.P261L Chr.1: 19284048-19284048 on Build GRCh38
4660930 c.778G>A p.A260T Chr.1: 19284052-19284052 on Build GRCh38
6123247 c.744G>A p.V248V Chr.1: 19284086-19284086 on Build GRCh38
6060161 c.728A>T p.E243V Chr.1: 19284102-19284102 on Build GRCh38
4508604 c.781C>T p.P261S Chr.1: 19284049-19284049 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748627699 Chr.1: 19284165-19284165 on Build GRCh38
rs145872076 Chr.1: 19284066-19284066 on Build GRCh38
rs375834244 Chr.1: 19284076-19284076 on Build GRCh38
rs564971201 Chr.1: 19284194-19284194 on Build GRCh38
rs142053998 Chr.1: 19283999-19283999 on Build GRCh38
rs116050237 Chr.1: 19284104-19284104 on Build GRCh38
rs762141039 Chr.1: 19284169-19284169 on Build GRCh38
rs1008549011 Chr.1: 19284376-19284376 on Build GRCh38
rs954905577 Chr.1: 19284036-19284036 on Build GRCh38
rs766240270 Chr.1: 19284024-19284024 on Build GRCh38
rs543771460 Chr.1: 19284232-19284232 on Build GRCh38
rs780563259 Chr.1: 19284090-19284090 on Build GRCh38
rs1038737217 Chr.1: 19284378-19284378 on Build GRCh38
rs993659204 Chr.1: 19284093-19284093 on Build GRCh38
rs777815379 Chr.1: 19284038-19284038 on Build GRCh38
rs746241251 Chr.1: 19284045-19284045 on Build GRCh38
rs754723873 Chr.1: 19284029-19284029 on Build GRCh38
rs764287638 Chr.1: 19284261-19284261 on Build GRCh38
rs373508687 Chr.1: 19284242-19284242 on Build GRCh38
rs750031518 Chr.1: 19283948-19283948 on Build GRCh38
rs757311716 Chr.1: 19284073-19284073 on Build GRCh38
rs917368207 Chr.1: 19284258-19284258 on Build GRCh38
rs777149433 Chr.1: 19284107-19284107 on Build GRCh38
rs570894526 Chr.1: 19284059-19284059 on Build GRCh38
rs779382228 Chr.1: 19284044-19284044 on Build GRCh38
rs757528748 Chr.1: 19284335-19284335 on Build GRCh38
rs762718170 Chr.1: 19284058-19284058 on Build GRCh38
rs758225676 Chr.1: 19283950-19283950 on Build GRCh38
rs1037504533 Chr.1: 19284355-19284355 on Build GRCh38
rs72651437 Chr.1: 19284314-19284314 on Build GRCh38
rs892355405 Chr.1: 19283936-19283936 on Build GRCh38
rs781418544 Chr.1: 19284341-19284341 on Build GRCh38
rs146835084 Chr.1: 19284065-19284065 on Build GRCh38
rs371463766 Chr.1: 19284199-19284199 on Build GRCh38
rs774573028 Chr.1: 19283990-19283990 on Build GRCh38
rs746045705 Chr.1: 19284084-19284084 on Build GRCh38
rs747038097 Chr.1: 19283962-19283962 on Build GRCh38
rs201763306 Chr.1: 19284005-19284005 on Build GRCh38
rs78167373 Chr.1: 19284318-19284318 on Build GRCh38
rs758890877 Chr.1: 19284087-19284087 on Build GRCh38
rs777971619 Chr.1: 19283984-19283984 on Build GRCh38
rs765796933 Chr.1: 19284161-19284161 on Build GRCh38
rs781558274 Chr.1: 19284164-19284164 on Build GRCh38
rs189114834 Chr.1: 19284366-19284366 on Build GRCh38
rs761413936 Chr.1: 19284010-19284010 on Build GRCh38
rs769267124 Chr.1: 19284105-19284105 on Build GRCh38
rs752560085 Chr.1: 19284032-19284032 on Build GRCh38
rs756195533 Chr.1: 19284360-19284360 on Build GRCh38
rs765002399 Chr.1: 19283947-19283947 on Build GRCh38
rs780616670 Chr.1: 19284160-19284160 on Build GRCh38
rs757287437 Chr.1: 19284141-19284141 on Build GRCh38
rs771026464 Chr.1: 19284042-19284042 on Build GRCh38
rs763958435 Chr.1: 19284137-19284137 on Build GRCh38
rs980098068 Chr.1: 19284262-19284262 on Build GRCh38
rs182478203 Chr.1: 19284053-19284053 on Build GRCh38
rs773885100 Chr.1: 19284116-19284116 on Build GRCh38
rs763980486 Chr.1: 19284098-19284098 on Build GRCh38
rs371040540 Chr.1: 19284152-19284152 on Build GRCh38
rs772568783 Chr.1: 19284055-19284055 on Build GRCh38
rs779701218 Chr.1: 19283959-19283959 on Build GRCh38
rs560229900 Chr.1: 19284364-19284364 on Build GRCh38
rs140288397 Chr.1: 19284035-19284035 on Build GRCh38
rs532118015 Chr.1: 19283964-19283964 on Build GRCh38
rs748784235 Chr.1: 19284110-19284110 on Build GRCh38
rs375648246 Chr.1: 19284163-19284163 on Build GRCh38
rs370516171 Chr.1: 19284115-19284115 on Build GRCh38
rs772524247 Chr.1: 19284049-19284049 on Build GRCh38
rs367816925 Chr.1: 19284140-19284140 on Build GRCh38
rs749410677 Chr.1: 19283987-19283987 on Build GRCh38
rs149649188 Chr.1: 19284068-19284068 on Build GRCh38
rs758663519 Chr.1: 19284159-19284159 on Build GRCh38
rs576887633 Chr.1: 19284230-19284230 on Build GRCh38
rs373639940 Chr.1: 19284134-19284134 on Build GRCh38
rs546113976 Chr.1: 19284136-19284136 on Build GRCh38
rs772727900 Chr.1: 19283995-19283995 on Build GRCh38
rs748252452 Chr.1: 19283977-19283977 on Build GRCh38
rs762510449 Chr.1: 19284021-19284021 on Build GRCh38
rs772683849 Chr.1: 19284002-19284002 on Build GRCh38
rs765347218 Chr.1: 19284151-19284151 on Build GRCh38
rs760443505 Chr.1: 19284031-19284031 on Build GRCh38
rs773905797 Chr.1: 19284175-19284175 on Build GRCh38
rs550188783 Chr.1: 19283976-19283976 on Build GRCh38
rs770401138 Chr.1: 19284166-19284166 on Build GRCh38
rs751292586 Chr.1: 19284025-19284025 on Build GRCh38
rs544271165 Chr.1: 19284368-19284368 on Build GRCh38
rs753250106 Chr.1: 19284206-19284206 on Build GRCh38
rs538211675 Chr.1: 19284064-19284064 on Build GRCh38
rs201279439 Chr.1: 19284006-19284006 on Build GRCh38
rs769821963 Chr.1: 19283979-19283979 on Build GRCh38
rs78549294 Chr.1: 19283989-19283989 on Build GRCh38
rs371333518 Chr.1: 19284052-19284052 on Build GRCh38
rs926519649 Chr.1: 19284294-19284294 on Build GRCh38
rs749588080 Chr.1: 19284040-19284040 on Build GRCh38
rs750150431 Chr.1: 19284018-19284018 on Build GRCh38
rs529893100 Chr.1: 19284256-19284256 on Build GRCh38
rs759171776 Chr.1: 19284130-19284130 on Build GRCh38
rs373169386 Chr.1: 19284067-19284067 on Build GRCh38
rs745556797 Chr.1: 19284177-19284177 on Build GRCh38

View Full Product Details