Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 20645170-20645705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PINK1
Entrez Gene ID
65018
Gene Name
PTEN induced putative kinase 1
Gene Aliases
BRPK, PARK6
UniGene
Hs.389171
Gene Chromosome Location
Chr.1: 20633455-20651511 on Build GRCh38
Transcripts
NM_032409.2 NR_046507.1

Gene Symbol
PINK1-AS
Entrez Gene ID
100861548
Gene Name
PINK1 antisense RNA
Gene Aliases
NAPINK1, PINK1-AS1, PINK1AS
UniGene
Hs.710259
Gene Chromosome Location
Chr.1: 20642657-20652193 on Build GRCh38
Transcripts
NM_032409.2 NR_046507.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6415979 c.1004G>A p.S335N Chr.1: 20645604-20645604 on Build GRCh38
6997600 c.1009C>T p.R337C Chr.1: 20645609-20645609 on Build GRCh38
7214424 c.1018G>A p.A340T Chr.1: 20645618-20645618 on Build GRCh38
6540162 c.1077G>T p.A359A Chr.1: 20645677-20645677 on Build GRCh38
1338544 c.1010G>A p.R337H Chr.1: 20645610-20645610 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748041977 Chr.1: 20645609-20645609 on Build GRCh38
rs762909782 Chr.1: 20645614-20645614 on Build GRCh38
rs776485113 Chr.1: 20645544-20645544 on Build GRCh38
rs3131713 Chr.1: 20645555-20645555 on Build GRCh38
rs954989173 Chr.1: 20645203-20645205 on Build GRCh38
rs944841142 Chr.1: 20645505-20645505 on Build GRCh38
rs778010106 Chr.1: 20645569-20645569 on Build GRCh38
rs189813937 Chr.1: 20645210-20645210 on Build GRCh38
rs141923445 Chr.1: 20645444-20645444 on Build GRCh38
rs768609926 Chr.1: 20645543-20645543 on Build GRCh38
rs764648391 Chr.1: 20645648-20645648 on Build GRCh38
rs3738136 Chr.1: 20645618-20645618 on Build GRCh38
rs989267180 Chr.1: 20645261-20645261 on Build GRCh38
rs752773249 Chr.1: 20645490-20645491 on Build GRCh38
rs988870376 Chr.1: 20645554-20645554 on Build GRCh38
rs771510405 Chr.1: 20645529-20645529 on Build GRCh38
rs370470902 Chr.1: 20645665-20645665 on Build GRCh38
rs1043542742 Chr.1: 20645240-20645240 on Build GRCh38
rs373417787 Chr.1: 20645568-20645568 on Build GRCh38
rs55946432 Chr.1: 20645504-20645505 on Build GRCh38
rs746502670 Chr.1: 20645578-20645578 on Build GRCh38
rs776594592 Chr.1: 20645605-20645605 on Build GRCh38
rs113910883 Chr.1: 20645559-20645559 on Build GRCh38
rs760248764 Chr.1: 20645542-20645542 on Build GRCh38
rs12121342 Chr.1: 20645322-20645322 on Build GRCh38
rs910563823 Chr.1: 20645224-20645224 on Build GRCh38
rs759620550 Chr.1: 20645621-20645621 on Build GRCh38
rs559882164 Chr.1: 20645316-20645316 on Build GRCh38
rs928992689 Chr.1: 20645440-20645440 on Build GRCh38
rs973358412 Chr.1: 20645411-20645411 on Build GRCh38
rs527602117 Chr.1: 20645381-20645381 on Build GRCh38
rs780285404 Chr.1: 20645507-20645507 on Build GRCh38
rs376445270 Chr.1: 20645532-20645532 on Build GRCh38
rs11379920 Chr.1: 20645487-20645488 on Build GRCh38
rs529870420 Chr.1: 20645217-20645217 on Build GRCh38
rs149187537 Chr.1: 20645611-20645611 on Build GRCh38
rs543038023 Chr.1: 20645467-20645467 on Build GRCh38
rs764822115 Chr.1: 20645486-20645487 on Build GRCh38
rs890245962 Chr.1: 20645502-20645502 on Build GRCh38
rs746287438 Chr.1: 20645500-20645501 on Build GRCh38
rs772421808 Chr.1: 20645501-20645502 on Build GRCh38
rs926061773 Chr.1: 20645253-20645253 on Build GRCh38
rs971477060 Chr.1: 20645552-20645552 on Build GRCh38
rs28940285 Chr.1: 20645640-20645640 on Build GRCh38
rs755530256 Chr.1: 20645515-20645515 on Build GRCh38
rs766766432 Chr.1: 20645553-20645553 on Build GRCh38
rs770076225 Chr.1: 20645521-20645521 on Build GRCh38
rs1026455828 Chr.1: 20645489-20645489 on Build GRCh38
rs766094815 Chr.1: 20645506-20645510 on Build GRCh38
rs3131712 Chr.1: 20645382-20645382 on Build GRCh38
rs867148175 Chr.1: 20645487-20645487 on Build GRCh38
rs201920474 Chr.1: 20645635-20645635 on Build GRCh38
rs144550985 Chr.1: 20645677-20645677 on Build GRCh38
rs919405996 Chr.1: 20645197-20645197 on Build GRCh38
rs776001129 Chr.1: 20645502-20645503 on Build GRCh38
rs779637743 Chr.1: 20645660-20645660 on Build GRCh38
rs76753586 Chr.1: 20645675-20645675 on Build GRCh38
rs35813094 Chr.1: 20645623-20645623 on Build GRCh38
rs55831733 Chr.1: 20645615-20645615 on Build GRCh38
rs143992206 Chr.1: 20645466-20645466 on Build GRCh38
rs934731465 Chr.1: 20645254-20645254 on Build GRCh38
rs780580396 Chr.1: 20645676-20645676 on Build GRCh38
rs745520017 Chr.1: 20645526-20645526 on Build GRCh38
rs1044281261 Chr.1: 20645366-20645366 on Build GRCh38
rs1024396038 Chr.1: 20645264-20645264 on Build GRCh38
rs926524477 Chr.1: 20645363-20645363 on Build GRCh38
rs895621838 Chr.1: 20645275-20645275 on Build GRCh38
rs767297594 Chr.1: 20645549-20645550 on Build GRCh38
rs769298471 Chr.1: 20645503-20645504 on Build GRCh38
rs762572298 Chr.1: 20645505-20645505 on Build GRCh38
rs751419017 Chr.1: 20645488-20645488 on Build GRCh38
rs115037254 Chr.1: 20645506-20645506 on Build GRCh38
rs764323136 Chr.1: 20645334-20645334 on Build GRCh38
rs376323248 Chr.1: 20645576-20645576 on Build GRCh38
rs756273070 Chr.1: 20645491-20645492 on Build GRCh38
rs137876853 Chr.1: 20645226-20645226 on Build GRCh38
rs779129205 Chr.1: 20645498-20645499 on Build GRCh38
rs756183137 Chr.1: 20645667-20645668 on Build GRCh38
rs56125389 Chr.1: 20645418-20645418 on Build GRCh38
rs749513325 Chr.1: 20645493-20645494 on Build GRCh38
rs554415859 Chr.1: 20645525-20645525 on Build GRCh38
rs925926343 Chr.1: 20645540-20645540 on Build GRCh38
rs199678436 Chr.1: 20645566-20645566 on Build GRCh38
rs956076577 Chr.1: 20645239-20645239 on Build GRCh38
rs763284142 Chr.1: 20645551-20645551 on Build GRCh38
rs754009779 Chr.1: 20645540-20645540 on Build GRCh38
rs184043309 Chr.1: 20645610-20645610 on Build GRCh38
rs900692291 Chr.1: 20645517-20645517 on Build GRCh38
rs750636834 Chr.1: 20645503-20645503 on Build GRCh38
rs778071671 Chr.1: 20645492-20645493 on Build GRCh38
rs981940191 Chr.1: 20645358-20645358 on Build GRCh38
rs773975173 Chr.1: 20645514-20645515 on Build GRCh38
rs757669987 Chr.1: 20645651-20645651 on Build GRCh38
rs562999056 Chr.1: 20645520-20645520 on Build GRCh38
rs890629217 Chr.1: 20645667-20645667 on Build GRCh38
rs768019187 Chr.1: 20645565-20645565 on Build GRCh38
rs142028165 Chr.1: 20645577-20645577 on Build GRCh38
rs747649012 Chr.1: 20645262-20645262 on Build GRCh38
rs879112454 Chr.1: 20645488-20645489 on Build GRCh38
rs1033401309 Chr.1: 20645227-20645227 on Build GRCh38
rs752683079 Chr.1: 20645618-20645618 on Build GRCh38
rs371943401 Chr.1: 20645549-20645549 on Build GRCh38
rs781588772 Chr.1: 20645518-20645518 on Build GRCh38
rs950026122 Chr.1: 20645305-20645305 on Build GRCh38
rs755060576 Chr.1: 20645562-20645562 on Build GRCh38
rs144330127 Chr.1: 20645603-20645603 on Build GRCh38
rs764252130 Chr.1: 20645678-20645679 on Build GRCh38
rs753186891 Chr.1: 20645624-20645624 on Build GRCh38
rs757561288 Chr.1: 20645494-20645495 on Build GRCh38

View Full Product Details