Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 78773956-78774451 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTS7
Entrez Gene ID
11173
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 7
Gene Aliases
ADAM-TS 7, ADAM-TS7, ADAMTS-7
UniGene
Hs.16441
Gene Chromosome Location
Chr.15: 78759203-78811464 on Build GRCh38
Transcripts
NM_014272.4 XM_005254137.3 XM_011521166.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6143021 c.1924G>A p.E642K Chr.15: 78774253-78774253 on Build GRCh38
4827473 c.1955C>T p.T652I Chr.15: 78774222-78774222 on Build GRCh38
1749335 c.1938C>T p.D646D Chr.15: 78774239-78774239 on Build GRCh38
1181946 c.1907C>T p.A636V Chr.15: 78774270-78774270 on Build GRCh38
3887563 c.1977C>T p.A659A Chr.15: 78774200-78774200 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1054001441 Chr.15: 78774363-78774363 on Build GRCh38
rs771174491 Chr.15: 78774079-78774079 on Build GRCh38
rs369820600 Chr.15: 78774341-78774341 on Build GRCh38
rs754611819 Chr.15: 78774210-78774210 on Build GRCh38
rs534697217 Chr.15: 78774101-78774101 on Build GRCh38
rs992656610 Chr.15: 78774277-78774277 on Build GRCh38
rs914165222 Chr.15: 78774373-78774373 on Build GRCh38
rs768360750 Chr.15: 78774407-78774407 on Build GRCh38
rs779733257 Chr.15: 78774116-78774116 on Build GRCh38
rs530384283 Chr.15: 78773980-78773980 on Build GRCh38
rs757504948 Chr.15: 78774314-78774314 on Build GRCh38
rs764661208 Chr.15: 78774195-78774195 on Build GRCh38
rs558614673 Chr.15: 78774089-78774089 on Build GRCh38
rs757937615 Chr.15: 78774270-78774270 on Build GRCh38
rs774952513 Chr.15: 78774244-78774244 on Build GRCh38
rs371964772 Chr.15: 78774197-78774197 on Build GRCh38
rs762538731 Chr.15: 78774347-78774347 on Build GRCh38
rs778574344 Chr.15: 78774264-78774264 on Build GRCh38
rs777586556 Chr.15: 78774143-78774143 on Build GRCh38
rs28639680 Chr.15: 78774096-78774096 on Build GRCh38
rs775693972 Chr.15: 78774184-78774184 on Build GRCh38
rs986155595 Chr.15: 78774042-78774042 on Build GRCh38
rs890516146 Chr.15: 78774416-78774416 on Build GRCh38
rs57852320 Chr.15: 78773984-78773984 on Build GRCh38
rs758381650 Chr.15: 78774228-78774228 on Build GRCh38
rs749478848 Chr.15: 78774296-78774296 on Build GRCh38
rs770010831 Chr.15: 78774180-78774180 on Build GRCh38
rs756540453 Chr.15: 78773982-78773982 on Build GRCh38
rs954832217 Chr.15: 78774039-78774039 on Build GRCh38
rs767733131 Chr.15: 78774204-78774204 on Build GRCh38
rs973191416 Chr.15: 78774070-78774070 on Build GRCh38
rs753382497 Chr.15: 78774117-78774117 on Build GRCh38
rs759227088 Chr.15: 78774256-78774256 on Build GRCh38
rs149091424 Chr.15: 78774272-78774272 on Build GRCh38
rs1030433620 Chr.15: 78774020-78774020 on Build GRCh38
rs374065655 Chr.15: 78774236-78774236 on Build GRCh38
rs62012625 Chr.15: 78774013-78774013 on Build GRCh38
rs773616459 Chr.15: 78774320-78774320 on Build GRCh38
rs770585045 Chr.15: 78774176-78774176 on Build GRCh38
rs764848621 Chr.15: 78774264-78774264 on Build GRCh38
rs548919757 Chr.15: 78774290-78774290 on Build GRCh38
rs775131744 Chr.15: 78774345-78774345 on Build GRCh38
rs529269045 Chr.15: 78774263-78774263 on Build GRCh38
rs774625747 Chr.15: 78774196-78774196 on Build GRCh38
rs563578765 Chr.15: 78774402-78774402 on Build GRCh38
rs770132401 Chr.15: 78774238-78774238 on Build GRCh38
rs752173584 Chr.15: 78774220-78774220 on Build GRCh38
rs200371521 Chr.15: 78774192-78774192 on Build GRCh38
rs74405922 Chr.15: 78774239-78774239 on Build GRCh38
rs781103969 Chr.15: 78774276-78774276 on Build GRCh38
rs553306524 Chr.15: 78774128-78774128 on Build GRCh38
rs34274005 Chr.15: 78774142-78774143 on Build GRCh38
rs745623749 Chr.15: 78774279-78774279 on Build GRCh38
rs112032898 Chr.15: 78774271-78774271 on Build GRCh38
rs375583815 Chr.15: 78774230-78774230 on Build GRCh38
rs570209308 Chr.15: 78774060-78774060 on Build GRCh38
rs780442904 Chr.15: 78774293-78774293 on Build GRCh38
rs564181061 Chr.15: 78774254-78774254 on Build GRCh38
rs531469164 Chr.15: 78774352-78774352 on Build GRCh38
rs371852402 Chr.15: 78774235-78774235 on Build GRCh38
rs892540840 Chr.15: 78773994-78773994 on Build GRCh38
rs146476597 Chr.15: 78774243-78774243 on Build GRCh38
rs963362503 Chr.15: 78774064-78774064 on Build GRCh38
rs762159131 Chr.15: 78774253-78774253 on Build GRCh38
rs57708073 Chr.15: 78774311-78774311 on Build GRCh38
rs945519597 Chr.15: 78774374-78774374 on Build GRCh38
rs778327640 Chr.15: 78774127-78774127 on Build GRCh38
rs772428118 Chr.15: 78774313-78774313 on Build GRCh38
rs561708272 Chr.15: 78774305-78774305 on Build GRCh38
rs746798651 Chr.15: 78774162-78774162 on Build GRCh38
rs753532595 Chr.15: 78774205-78774205 on Build GRCh38
rs761052271 Chr.15: 78774330-78774330 on Build GRCh38
rs866812330 Chr.15: 78774410-78774410 on Build GRCh38
rs537192474 Chr.15: 78774061-78774061 on Build GRCh38
rs748756654 Chr.15: 78774312-78774312 on Build GRCh38
rs771992982 Chr.15: 78774138-78774138 on Build GRCh38
rs140051491 Chr.15: 78774237-78774237 on Build GRCh38
rs754513847 Chr.15: 78774124-78774124 on Build GRCh38
rs755937777 Chr.15: 78774269-78774269 on Build GRCh38
rs368069631 Chr.15: 78774178-78774178 on Build GRCh38
rs778196386 Chr.15: 78774029-78774029 on Build GRCh38
rs370343072 Chr.15: 78774179-78774179 on Build GRCh38
rs771248578 Chr.15: 78774338-78774338 on Build GRCh38
rs764795450 Chr.15: 78774219-78774219 on Build GRCh38

View Full Product Details