Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 20483087-20483627 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSM2A
Entrez Gene ID
123876
Gene Name
acyl-CoA synthetase medium-chain family member 2A
Gene Aliases
A-923A4.1, ACSM2
UniGene
Hs.298252
Gene Chromosome Location
Chr.16: 20451461-20487669 on Build GRCh38
Transcripts
NM_001308169.1 NM_001308172.1 NM_001308954.1 XM_017022923.1 XM_017022926.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5805407 c.1613A>T p.Y538F Chr.16: 20483161-20483161 on Build GRCh38
6143761 c.1629+1G>T p.? Chr.16: 20483178-20483178 on Build GRCh38
1708850 c.1550C>T p.S517F Chr.16: 20483098-20483098 on Build GRCh38
2129912 c.1593G>A p.V531V Chr.16: 20483141-20483141 on Build GRCh38
4431329 c.1629G>C p.K543N Chr.16: 20483177-20483177 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs112576998 Chr.16: 20483427-20483427 on Build GRCh38
rs766707267 Chr.16: 20483162-20483162 on Build GRCh38
rs1006585854 Chr.16: 20483545-20483545 on Build GRCh38
rs34519917 Chr.16: 20483392-20483392 on Build GRCh38
rs773452500 Chr.16: 20483159-20483159 on Build GRCh38
rs980415858 Chr.16: 20483389-20483389 on Build GRCh38
rs749480960 Chr.16: 20483224-20483224 on Build GRCh38
rs758208404 Chr.16: 20483210-20483210 on Build GRCh38
rs771251674 Chr.16: 20483229-20483229 on Build GRCh38
rs917929133 Chr.16: 20483377-20483377 on Build GRCh38
rs775526706 Chr.16: 20483300-20483300 on Build GRCh38
rs553851351 Chr.16: 20483506-20483506 on Build GRCh38
rs917139959 Chr.16: 20483127-20483127 on Build GRCh38
rs868114576 Chr.16: 20483416-20483416 on Build GRCh38
rs777582294 Chr.16: 20483122-20483122 on Build GRCh38
rs890768721 Chr.16: 20483542-20483542 on Build GRCh38
rs992143438 Chr.16: 20483362-20483362 on Build GRCh38
rs776556869 Chr.16: 20483146-20483146 on Build GRCh38
rs370522235 Chr.16: 20483126-20483126 on Build GRCh38
rs1025645938 Chr.16: 20483340-20483340 on Build GRCh38
rs759956173 Chr.16: 20483163-20483163 on Build GRCh38
rs752320053 Chr.16: 20483114-20483114 on Build GRCh38
rs567203092 Chr.16: 20483548-20483549 on Build GRCh38
rs1016412627 Chr.16: 20483344-20483344 on Build GRCh38
rs548701132 Chr.16: 20483255-20483255 on Build GRCh38
rs555213827 Chr.16: 20483293-20483293 on Build GRCh38
rs772091778 Chr.16: 20483140-20483140 on Build GRCh38
rs199504117 Chr.16: 20483263-20483263 on Build GRCh38
rs575406851 Chr.16: 20483375-20483375 on Build GRCh38
rs755691108 Chr.16: 20483179-20483179 on Build GRCh38
rs948105378 Chr.16: 20483385-20483385 on Build GRCh38
rs1009865599 Chr.16: 20483286-20483286 on Build GRCh38
rs761892143 Chr.16: 20483155-20483155 on Build GRCh38
rs138375901 Chr.16: 20483135-20483135 on Build GRCh38
rs151324005 Chr.16: 20483287-20483288 on Build GRCh38
rs200160908 Chr.16: 20483166-20483166 on Build GRCh38
rs144620533 Chr.16: 20483124-20483124 on Build GRCh38
rs1050759529 Chr.16: 20483525-20483525 on Build GRCh38
rs12920486 Chr.16: 20483602-20483602 on Build GRCh38
rs773328012 Chr.16: 20483222-20483222 on Build GRCh38
rs755815714 Chr.16: 20483115-20483115 on Build GRCh38
rs568574847 Chr.16: 20483323-20483323 on Build GRCh38
rs780042157 Chr.16: 20483212-20483212 on Build GRCh38
rs768127780 Chr.16: 20483169-20483169 on Build GRCh38
rs557560338 Chr.16: 20483342-20483342 on Build GRCh38
rs539761155 Chr.16: 20483399-20483399 on Build GRCh38
rs572904158 Chr.16: 20483519-20483519 on Build GRCh38
rs561923469 Chr.16: 20483555-20483555 on Build GRCh38
rs397855149 Chr.16: 20483394-20483394 on Build GRCh38
rs778545036 Chr.16: 20483209-20483209 on Build GRCh38
rs540276419 Chr.16: 20483554-20483554 on Build GRCh38
rs777530765 Chr.16: 20483188-20483188 on Build GRCh38
rs867521600 Chr.16: 20483521-20483521 on Build GRCh38
rs538131595 Chr.16: 20483261-20483261 on Build GRCh38
rs1054291445 Chr.16: 20483428-20483428 on Build GRCh38
rs745774062 Chr.16: 20483136-20483136 on Build GRCh38
rs1018933322 Chr.16: 20483553-20483553 on Build GRCh38
rs757023048 Chr.16: 20483206-20483206 on Build GRCh38
rs950022676 Chr.16: 20483144-20483144 on Build GRCh38
rs60606533 Chr.16: 20483600-20483602 on Build GRCh38
rs180888298 Chr.16: 20483331-20483331 on Build GRCh38
rs769933923 Chr.16: 20483157-20483157 on Build GRCh38
rs1044216347 Chr.16: 20483149-20483149 on Build GRCh38
rs746087264 Chr.16: 20483107-20483107 on Build GRCh38
rs891346153 Chr.16: 20483290-20483290 on Build GRCh38
rs1018551893 Chr.16: 20483307-20483307 on Build GRCh38
rs533724330 Chr.16: 20483220-20483220 on Build GRCh38
rs752090939 Chr.16: 20483172-20483172 on Build GRCh38
rs974245634 Chr.16: 20483335-20483335 on Build GRCh38
rs891515233 Chr.16: 20483473-20483473 on Build GRCh38
rs1007570918 Chr.16: 20483303-20483303 on Build GRCh38
rs753569866 Chr.16: 20483190-20483190 on Build GRCh38
rs111396853 Chr.16: 20483257-20483257 on Build GRCh38
rs945789372 Chr.16: 20483488-20483488 on Build GRCh38
rs763253064 Chr.16: 20483160-20483160 on Build GRCh38
rs374630512 Chr.16: 20483217-20483217 on Build GRCh38
rs962837110 Chr.16: 20483321-20483321 on Build GRCh38

View Full Product Details