Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 20486800-20487312 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSM2A
Entrez Gene ID
123876
Gene Name
acyl-CoA synthetase medium-chain family member 2A
Gene Aliases
A-923A4.1, ACSM2
UniGene
Hs.298252
Gene Chromosome Location
Chr.16: 20451461-20487669 on Build GRCh38
Transcripts
NM_001308169.1 NM_001308172.1 NM_001308954.1 XM_017022923.1 XM_017022926.1

SNP Detail

SNP ID Chromosome Location
rs186143866 Chr.16: 20486976-20486976 on Build GRCh38
rs1055008 Chr.16: 20486829-20486829 on Build GRCh38
rs1055012 Chr.16: 20486897-20486897 on Build GRCh38
rs1007388413 Chr.16: 20486971-20486971 on Build GRCh38
rs868540081 Chr.16: 20487159-20487159 on Build GRCh38
rs1055007 Chr.16: 20486824-20486824 on Build GRCh38
rs934759907 Chr.16: 20487130-20487130 on Build GRCh38
rs570006486 Chr.16: 20487096-20487097 on Build GRCh38
rs183726353 Chr.16: 20486913-20486913 on Build GRCh38
rs1015978457 Chr.16: 20486977-20486977 on Build GRCh38
rs575106033 Chr.16: 20486853-20486853 on Build GRCh38
rs201108656 Chr.16: 20487064-20487064 on Build GRCh38
rs1051632097 Chr.16: 20487138-20487138 on Build GRCh38
rs1026416266 Chr.16: 20487023-20487023 on Build GRCh38
rs993147772 Chr.16: 20486994-20486994 on Build GRCh38
rs993064898 Chr.16: 20487042-20487042 on Build GRCh38
rs182537812 Chr.16: 20487275-20487275 on Build GRCh38
rs142302909 Chr.16: 20487278-20487278 on Build GRCh38
rs867835932 Chr.16: 20487004-20487004 on Build GRCh38
rs528569251 Chr.16: 20486980-20486980 on Build GRCh38
rs751125713 Chr.16: 20486931-20486931 on Build GRCh38
rs922582710 Chr.16: 20487132-20487135 on Build GRCh38
rs768868720 Chr.16: 20486997-20486997 on Build GRCh38
rs144005474 Chr.16: 20487141-20487141 on Build GRCh38
rs373739790 Chr.16: 20487222-20487222 on Build GRCh38
rs993634343 Chr.16: 20487225-20487225 on Build GRCh38
rs532909922 Chr.16: 20487203-20487203 on Build GRCh38
rs1037950383 Chr.16: 20487219-20487219 on Build GRCh38
rs147663226 Chr.16: 20487182-20487182 on Build GRCh38
rs926069647 Chr.16: 20486837-20486837 on Build GRCh38
rs1055011 Chr.16: 20486846-20486846 on Build GRCh38
rs916198653 Chr.16: 20487045-20487045 on Build GRCh38
rs542130720 Chr.16: 20486861-20486861 on Build GRCh38
rs1053543513 Chr.16: 20486922-20486922 on Build GRCh38
rs763758573 Chr.16: 20487094-20487097 on Build GRCh38
rs963463700 Chr.16: 20486986-20486986 on Build GRCh38
rs879872404 Chr.16: 20487256-20487256 on Build GRCh38
rs1055014 Chr.16: 20486938-20486938 on Build GRCh38
rs943169711 Chr.16: 20487211-20487211 on Build GRCh38
rs563358363 Chr.16: 20486893-20486893 on Build GRCh38
rs737900 Chr.16: 20487078-20487078 on Build GRCh38
rs934645152 Chr.16: 20486838-20486838 on Build GRCh38
rs139949569 Chr.16: 20487026-20487026 on Build GRCh38
rs145360163 Chr.16: 20487061-20487061 on Build GRCh38
rs1051661530 Chr.16: 20486970-20486972 on Build GRCh38
rs1055009 Chr.16: 20486839-20486839 on Build GRCh38
rs867162513 Chr.16: 20487013-20487013 on Build GRCh38
rs997370872 Chr.16: 20486939-20486939 on Build GRCh38
rs543556152 Chr.16: 20487137-20487148 on Build GRCh38
rs888943022 Chr.16: 20486970-20486970 on Build GRCh38
rs189556986 Chr.16: 20487131-20487131 on Build GRCh38
rs978933740 Chr.16: 20487081-20487081 on Build GRCh38
rs1047978015 Chr.16: 20487270-20487270 on Build GRCh38
rs1026091848 Chr.16: 20487241-20487241 on Build GRCh38
rs202194340 Chr.16: 20487067-20487067 on Build GRCh38
rs926031278 Chr.16: 20487090-20487093 on Build GRCh38

View Full Product Details