Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 20661708-20662208 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSM1
Entrez Gene ID
116285
Gene Name
acyl-CoA synthetase medium-chain family member 1
Gene Aliases
BUCS1, MACS1
UniGene
Hs.306812
Gene Chromosome Location
Chr.16: 20623235-20697788 on Build GRCh38
Transcripts
NM_001318890.1 NM_052956.2 NR_134918.1 XM_006721016.2 XM_006721017.2 XM_006721018.2 XM_011545729.2 XM_011545730.2 XM_011545731.2 XM_011545732.2 XM_017022914.1 XM_017022915.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3507034 c.944G>T p.W315L Chr.16: 20661842-20661842 on Build GRCh38
7196374 c.941T>C p.F314S Chr.16: 20661845-20661845 on Build GRCh38
3507032 c.981G>A p.Q327Q Chr.16: 20661805-20661805 on Build GRCh38
3507033 c.956C>T p.S319F Chr.16: 20661830-20661830 on Build GRCh38
702447 c.913-1G>C p.? Chr.16: 20661874-20661874 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs541839746 Chr.16: 20662006-20662023 on Build GRCh38
rs1018691293 Chr.16: 20662066-20662066 on Build GRCh38
rs952298230 Chr.16: 20662149-20662149 on Build GRCh38
rs995608713 Chr.16: 20662088-20662088 on Build GRCh38
rs145893961 Chr.16: 20661842-20661842 on Build GRCh38
rs985459828 Chr.16: 20662153-20662153 on Build GRCh38
rs762313790 Chr.16: 20661825-20661825 on Build GRCh38
rs751345801 Chr.16: 20661775-20661775 on Build GRCh38
rs376985029 Chr.16: 20661822-20661822 on Build GRCh38
rs754844249 Chr.16: 20661780-20661780 on Build GRCh38
rs748159478 Chr.16: 20661791-20661791 on Build GRCh38
rs970446826 Chr.16: 20662159-20662159 on Build GRCh38
rs377430734 Chr.16: 20661846-20661846 on Build GRCh38
rs370280979 Chr.16: 20661909-20661909 on Build GRCh38
rs551674375 Chr.16: 20662075-20662075 on Build GRCh38
rs770320691 Chr.16: 20661910-20661910 on Build GRCh38
rs563852384 Chr.16: 20662076-20662076 on Build GRCh38
rs927117326 Chr.16: 20661833-20661833 on Build GRCh38
rs756142236 Chr.16: 20661851-20661851 on Build GRCh38
rs764670497 Chr.16: 20661827-20661827 on Build GRCh38
rs888795178 Chr.16: 20662008-20662008 on Build GRCh38
rs747324224 Chr.16: 20661887-20661887 on Build GRCh38
rs145195826 Chr.16: 20661849-20661849 on Build GRCh38
rs981496013 Chr.16: 20662168-20662168 on Build GRCh38
rs762630239 Chr.16: 20661839-20661839 on Build GRCh38
rs772056578 Chr.16: 20662083-20662083 on Build GRCh38
rs117587770 Chr.16: 20662030-20662030 on Build GRCh38
rs780974879 Chr.16: 20661788-20661788 on Build GRCh38
rs1009833004 Chr.16: 20661955-20661955 on Build GRCh38
rs758064480 Chr.16: 20661759-20661759 on Build GRCh38
rs140739677 Chr.16: 20661831-20661831 on Build GRCh38
rs768837248 Chr.16: 20661889-20661889 on Build GRCh38
rs915890288 Chr.16: 20662156-20662156 on Build GRCh38
rs146842441 Chr.16: 20661821-20661821 on Build GRCh38
rs1011247582 Chr.16: 20661968-20661968 on Build GRCh38
rs1046253005 Chr.16: 20661902-20661902 on Build GRCh38
rs764795498 Chr.16: 20661752-20661752 on Build GRCh38
rs928748248 Chr.16: 20661908-20661908 on Build GRCh38
rs750011994 Chr.16: 20661754-20661754 on Build GRCh38
rs754722445 Chr.16: 20661843-20661843 on Build GRCh38
rs963171653 Chr.16: 20662059-20662059 on Build GRCh38
rs757336581 Chr.16: 20661793-20661793 on Build GRCh38
rs140011050 Chr.16: 20662067-20662067 on Build GRCh38
rs775819776 Chr.16: 20661817-20661817 on Build GRCh38
rs765919986 Chr.16: 20661840-20661840 on Build GRCh38
rs531447696 Chr.16: 20662026-20662026 on Build GRCh38
rs746038493 Chr.16: 20661799-20661799 on Build GRCh38
rs1007248147 Chr.16: 20662049-20662049 on Build GRCh38
rs928718976 Chr.16: 20662177-20662177 on Build GRCh38
rs776994288 Chr.16: 20661905-20661905 on Build GRCh38
rs1043367881 Chr.16: 20661978-20661978 on Build GRCh38
rs778928233 Chr.16: 20661920-20661923 on Build GRCh38
rs1051296279 Chr.16: 20661963-20661963 on Build GRCh38
rs892345035 Chr.16: 20661924-20661924 on Build GRCh38
rs773751250 Chr.16: 20661917-20661917 on Build GRCh38
rs182085130 Chr.16: 20661972-20661972 on Build GRCh38
rs993664879 Chr.16: 20662095-20662095 on Build GRCh38
rs758552861 Chr.16: 20661883-20661883 on Build GRCh38
rs915908339 Chr.16: 20661829-20661829 on Build GRCh38
rs745909979 Chr.16: 20661877-20661877 on Build GRCh38
rs571345979 Chr.16: 20662037-20662037 on Build GRCh38
rs772735675 Chr.16: 20661838-20661838 on Build GRCh38
rs766152268 Chr.16: 20661761-20661761 on Build GRCh38
rs762349420 Chr.16: 20661919-20661919 on Build GRCh38
rs772214028 Chr.16: 20661811-20661811 on Build GRCh38
rs1026489165 Chr.16: 20662124-20662124 on Build GRCh38
rs779095607 Chr.16: 20661858-20661858 on Build GRCh38
rs143826162 Chr.16: 20661796-20661796 on Build GRCh38

View Full Product Details