Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67229701-67230193 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FHOD1
Entrez Gene ID
29109
Gene Name
formin homology 2 domain containing 1
Gene Aliases
FHOS
UniGene
Hs.95231
Gene Chromosome Location
Chr.16: 67229389-67247522 on Build GRCh38
Transcripts
NM_001318202.1 NM_013241.2 XM_006721180.1 XM_011523043.2 XM_011523044.1 XM_011523045.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
972356 c.3204C>T p.R1068R Chr.16: 67230076-67230076 on Build GRCh38
3387535 c.3399C>A p.G1133G Chr.16: 67229806-67229806 on Build GRCh38
5706174 c.3327C>A p.D1109E Chr.16: 67229878-67229878 on Build GRCh38
972355 c.3258C>A p.P1086P Chr.16: 67229947-67229947 on Build GRCh38
6663964 c.3178A>G p.R1060G Chr.16: 67230102-67230102 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750859651 Chr.16: 67229752-67229752 on Build GRCh38
rs1007067939 Chr.16: 67230081-67230081 on Build GRCh38
rs781372321 Chr.16: 67230156-67230156 on Build GRCh38
rs770847586 Chr.16: 67229802-67229802 on Build GRCh38
rs747207376 Chr.16: 67229964-67229964 on Build GRCh38
rs145161451 Chr.16: 67230074-67230074 on Build GRCh38
rs775105803 Chr.16: 67229811-67229811 on Build GRCh38
rs777850281 Chr.16: 67229810-67229810 on Build GRCh38
rs755096285 Chr.16: 67229873-67229873 on Build GRCh38
rs922908489 Chr.16: 67229786-67229786 on Build GRCh38
rs754043657 Chr.16: 67229779-67229779 on Build GRCh38
rs139955742 Chr.16: 67229821-67229821 on Build GRCh38
rs994894216 Chr.16: 67229934-67229934 on Build GRCh38
rs751662917 Chr.16: 67230088-67230088 on Build GRCh38
rs754002414 Chr.16: 67229864-67229864 on Build GRCh38
rs772730223 Chr.16: 67230032-67230032 on Build GRCh38
rs763111646 Chr.16: 67230077-67230077 on Build GRCh38
rs570015015 Chr.16: 67229922-67229922 on Build GRCh38
rs371052270 Chr.16: 67230064-67230064 on Build GRCh38
rs886581292 Chr.16: 67230013-67230013 on Build GRCh38
rs983111425 Chr.16: 67230153-67230153 on Build GRCh38
rs745776764 Chr.16: 67229854-67229855 on Build GRCh38
rs758732717 Chr.16: 67229999-67229999 on Build GRCh38
rs752991729 Chr.16: 67229996-67229996 on Build GRCh38
rs905983421 Chr.16: 67229884-67229884 on Build GRCh38
rs756157624 Chr.16: 67229754-67229754 on Build GRCh38
rs760072377 Chr.16: 67230060-67230060 on Build GRCh38
rs925773671 Chr.16: 67229745-67229745 on Build GRCh38
rs774001965 Chr.16: 67229954-67229954 on Build GRCh38
rs765351934 Chr.16: 67229991-67229991 on Build GRCh38
rs778956538 Chr.16: 67229788-67229788 on Build GRCh38
rs555521387 Chr.16: 67229966-67229966 on Build GRCh38
rs753800789 Chr.16: 67230099-67230099 on Build GRCh38
rs764198081 Chr.16: 67230004-67230004 on Build GRCh38
rs369591941 Chr.16: 67230078-67230078 on Build GRCh38
rs372458150 Chr.16: 67229940-67229940 on Build GRCh38
rs955879919 Chr.16: 67230112-67230112 on Build GRCh38
rs773670831 Chr.16: 67229967-67229967 on Build GRCh38
rs747474921 Chr.16: 67230038-67230038 on Build GRCh38
rs745762059 Chr.16: 67229809-67229809 on Build GRCh38
rs376653596 Chr.16: 67230122-67230122 on Build GRCh38
rs368550201 Chr.16: 67229827-67229827 on Build GRCh38
rs754951818 Chr.16: 67230108-67230108 on Build GRCh38
rs368888802 Chr.16: 67230142-67230142 on Build GRCh38
rs771797116 Chr.16: 67229907-67229907 on Build GRCh38
rs776135636 Chr.16: 67230162-67230162 on Build GRCh38
rs777000933 Chr.16: 67229985-67229985 on Build GRCh38
rs752899995 Chr.16: 67229877-67229877 on Build GRCh38
rs766767305 Chr.16: 67229857-67229857 on Build GRCh38
rs373710737 Chr.16: 67230150-67230150 on Build GRCh38
rs757844941 Chr.16: 67230125-67230125 on Build GRCh38
rs577717314 Chr.16: 67229726-67229726 on Build GRCh38
rs768077398 Chr.16: 67229746-67229746 on Build GRCh38
rs755853877 Chr.16: 67230014-67230014 on Build GRCh38
rs895068442 Chr.16: 67229896-67229896 on Build GRCh38
rs886507546 Chr.16: 67230168-67230168 on Build GRCh38
rs775142945 Chr.16: 67230010-67230010 on Build GRCh38
rs748689233 Chr.16: 67229731-67229731 on Build GRCh38
rs767808985 Chr.16: 67229822-67229822 on Build GRCh38
rs746584932 Chr.16: 67230149-67230149 on Build GRCh38
rs746859450 Chr.16: 67229801-67229801 on Build GRCh38
rs567735938 Chr.16: 67230029-67230029 on Build GRCh38
rs150018904 Chr.16: 67229875-67229875 on Build GRCh38
rs1005111288 Chr.16: 67230020-67230020 on Build GRCh38
rs766037000 Chr.16: 67230093-67230093 on Build GRCh38
rs191057325 Chr.16: 67229730-67229730 on Build GRCh38
rs539707576 Chr.16: 67229817-67229817 on Build GRCh38
rs762346925 Chr.16: 67229816-67229816 on Build GRCh38
rs999475988 Chr.16: 67229992-67229992 on Build GRCh38
rs748893339 Chr.16: 67229931-67229931 on Build GRCh38
rs777210595 Chr.16: 67230048-67230048 on Build GRCh38
rs546547630 Chr.16: 67230160-67230160 on Build GRCh38
rs199940256 Chr.16: 67229759-67229759 on Build GRCh38
rs1049123335 Chr.16: 67230161-67230161 on Build GRCh38
rs142647972 Chr.16: 67230123-67230123 on Build GRCh38
rs781092903 Chr.16: 67229903-67229903 on Build GRCh38
rs761208251 Chr.16: 67229854-67229854 on Build GRCh38
rs375544264 Chr.16: 67229982-67229982 on Build GRCh38
rs760527651 Chr.16: 67230091-67230091 on Build GRCh38
rs758366092 Chr.16: 67229808-67229808 on Build GRCh38
rs771134713 Chr.16: 67230040-67230040 on Build GRCh38
rs1027949280 Chr.16: 67230084-67230084 on Build GRCh38
rs780041932 Chr.16: 67230017-67230017 on Build GRCh38
rs1052999923 Chr.16: 67229807-67229807 on Build GRCh38
rs757132766 Chr.16: 67229882-67229882 on Build GRCh38
rs750289724 Chr.16: 67230005-67230005 on Build GRCh38
rs1018830041 Chr.16: 67230086-67230086 on Build GRCh38
rs202229073 Chr.16: 67229925-67229925 on Build GRCh38
rs756124197 Chr.16: 67229924-67229924 on Build GRCh38
rs775759444 Chr.16: 67230075-67230075 on Build GRCh38
rs771765804 Chr.16: 67230031-67230031 on Build GRCh38
rs759693508 Chr.16: 67229986-67229986 on Build GRCh38
rs779259038 Chr.16: 67229784-67229784 on Build GRCh38
rs551708643 Chr.16: 67229843-67229843 on Build GRCh38
rs908962749 Chr.16: 67230167-67230167 on Build GRCh38
rs949948364 Chr.16: 67230139-67230139 on Build GRCh38
rs941911219 Chr.16: 67229832-67229832 on Build GRCh38
rs754502928 Chr.16: 67230026-67230026 on Build GRCh38
rs749120308 Chr.16: 67230018-67230018 on Build GRCh38
rs577832197 Chr.16: 67230159-67230159 on Build GRCh38
rs772049359 Chr.16: 67230007-67230007 on Build GRCh38
rs760930009 Chr.16: 67229968-67229968 on Build GRCh38
rs773986113 Chr.16: 67229818-67229818 on Build GRCh38

View Full Product Details