Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67230444-67230943 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FHOD1
Entrez Gene ID
29109
Gene Name
formin homology 2 domain containing 1
Gene Aliases
FHOS
UniGene
Hs.95231
Gene Chromosome Location
Chr.16: 67229389-67247522 on Build GRCh38
Transcripts
NM_001318202.1 NM_013241.2 XM_006721180.1 XM_011523043.2 XM_011523044.1 XM_011523045.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1242281 c.2718C>T p.S906S Chr.16: 67230741-67230741 on Build GRCh38
6340294 c.2668-9C>A p.? Chr.16: 67230800-67230800 on Build GRCh38
3421088 c.2686A>G p.T896A Chr.16: 67230773-67230773 on Build GRCh38
3510968 c.2832G>A p.R944R Chr.16: 67230627-67230627 on Build GRCh38
6663961 c.2711G>A p.R904H Chr.16: 67230748-67230748 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761688398 Chr.16: 67230740-67230740 on Build GRCh38
rs752798062 Chr.16: 67230739-67230739 on Build GRCh38
rs759695675 Chr.16: 67230805-67230805 on Build GRCh38
rs555307904 Chr.16: 67230571-67230571 on Build GRCh38
rs143345712 Chr.16: 67230714-67230714 on Build GRCh38
rs756999242 Chr.16: 67230672-67230672 on Build GRCh38
rs779953131 Chr.16: 67230749-67230749 on Build GRCh38
rs137860084 Chr.16: 67230603-67230603 on Build GRCh38
rs758203925 Chr.16: 67230528-67230528 on Build GRCh38
rs756404268 Chr.16: 67230519-67230519 on Build GRCh38
rs776903658 Chr.16: 67230731-67230731 on Build GRCh38
rs143903337 Chr.16: 67230643-67230643 on Build GRCh38
rs754508833 Chr.16: 67230834-67230834 on Build GRCh38
rs771534266 Chr.16: 67230729-67230729 on Build GRCh38
rs760026274 Chr.16: 67230548-67230548 on Build GRCh38
rs113153021 Chr.16: 67230526-67230526 on Build GRCh38
rs767884897 Chr.16: 67230573-67230573 on Build GRCh38
rs976040730 Chr.16: 67230769-67230769 on Build GRCh38
rs748769560 Chr.16: 67230697-67230697 on Build GRCh38
rs748898328 Chr.16: 67230614-67230614 on Build GRCh38
rs752942969 Chr.16: 67230643-67230645 on Build GRCh38
rs770378710 Chr.16: 67230533-67230533 on Build GRCh38
rs982901713 Chr.16: 67230868-67230868 on Build GRCh38
rs372303815 Chr.16: 67230590-67230590 on Build GRCh38
rs375829375 Chr.16: 67230747-67230747 on Build GRCh38
rs768265790 Chr.16: 67230622-67230622 on Build GRCh38
rs752003675 Chr.16: 67230512-67230512 on Build GRCh38
rs779578738 Chr.16: 67230696-67230696 on Build GRCh38
rs373561996 Chr.16: 67230470-67230470 on Build GRCh38
rs541072158 Chr.16: 67230595-67230595 on Build GRCh38
rs1054133982 Chr.16: 67230710-67230710 on Build GRCh38
rs760835500 Chr.16: 67230690-67230690 on Build GRCh38
rs758950896 Chr.16: 67230908-67230908 on Build GRCh38
rs190207170 Chr.16: 67230465-67230465 on Build GRCh38
rs760457639 Chr.16: 67230828-67230828 on Build GRCh38
rs891440219 Chr.16: 67230711-67230711 on Build GRCh38
rs767996597 Chr.16: 67230574-67230575 on Build GRCh38
rs753156211 Chr.16: 67230493-67230493 on Build GRCh38
rs6499117 Chr.16: 67230726-67230726 on Build GRCh38
rs147506405 Chr.16: 67230718-67230718 on Build GRCh38
rs777277340 Chr.16: 67230801-67230801 on Build GRCh38
rs573563532 Chr.16: 67230579-67230579 on Build GRCh38
rs754148641 Chr.16: 67230646-67230646 on Build GRCh38
rs781200583 Chr.16: 67230676-67230676 on Build GRCh38
rs976114086 Chr.16: 67230626-67230626 on Build GRCh38
rs750277750 Chr.16: 67230682-67230682 on Build GRCh38
rs7206105 Chr.16: 67230552-67230552 on Build GRCh38
rs962042374 Chr.16: 67230755-67230755 on Build GRCh38
rs768133756 Chr.16: 67230633-67230633 on Build GRCh38
rs370200167 Chr.16: 67230732-67230732 on Build GRCh38
rs764614774 Chr.16: 67230841-67230841 on Build GRCh38
rs750435091 Chr.16: 67230518-67230518 on Build GRCh38
rs775809499 Chr.16: 67230810-67230810 on Build GRCh38
rs764350944 Chr.16: 67230825-67230825 on Build GRCh38
rs754058792 Chr.16: 67230589-67230589 on Build GRCh38
rs757268521 Chr.16: 67230597-67230597 on Build GRCh38
rs201464817 Chr.16: 67230515-67230515 on Build GRCh38
rs200719864 Chr.16: 67230570-67230570 on Build GRCh38
rs140269091 Chr.16: 67230471-67230471 on Build GRCh38
rs561524743 Chr.16: 67230811-67230811 on Build GRCh38
rs778529897 Chr.16: 67230708-67230708 on Build GRCh38
rs200564211 Chr.16: 67230500-67230500 on Build GRCh38
rs933968341 Chr.16: 67230866-67230866 on Build GRCh38
rs975830537 Chr.16: 67230850-67230850 on Build GRCh38
rs777870268 Chr.16: 67230530-67230530 on Build GRCh38
rs774055683 Chr.16: 67230624-67230624 on Build GRCh38
rs112730433 Chr.16: 67230745-67230745 on Build GRCh38
rs941514075 Chr.16: 67230895-67230895 on Build GRCh38
rs766630962 Chr.16: 67230582-67230582 on Build GRCh38
rs1045418775 Chr.16: 67230888-67230888 on Build GRCh38
rs769550151 Chr.16: 67230543-67230543 on Build GRCh38
rs935221389 Chr.16: 67230476-67230476 on Build GRCh38
rs867239418 Chr.16: 67230831-67230831 on Build GRCh38
rs907847751 Chr.16: 67230616-67230616 on Build GRCh38
rs139347380 Chr.16: 67230686-67230686 on Build GRCh38
rs998356588 Chr.16: 67230561-67230561 on Build GRCh38
rs758294040 Chr.16: 67230591-67230591 on Build GRCh38
rs769943196 Chr.16: 67230809-67230809 on Build GRCh38
rs922602952 Chr.16: 67230853-67230853 on Build GRCh38
rs763319950 Chr.16: 67230556-67230556 on Build GRCh38
rs769125781 Chr.16: 67230613-67230613 on Build GRCh38
rs750570717 Chr.16: 67230578-67230578 on Build GRCh38
rs764392630 Chr.16: 67230567-67230567 on Build GRCh38
rs553201423 Chr.16: 67230479-67230479 on Build GRCh38
rs765662304 Chr.16: 67230734-67230734 on Build GRCh38
rs183151381 Chr.16: 67230833-67230833 on Build GRCh38
rs201273155 Chr.16: 67230531-67230531 on Build GRCh38
rs745452115 Chr.16: 67230606-67230606 on Build GRCh38
rs750103540 Chr.16: 67230719-67230719 on Build GRCh38
rs764765882 Chr.16: 67230481-67230481 on Build GRCh38
rs745349213 Chr.16: 67230541-67230541 on Build GRCh38
rs201732128 Chr.16: 67230647-67230647 on Build GRCh38
rs751430927 Chr.16: 67230666-67230666 on Build GRCh38
rs766521087 Chr.16: 67230644-67230644 on Build GRCh38
rs776246617 Chr.16: 67230538-67230538 on Build GRCh38
rs747381605 Chr.16: 67230788-67230788 on Build GRCh38
rs892706225 Chr.16: 67230851-67230851 on Build GRCh38
rs773281297 Chr.16: 67230466-67230466 on Build GRCh38
rs376886788 Chr.16: 67230596-67230596 on Build GRCh38
rs1030769147 Chr.16: 67230656-67230656 on Build GRCh38
rs199955926 Chr.16: 67230800-67230800 on Build GRCh38
rs753412615 Chr.16: 67230752-67230752 on Build GRCh38
rs778039880 Chr.16: 67230775-67230775 on Build GRCh38
rs563505509 Chr.16: 67230663-67230663 on Build GRCh38
rs780123195 Chr.16: 67230524-67230524 on Build GRCh38
rs975398430 Chr.16: 67230702-67230702 on Build GRCh38
rs755804447 Chr.16: 67230820-67230821 on Build GRCh38
rs1014894880 Chr.16: 67230750-67230750 on Build GRCh38
rs932053764 Chr.16: 67230791-67230791 on Build GRCh38
rs752220377 Chr.16: 67230842-67230842 on Build GRCh38
rs111846078 Chr.16: 67230627-67230627 on Build GRCh38
rs755679813 Chr.16: 67230683-67230683 on Build GRCh38
rs781291525 Chr.16: 67230517-67230517 on Build GRCh38
rs773523134 Chr.16: 67230634-67230634 on Build GRCh38
rs375713096 Chr.16: 67230746-67230746 on Build GRCh38
rs764586981 Chr.16: 67230507-67230507 on Build GRCh38
rs150643297 Chr.16: 67230768-67230768 on Build GRCh38
rs1039067687 Chr.16: 67230744-67230744 on Build GRCh38
rs766454816 Chr.16: 67230480-67230480 on Build GRCh38
rs1005832785 Chr.16: 67230593-67230593 on Build GRCh38
rs34590217 Chr.16: 67230707-67230707 on Build GRCh38
rs374066939 Chr.16: 67230826-67230826 on Build GRCh38
rs754416899 Chr.16: 67230754-67230754 on Build GRCh38
rs777801420 Chr.16: 67230914-67230914 on Build GRCh38
rs906991167 Chr.16: 67230896-67230896 on Build GRCh38
rs767173203 Chr.16: 67230741-67230741 on Build GRCh38
rs547019932 Chr.16: 67230761-67230761 on Build GRCh38
rs981924024 Chr.16: 67230653-67230653 on Build GRCh38
rs201674604 Chr.16: 67230464-67230464 on Build GRCh38
rs945723969 Chr.16: 67230852-67230852 on Build GRCh38
rs879482441 Chr.16: 67230865-67230865 on Build GRCh38

View Full Product Details