Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67482486-67482980 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGRP
Entrez Gene ID
181
Gene Name
agouti related neuropeptide
Gene Aliases
AGRT, ART, ASIP2
UniGene
Hs.104633
Gene Chromosome Location
Chr.16: 67482571-67483813 on Build GRCh38
Transcripts
NM_001138.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6596475 c.254G>A p.R85H Chr.16: 67482781-67482781 on Build GRCh38
4484350 c.279C>T p.S93S Chr.16: 67482756-67482756 on Build GRCh38
7134007 c.348C>G p.F116L Chr.16: 67482687-67482687 on Build GRCh38
4579161 c.261C>T p.C87C Chr.16: 67482774-67482774 on Build GRCh38
6298205 c.358C>A p.R120S Chr.16: 67482677-67482677 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758156210 Chr.16: 67482835-67482835 on Build GRCh38
rs370825180 Chr.16: 67482829-67482829 on Build GRCh38
rs577037909 Chr.16: 67482572-67482572 on Build GRCh38
rs987185746 Chr.16: 67482791-67482791 on Build GRCh38
rs750837166 Chr.16: 67482658-67482658 on Build GRCh38
rs374535153 Chr.16: 67482515-67482515 on Build GRCh38
rs544523254 Chr.16: 67482906-67482906 on Build GRCh38
rs766377301 Chr.16: 67482516-67482516 on Build GRCh38
rs757155892 Chr.16: 67482630-67482630 on Build GRCh38
rs768146316 Chr.16: 67482749-67482749 on Build GRCh38
rs556803694 Chr.16: 67482930-67482930 on Build GRCh38
rs376534329 Chr.16: 67482799-67482799 on Build GRCh38
rs778045451 Chr.16: 67482838-67482838 on Build GRCh38
rs1012745439 Chr.16: 67482587-67482587 on Build GRCh38
rs563040861 Chr.16: 67482597-67482597 on Build GRCh38
rs544461250 Chr.16: 67482582-67482582 on Build GRCh38
rs150939676 Chr.16: 67482659-67482659 on Build GRCh38
rs199927717 Chr.16: 67482703-67482703 on Build GRCh38
rs964545025 Chr.16: 67482779-67482779 on Build GRCh38
rs778591661 Chr.16: 67482799-67482807 on Build GRCh38
rs777638183 Chr.16: 67482625-67482625 on Build GRCh38
rs935945327 Chr.16: 67482860-67482860 on Build GRCh38
rs763687403 Chr.16: 67482612-67482624 on Build GRCh38
rs756061493 Chr.16: 67482800-67482800 on Build GRCh38
rs750456078 Chr.16: 67482607-67482607 on Build GRCh38
rs200972106 Chr.16: 67482700-67482700 on Build GRCh38
rs748708394 Chr.16: 67482803-67482803 on Build GRCh38
rs765334298 Chr.16: 67482782-67482782 on Build GRCh38
rs748983615 Chr.16: 67482737-67482737 on Build GRCh38
rs1045776378 Chr.16: 67482899-67482899 on Build GRCh38
rs763723672 Chr.16: 67482653-67482653 on Build GRCh38
rs1015192084 Chr.16: 67482952-67482952 on Build GRCh38
rs911126169 Chr.16: 67482836-67482836 on Build GRCh38
rs755684740 Chr.16: 67482715-67482715 on Build GRCh38
rs913040575 Chr.16: 67482868-67482868 on Build GRCh38
rs145247710 Chr.16: 67482798-67482798 on Build GRCh38
rs111598170 Chr.16: 67482891-67482891 on Build GRCh38
rs752398073 Chr.16: 67482618-67482618 on Build GRCh38
rs899002440 Chr.16: 67482563-67482563 on Build GRCh38
rs373967500 Chr.16: 67482867-67482867 on Build GRCh38
rs748603132 Chr.16: 67482827-67482827 on Build GRCh38
rs754701071 Chr.16: 67482821-67482821 on Build GRCh38
rs769073853 Chr.16: 67482677-67482677 on Build GRCh38
rs749741094 Chr.16: 67482676-67482676 on Build GRCh38
rs36017515 Chr.16: 67482557-67482558 on Build GRCh38
rs776630122 Chr.16: 67482774-67482774 on Build GRCh38
rs1006450278 Chr.16: 67482933-67482933 on Build GRCh38
rs775270930 Chr.16: 67482675-67482675 on Build GRCh38
rs907321201 Chr.16: 67482907-67482907 on Build GRCh38
rs533994002 Chr.16: 67482797-67482797 on Build GRCh38
rs767874407 Chr.16: 67482684-67482684 on Build GRCh38
rs1035022181 Chr.16: 67482512-67482512 on Build GRCh38
rs773694020 Chr.16: 67482769-67482769 on Build GRCh38
rs773319622 Chr.16: 67482691-67482691 on Build GRCh38
rs780217653 Chr.16: 67482614-67482614 on Build GRCh38
rs142497424 Chr.16: 67482645-67482645 on Build GRCh38
rs753789214 Chr.16: 67482623-67482624 on Build GRCh38
rs140670389 Chr.16: 67482750-67482750 on Build GRCh38
rs191817654 Chr.16: 67482534-67482534 on Build GRCh38
rs922262842 Chr.16: 67482617-67482617 on Build GRCh38
rs745432077 Chr.16: 67482714-67482714 on Build GRCh38
rs933656758 Chr.16: 67482626-67482626 on Build GRCh38
rs781256491 Chr.16: 67482710-67482710 on Build GRCh38
rs529193631 Chr.16: 67482541-67482541 on Build GRCh38
rs569103942 Chr.16: 67482759-67482759 on Build GRCh38
rs759106876 Chr.16: 67482590-67482590 on Build GRCh38
rs975628688 Chr.16: 67482781-67482781 on Build GRCh38
rs1012110755 Chr.16: 67482704-67482704 on Build GRCh38
rs34730014 Chr.16: 67482702-67482702 on Build GRCh38
rs866590433 Chr.16: 67482544-67482544 on Build GRCh38
rs748432517 Chr.16: 67482756-67482756 on Build GRCh38
rs746966710 Chr.16: 67482627-67482627 on Build GRCh38
rs747524452 Chr.16: 67482841-67482841 on Build GRCh38
rs906371146 Chr.16: 67482953-67482953 on Build GRCh38
rs745327861 Chr.16: 67482657-67482657 on Build GRCh38
rs201485380 Chr.16: 67482644-67482644 on Build GRCh38
rs201283824 Chr.16: 67482801-67482801 on Build GRCh38
rs145840661 Chr.16: 67482771-67482771 on Build GRCh38
rs937350841 Chr.16: 67482920-67482920 on Build GRCh38
rs757247762 Chr.16: 67482705-67482705 on Build GRCh38
rs376792035 Chr.16: 67482591-67482591 on Build GRCh38
rs956734327 Chr.16: 67482762-67482762 on Build GRCh38
rs778405821 Chr.16: 67482825-67482825 on Build GRCh38
rs1056222717 Chr.16: 67482921-67482921 on Build GRCh38
rs1044892913 Chr.16: 67482943-67482943 on Build GRCh38
rs764592588 Chr.16: 67482610-67482610 on Build GRCh38
rs554693954 Chr.16: 67482773-67482773 on Build GRCh38
rs139948841 Chr.16: 67482678-67482678 on Build GRCh38
rs766645961 Chr.16: 67482696-67482696 on Build GRCh38
rs199826914 Chr.16: 67482693-67482693 on Build GRCh38
rs922789896 Chr.16: 67482786-67482786 on Build GRCh38
rs771375426 Chr.16: 67482863-67482863 on Build GRCh38
rs772682095 Chr.16: 67482763-67482763 on Build GRCh38
rs759822430 Chr.16: 67482778-67482778 on Build GRCh38
rs779648976 Chr.16: 67482735-67482735 on Build GRCh38
rs181700332 Chr.16: 67482790-67482790 on Build GRCh38
rs758198920 Chr.16: 67482621-67482621 on Build GRCh38
rs780644002 Chr.16: 67482765-67482765 on Build GRCh38
rs750189420 Chr.16: 67482810-67482816 on Build GRCh38
rs762405425 Chr.16: 67482683-67482683 on Build GRCh38

View Full Product Details