Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 8109084-8109582 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALOXE3
Entrez Gene ID
59344
Gene Name
arachidonate lipoxygenase 3
Gene Aliases
ARCI3, E-LOX, eLOX-3, eLOX3
UniGene
Hs.232770
Gene Chromosome Location
Chr.17: 8095900-8118916 on Build GRCh38
Transcripts
NM_001165960.1 NM_021628.2 XM_017024921.1 XM_017024922.1 XM_017024923.1 XM_017024924.1 XM_017024925.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1679543 c.1929C>A p.D643E Chr.17: 8109203-8109203 on Build GRCh38
6390451 c.1789G>T p.V597F Chr.17: 8109343-8109343 on Build GRCh38
5387695 c.1905C>T p.I635I Chr.17: 8109227-8109227 on Build GRCh38
437824 c.1451delT p.F484fs*30 Chr.17: 8109285-8109285 on Build GRCh38
179991 c.1530C>T p.D510D Chr.17: 8109206-8109206 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757289063 Chr.17: 8109170-8109170 on Build GRCh38
rs115159829 Chr.17: 8109158-8109158 on Build GRCh38
rs775085045 Chr.17: 8109123-8109123 on Build GRCh38
rs73241786 Chr.17: 8109141-8109141 on Build GRCh38
rs145695120 Chr.17: 8109297-8109297 on Build GRCh38
rs1041398207 Chr.17: 8109485-8109485 on Build GRCh38
rs929566867 Chr.17: 8109514-8109514 on Build GRCh38
rs766466825 Chr.17: 8109222-8109222 on Build GRCh38
rs772097833 Chr.17: 8109370-8109370 on Build GRCh38
rs148978476 Chr.17: 8109273-8109273 on Build GRCh38
rs745763413 Chr.17: 8109368-8109368 on Build GRCh38
rs368428897 Chr.17: 8109229-8109229 on Build GRCh38
rs372053788 Chr.17: 8109300-8109300 on Build GRCh38
rs769443073 Chr.17: 8109189-8109189 on Build GRCh38
rs3027205 Chr.17: 8109193-8109193 on Build GRCh38
rs762784387 Chr.17: 8109154-8109154 on Build GRCh38
rs767246830 Chr.17: 8109299-8109299 on Build GRCh38
rs939369437 Chr.17: 8109544-8109544 on Build GRCh38
rs753946891 Chr.17: 8109167-8109167 on Build GRCh38
rs755813396 Chr.17: 8109339-8109339 on Build GRCh38
rs374867848 Chr.17: 8109203-8109203 on Build GRCh38
rs977098204 Chr.17: 8109163-8109163 on Build GRCh38
rs184874325 Chr.17: 8109545-8109545 on Build GRCh38
rs954361172 Chr.17: 8109432-8109432 on Build GRCh38
rs777633445 Chr.17: 8109354-8109354 on Build GRCh38
rs765379087 Chr.17: 8109377-8109377 on Build GRCh38
rs747784080 Chr.17: 8109261-8109261 on Build GRCh38
rs193042111 Chr.17: 8109484-8109484 on Build GRCh38
rs968653406 Chr.17: 8109412-8109412 on Build GRCh38
rs747336341 Chr.17: 8109245-8109245 on Build GRCh38
rs373920814 Chr.17: 8109493-8109493 on Build GRCh38
rs551298982 Chr.17: 8109115-8109115 on Build GRCh38
rs376963513 Chr.17: 8109159-8109159 on Build GRCh38
rs772495163 Chr.17: 8109480-8109480 on Build GRCh38
rs892692954 Chr.17: 8109476-8109476 on Build GRCh38
rs910315104 Chr.17: 8109465-8109465 on Build GRCh38
rs762975670 Chr.17: 8109390-8109390 on Build GRCh38
rs1010335929 Chr.17: 8109246-8109246 on Build GRCh38
rs768920939 Chr.17: 8109247-8109247 on Build GRCh38
rs780844389 Chr.17: 8109260-8109260 on Build GRCh38
rs905397929 Chr.17: 8109224-8109224 on Build GRCh38
rs552634459 Chr.17: 8109239-8109239 on Build GRCh38
rs760291682 Chr.17: 8109134-8109134 on Build GRCh38
rs965775435 Chr.17: 8109255-8109255 on Build GRCh38
rs1003544498 Chr.17: 8109303-8109303 on Build GRCh38
rs758895916 Chr.17: 8109242-8109242 on Build GRCh38
rs121434232 Chr.17: 8109238-8109238 on Build GRCh38
rs781376374 Chr.17: 8109332-8109332 on Build GRCh38
rs758837775 Chr.17: 8109159-8109159 on Build GRCh38
rs145838099 Chr.17: 8109233-8109233 on Build GRCh38
rs940639657 Chr.17: 8109337-8109337 on Build GRCh38
rs776788351 Chr.17: 8109391-8109392 on Build GRCh38
rs750553644 Chr.17: 8109241-8109241 on Build GRCh38
rs985113517 Chr.17: 8109169-8109169 on Build GRCh38
rs552323537 Chr.17: 8109170-8109171 on Build GRCh38
rs746406430 Chr.17: 8109180-8109180 on Build GRCh38
rs371186338 Chr.17: 8109371-8109371 on Build GRCh38
rs557069937 Chr.17: 8109470-8109470 on Build GRCh38
rs576402170 Chr.17: 8109221-8109221 on Build GRCh38
rs772822868 Chr.17: 8109274-8109274 on Build GRCh38
rs776193858 Chr.17: 8109137-8109137 on Build GRCh38
rs767373651 Chr.17: 8109243-8109243 on Build GRCh38
rs751385208 Chr.17: 8109226-8109226 on Build GRCh38
rs768352670 Chr.17: 8109202-8109202 on Build GRCh38
rs138290721 Chr.17: 8109321-8109321 on Build GRCh38
rs987182221 Chr.17: 8109435-8109435 on Build GRCh38
rs754701941 Chr.17: 8109211-8109211 on Build GRCh38
rs867780319 Chr.17: 8109250-8109250 on Build GRCh38
rs769303884 Chr.17: 8109213-8109213 on Build GRCh38
rs927935449 Chr.17: 8109534-8109534 on Build GRCh38
rs541619175 Chr.17: 8109301-8109301 on Build GRCh38
rs766396693 Chr.17: 8109395-8109395 on Build GRCh38
rs1029171531 Chr.17: 8109527-8109527 on Build GRCh38
rs905326805 Chr.17: 8109426-8109426 on Build GRCh38
rs774305049 Chr.17: 8109363-8109363 on Build GRCh38
rs746832227 Chr.17: 8109106-8109106 on Build GRCh38
rs765159284 Chr.17: 8109144-8109144 on Build GRCh38
rs973577499 Chr.17: 8109489-8109489 on Build GRCh38
rs536043671 Chr.17: 8109114-8109114 on Build GRCh38
rs368452121 Chr.17: 8109278-8109278 on Build GRCh38
rs1045200799 Chr.17: 8109216-8109216 on Build GRCh38
rs755325552 Chr.17: 8109327-8109327 on Build GRCh38
rs770577900 Chr.17: 8109357-8109357 on Build GRCh38
rs762696128 Chr.17: 8109108-8109108 on Build GRCh38
rs998808467 Chr.17: 8109424-8109424 on Build GRCh38
rs953212312 Chr.17: 8109540-8109540 on Build GRCh38
rs748592926 Chr.17: 8109334-8109334 on Build GRCh38
rs759316171 Chr.17: 8109227-8109227 on Build GRCh38
rs370462522 Chr.17: 8109194-8109194 on Build GRCh38
rs772608978 Chr.17: 8109181-8109181 on Build GRCh38
rs754029651 Chr.17: 8109232-8109232 on Build GRCh38
rs953341224 Chr.17: 8109275-8109275 on Build GRCh38
rs370665382 Chr.17: 8109155-8109155 on Build GRCh38
rs761332889 Chr.17: 8109205-8109205 on Build GRCh38
rs1031340740 Chr.17: 8109425-8109425 on Build GRCh38
rs368485942 Chr.17: 8109376-8109376 on Build GRCh38
rs1045129416 Chr.17: 8109414-8109414 on Build GRCh38
rs141735745 Chr.17: 8109220-8109221 on Build GRCh38
rs770986562 Chr.17: 8109293-8109293 on Build GRCh38
rs780702320 Chr.17: 8109161-8109162 on Build GRCh38
rs746198347 Chr.17: 8109148-8109148 on Build GRCh38
rs1006446070 Chr.17: 8109546-8109546 on Build GRCh38
rs1054018314 Chr.17: 8109464-8109464 on Build GRCh38
rs987255862 Chr.17: 8109225-8109225 on Build GRCh38
rs376787640 Chr.17: 8109151-8109151 on Build GRCh38
rs1047969445 Chr.17: 8109515-8109515 on Build GRCh38
rs748739747 Chr.17: 8109355-8109355 on Build GRCh38
rs375080695 Chr.17: 8109244-8109244 on Build GRCh38
rs376653779 Chr.17: 8109379-8109379 on Build GRCh38
rs368012174 Chr.17: 8109304-8109304 on Build GRCh38
rs773240392 Chr.17: 8109215-8109215 on Build GRCh38
rs758480121 Chr.17: 8109306-8109306 on Build GRCh38

View Full Product Details