Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 35575920-35576419 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PEX12
Entrez Gene ID
5193
Gene Name
peroxisomal biogenesis factor 12
Gene Aliases
PAF-3, PBD3A
UniGene
Hs.591190
Gene Chromosome Location
Chr.17: 35574795-35578637 on Build GRCh38
Transcripts
NM_000286.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1382512 c.738C>A p.S246S Chr.17: 35576124-35576124 on Build GRCh38
1382511 c.822G>T p.K274N Chr.17: 35576040-35576040 on Build GRCh38
5725328 c.885C>T p.P295P Chr.17: 35575977-35575977 on Build GRCh38
6872224 c.861C>A p.H287Q Chr.17: 35576001-35576001 on Build GRCh38
1302669 c.768C>G p.F256L Chr.17: 35576094-35576094 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs61752111 Chr.17: 35575975-35575975 on Build GRCh38
rs980992759 Chr.17: 35576144-35576144 on Build GRCh38
rs767070181 Chr.17: 35575978-35575978 on Build GRCh38
rs757306012 Chr.17: 35576081-35576081 on Build GRCh38
rs781455518 Chr.17: 35576090-35576090 on Build GRCh38
rs61752108 Chr.17: 35576117-35576118 on Build GRCh38
rs398123301 Chr.17: 35575973-35575974 on Build GRCh38
rs754193088 Chr.17: 35576081-35576081 on Build GRCh38
rs757399505 Chr.17: 35576215-35576215 on Build GRCh38
rs146527335 Chr.17: 35576211-35576211 on Build GRCh38
rs142726996 Chr.17: 35576069-35576069 on Build GRCh38
rs764241263 Chr.17: 35576219-35576219 on Build GRCh38
rs1030736075 Chr.17: 35576123-35576123 on Build GRCh38
rs761590834 Chr.17: 35575976-35575976 on Build GRCh38
rs531962872 Chr.17: 35576265-35576266 on Build GRCh38
rs201442874 Chr.17: 35576052-35576052 on Build GRCh38
rs928221608 Chr.17: 35576169-35576169 on Build GRCh38
rs138568975 Chr.17: 35576183-35576184 on Build GRCh38
rs867245161 Chr.17: 35575951-35575952 on Build GRCh38
rs772194164 Chr.17: 35575950-35575950 on Build GRCh38
rs940043493 Chr.17: 35576241-35576241 on Build GRCh38
rs765982515 Chr.17: 35576138-35576138 on Build GRCh38
rs202159096 Chr.17: 35576033-35576033 on Build GRCh38
rs1057519507 Chr.17: 35576095-35576096 on Build GRCh38
rs190481912 Chr.17: 35576257-35576257 on Build GRCh38
rs768447498 Chr.17: 35576102-35576102 on Build GRCh38
rs768427035 Chr.17: 35575972-35575972 on Build GRCh38
rs749859287 Chr.17: 35575990-35575990 on Build GRCh38
rs569741823 Chr.17: 35576051-35576051 on Build GRCh38
rs145574816 Chr.17: 35575959-35575959 on Build GRCh38
rs932673242 Chr.17: 35576267-35576267 on Build GRCh38
rs748595918 Chr.17: 35575947-35575947 on Build GRCh38
rs573624630 Chr.17: 35576106-35576106 on Build GRCh38
rs753527027 Chr.17: 35576015-35576015 on Build GRCh38
rs968180697 Chr.17: 35576174-35576174 on Build GRCh38
rs371277434 Chr.17: 35575956-35575956 on Build GRCh38
rs62642859 Chr.17: 35576175-35576178 on Build GRCh38
rs765654384 Chr.17: 35576158-35576158 on Build GRCh38
rs974191182 Chr.17: 35576239-35576239 on Build GRCh38
rs533141936 Chr.17: 35576246-35576246 on Build GRCh38
rs781186474 Chr.17: 35576228-35576228 on Build GRCh38
rs776427149 Chr.17: 35576139-35576139 on Build GRCh38
rs796327178 Chr.17: 35576306-35576306 on Build GRCh38
rs200413804 Chr.17: 35576125-35576125 on Build GRCh38
rs755457602 Chr.17: 35576028-35576028 on Build GRCh38
rs104894616 Chr.17: 35576171-35576171 on Build GRCh38
rs777654572 Chr.17: 35575955-35575955 on Build GRCh38
rs1021171631 Chr.17: 35576160-35576160 on Build GRCh38
rs373860467 Chr.17: 35575957-35575957 on Build GRCh38
rs765449979 Chr.17: 35576046-35576046 on Build GRCh38
rs61752109 Chr.17: 35575986-35575987 on Build GRCh38
rs759584047 Chr.17: 35576075-35576077 on Build GRCh38
rs888969753 Chr.17: 35576301-35576301 on Build GRCh38
rs773175740 Chr.17: 35576126-35576126 on Build GRCh38
rs61752107 Chr.17: 35576128-35576129 on Build GRCh38
rs761641358 Chr.17: 35576220-35576223 on Build GRCh38
rs758784930 Chr.17: 35576066-35576066 on Build GRCh38
rs1007698011 Chr.17: 35576305-35576305 on Build GRCh38
rs746089337 Chr.17: 35576099-35576099 on Build GRCh38
rs965759382 Chr.17: 35576137-35576137 on Build GRCh38
rs139417458 Chr.17: 35576140-35576140 on Build GRCh38
rs773962549 Chr.17: 35576104-35576104 on Build GRCh38
rs371597789 Chr.17: 35576117-35576117 on Build GRCh38
rs762789314 Chr.17: 35575969-35575969 on Build GRCh38
rs1048955680 Chr.17: 35576276-35576276 on Build GRCh38
rs776806092 Chr.17: 35575963-35575963 on Build GRCh38
rs570156139 Chr.17: 35576319-35576319 on Build GRCh38
rs779100543 Chr.17: 35576220-35576221 on Build GRCh38
rs398123302 Chr.17: 35575968-35575968 on Build GRCh38
rs941482060 Chr.17: 35576237-35576237 on Build GRCh38
rs371906447 Chr.17: 35576196-35576196 on Build GRCh38
rs766182813 Chr.17: 35576006-35576006 on Build GRCh38
rs774415243 Chr.17: 35575973-35575973 on Build GRCh38
rs771406750 Chr.17: 35575945-35575946 on Build GRCh38
rs747099919 Chr.17: 35576073-35576073 on Build GRCh38
rs187526749 Chr.17: 35576183-35576183 on Build GRCh38
rs138720627 Chr.17: 35575995-35575995 on Build GRCh38
rs200463087 Chr.17: 35576213-35576213 on Build GRCh38
rs746106325 Chr.17: 35576233-35576233 on Build GRCh38
rs12941376 Chr.17: 35576129-35576129 on Build GRCh38

View Full Product Details