Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 39687788-39688297 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERBB2
Entrez Gene ID
2064
Gene Name
erb-b2 receptor tyrosine kinase 2
Gene Aliases
CD340, HER-2, HER-2/neu, HER2, MLN 19, NEU, NGL, TKR1
UniGene
Hs.446352
Gene Chromosome Location
Chr.17: 39688084-39728662 on Build GRCh38
Transcripts
NM_001005862.2 NM_001289936.1 NM_001289938.1 NM_001291726.1 NM_001291728.1 NM_001291730.1 NM_001291732.1 NM_001291733.1 NM_033419.4 XM_011525480.2 XM_011525481.2

Gene Symbol
PGAP3
Entrez Gene ID
93210
Gene Name
post-GPI attachment to proteins 3
Gene Aliases
AGLA546, CAB2, PERLD1, PP1498, hCOS16
UniGene
Hs.462971
Gene Chromosome Location
Chr.17: 39671122-39688070 on Build GRCh38
Transcripts
NM_001005862.2 NM_001289936.1 NM_001289938.1 NM_001291726.1 NM_001291728.1 NM_001291730.1 NM_001291732.1 NM_001291733.1 NM_033419.4 XM_011525480.2 XM_011525481.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4713777 c.178G>A p.A60T Chr.17: 39687837-39687837 on Build GRCh38
5657326 c.47C>T p.A16V Chr.17: 39687968-39687968 on Build GRCh38
7297482 c.59G>A p.G20D Chr.17: 39687956-39687956 on Build GRCh38
6195354 c.54G>A p.A18A Chr.17: 39687961-39687961 on Build GRCh38
116759 c.163A>G p.I55V Chr.17: 39687852-39687852 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs946530794 Chr.17: 39687947-39687947 on Build GRCh38
rs769356560 Chr.17: 39687957-39687957 on Build GRCh38
rs766556749 Chr.17: 39688023-39688023 on Build GRCh38
rs767944583 Chr.17: 39688007-39688007 on Build GRCh38
rs548386033 Chr.17: 39688237-39688237 on Build GRCh38
rs745639362 Chr.17: 39687956-39687956 on Build GRCh38
rs202046464 Chr.17: 39688019-39688019 on Build GRCh38
rs761050510 Chr.17: 39688018-39688018 on Build GRCh38
rs758798118 Chr.17: 39688077-39688077 on Build GRCh38
rs755171864 Chr.17: 39688034-39688034 on Build GRCh38
rs780280407 Chr.17: 39687934-39687934 on Build GRCh38
rs775187703 Chr.17: 39687964-39687964 on Build GRCh38
rs891877528 Chr.17: 39687948-39687948 on Build GRCh38
rs1051112174 Chr.17: 39688220-39688220 on Build GRCh38
rs777801047 Chr.17: 39687865-39687865 on Build GRCh38
rs777894640 Chr.17: 39687940-39687940 on Build GRCh38
rs372203566 Chr.17: 39688001-39688001 on Build GRCh38
rs1017397801 Chr.17: 39688030-39688030 on Build GRCh38
rs1041059830 Chr.17: 39687817-39687817 on Build GRCh38
rs780010071 Chr.17: 39687860-39687860 on Build GRCh38
rs975996306 Chr.17: 39688149-39688149 on Build GRCh38
rs773564341 Chr.17: 39688060-39688060 on Build GRCh38
rs200697953 Chr.17: 39687880-39687880 on Build GRCh38
rs770939421 Chr.17: 39687945-39687945 on Build GRCh38
rs564810973 Chr.17: 39688027-39688027 on Build GRCh38
rs949013206 Chr.17: 39688121-39688121 on Build GRCh38
rs769425361 Chr.17: 39687883-39687883 on Build GRCh38
rs762320296 Chr.17: 39688005-39688005 on Build GRCh38
rs150801318 Chr.17: 39687891-39687891 on Build GRCh38
rs944660856 Chr.17: 39687963-39687964 on Build GRCh38
rs998225576 Chr.17: 39687982-39687982 on Build GRCh38
rs952827079 Chr.17: 39687882-39687882 on Build GRCh38
rs944603192 Chr.17: 39688146-39688146 on Build GRCh38
rs954666246 Chr.17: 39687988-39687988 on Build GRCh38
rs750606431 Chr.17: 39687848-39687848 on Build GRCh38
rs777149964 Chr.17: 39688022-39688022 on Build GRCh38
rs368152790 Chr.17: 39687925-39687925 on Build GRCh38
rs897406255 Chr.17: 39688097-39688097 on Build GRCh38
rs750385176 Chr.17: 39687819-39687819 on Build GRCh38
rs1005687770 Chr.17: 39688074-39688074 on Build GRCh38
rs891908735 Chr.17: 39688197-39688197 on Build GRCh38
rs563212698 Chr.17: 39688101-39688101 on Build GRCh38
rs541182612 Chr.17: 39688106-39688106 on Build GRCh38
rs967780100 Chr.17: 39688123-39688123 on Build GRCh38
rs752032433 Chr.17: 39687901-39687901 on Build GRCh38
rs765928877 Chr.17: 39687971-39687972 on Build GRCh38
rs535959708 Chr.17: 39687886-39687886 on Build GRCh38
rs998651699 Chr.17: 39688216-39688216 on Build GRCh38
rs573315299 Chr.17: 39688112-39688112 on Build GRCh38
rs904663988 Chr.17: 39687924-39687924 on Build GRCh38
rs762232354 Chr.17: 39687902-39687902 on Build GRCh38
rs77251976 Chr.17: 39688122-39688122 on Build GRCh38
rs746846864 Chr.17: 39687867-39687867 on Build GRCh38
rs1034729768 Chr.17: 39687899-39687899 on Build GRCh38
rs1042258551 Chr.17: 39687812-39687812 on Build GRCh38
rs919044456 Chr.17: 39688061-39688061 on Build GRCh38
rs923815405 Chr.17: 39688134-39688134 on Build GRCh38
rs768282987 Chr.17: 39688002-39688002 on Build GRCh38
rs1030058305 Chr.17: 39687985-39687985 on Build GRCh38
rs774897725 Chr.17: 39687890-39687890 on Build GRCh38
rs367611030 Chr.17: 39687815-39687815 on Build GRCh38
rs771589861 Chr.17: 39688125-39688125 on Build GRCh38
rs770814890 Chr.17: 39687875-39687875 on Build GRCh38
rs746488673 Chr.17: 39687834-39687834 on Build GRCh38
rs948967286 Chr.17: 39687866-39687866 on Build GRCh38
rs80085819 Chr.17: 39688142-39688142 on Build GRCh38
rs200611113 Chr.17: 39688049-39688049 on Build GRCh38
rs199645746 Chr.17: 39688009-39688009 on Build GRCh38
rs535183604 Chr.17: 39687835-39687835 on Build GRCh38
rs902458903 Chr.17: 39687814-39687814 on Build GRCh38
rs554689197 Chr.17: 39687998-39687998 on Build GRCh38
rs756294252 Chr.17: 39687928-39687928 on Build GRCh38
rs781433220 Chr.17: 39687840-39687840 on Build GRCh38
rs752197704 Chr.17: 39687810-39687810 on Build GRCh38
rs56350074 Chr.17: 39688086-39688086 on Build GRCh38
rs762412326 Chr.17: 39687974-39687974 on Build GRCh38
rs888522675 Chr.17: 39688226-39688226 on Build GRCh38
rs901202816 Chr.17: 39687971-39687971 on Build GRCh38
rs916877176 Chr.17: 39688116-39688116 on Build GRCh38
rs936187754 Chr.17: 39688152-39688152 on Build GRCh38
rs774896794 Chr.17: 39688000-39688000 on Build GRCh38
rs765390691 Chr.17: 39688046-39688046 on Build GRCh38
rs756275423 Chr.17: 39687854-39687854 on Build GRCh38
rs963144673 Chr.17: 39688056-39688056 on Build GRCh38
rs1041440997 Chr.17: 39687965-39687965 on Build GRCh38
rs1053267751 Chr.17: 39688187-39688187 on Build GRCh38
rs941463394 Chr.17: 39688243-39688243 on Build GRCh38
rs35771148 Chr.17: 39688221-39688221 on Build GRCh38
rs373850048 Chr.17: 39687895-39687895 on Build GRCh38
rs959363367 Chr.17: 39687932-39687932 on Build GRCh38
rs375234528 Chr.17: 39687946-39687946 on Build GRCh38
rs553299603 Chr.17: 39687862-39687862 on Build GRCh38
rs371801150 Chr.17: 39687818-39687818 on Build GRCh38
rs762614708 Chr.17: 39687901-39687905 on Build GRCh38
rs776488270 Chr.17: 39687878-39687878 on Build GRCh38
rs769873219 Chr.17: 39688178-39688178 on Build GRCh38
rs923826853 Chr.17: 39687959-39687959 on Build GRCh38
rs768151075 Chr.17: 39687987-39687987 on Build GRCh38
rs746934856 Chr.17: 39687942-39687942 on Build GRCh38
rs753883552 Chr.17: 39687935-39687935 on Build GRCh38
rs767854862 Chr.17: 39687915-39687915 on Build GRCh38

View Full Product Details