Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 69251816-69252360 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA5
Entrez Gene ID
23461
Gene Name
ATP binding cassette subfamily A member 5
Gene Aliases
ABC13, EST90625
UniGene
Hs.731824
Gene Chromosome Location
Chr.17: 69244435-69327182 on Build GRCh38
Transcripts
NM_018672.4 NM_172232.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4828034 c.4430C>A p.T1477N Chr.17: 69251852-69251852 on Build GRCh38
259265 c.4417C>T p.R1473* Chr.17: 69251865-69251865 on Build GRCh38
4495109 c.4466C>T p.T1489I Chr.17: 69251816-69251816 on Build GRCh38
3521457 c.4442A>G p.N1481S Chr.17: 69251840-69251840 on Build GRCh38
5387157 c.4426C>T p.R1476* Chr.17: 69251856-69251856 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs535829174 Chr.17: 69252099-69252099 on Build GRCh38
rs1001356177 Chr.17: 69252323-69252323 on Build GRCh38
rs554273888 Chr.17: 69252147-69252147 on Build GRCh38
rs900414067 Chr.17: 69252083-69252083 on Build GRCh38
rs368231532 Chr.17: 69251910-69251910 on Build GRCh38
rs544003295 Chr.17: 69252305-69252305 on Build GRCh38
rs1015676218 Chr.17: 69252157-69252157 on Build GRCh38
rs35678568 Chr.17: 69252179-69252180 on Build GRCh38
rs368173901 Chr.17: 69251944-69251944 on Build GRCh38
rs71144665 Chr.17: 69252016-69252017 on Build GRCh38
rs143193092 Chr.17: 69251865-69251865 on Build GRCh38
rs776759169 Chr.17: 69251979-69251979 on Build GRCh38
rs935670435 Chr.17: 69251859-69251859 on Build GRCh38
rs1038973541 Chr.17: 69252077-69252077 on Build GRCh38
rs1050024614 Chr.17: 69251983-69251983 on Build GRCh38
rs201178598 Chr.17: 69251913-69251914 on Build GRCh38
rs964104290 Chr.17: 69251898-69251898 on Build GRCh38
rs752814217 Chr.17: 69251864-69251864 on Build GRCh38
rs1037063100 Chr.17: 69252259-69252259 on Build GRCh38
rs1023252273 Chr.17: 69252204-69252204 on Build GRCh38
rs1032885161 Chr.17: 69252329-69252329 on Build GRCh38
rs993124997 Chr.17: 69251973-69251973 on Build GRCh38
rs925597618 Chr.17: 69251846-69251846 on Build GRCh38
rs778216537 Chr.17: 69251881-69251881 on Build GRCh38
rs957251453 Chr.17: 69252135-69252135 on Build GRCh38
rs922743862 Chr.17: 69252201-69252201 on Build GRCh38
rs553754046 Chr.17: 69252075-69252075 on Build GRCh38
rs1016087142 Chr.17: 69252068-69252068 on Build GRCh38
rs925586167 Chr.17: 69252043-69252043 on Build GRCh38
rs753638194 Chr.17: 69251849-69251849 on Build GRCh38
rs925638584 Chr.17: 69252239-69252239 on Build GRCh38
rs961524215 Chr.17: 69252074-69252074 on Build GRCh38
rs913012105 Chr.17: 69251907-69251907 on Build GRCh38
rs952668886 Chr.17: 69252102-69252102 on Build GRCh38
rs925879402 Chr.17: 69252112-69252112 on Build GRCh38
rs979947845 Chr.17: 69252107-69252107 on Build GRCh38
rs549702553 Chr.17: 69252035-69252035 on Build GRCh38
rs988532191 Chr.17: 69252153-69252153 on Build GRCh38
rs1005963517 Chr.17: 69252257-69252257 on Build GRCh38
rs552382425 Chr.17: 69251906-69251909 on Build GRCh38
rs530230882 Chr.17: 69251856-69251856 on Build GRCh38
rs766585634 Chr.17: 69251943-69251943 on Build GRCh38
rs934296931 Chr.17: 69252249-69252249 on Build GRCh38
rs757113904 Chr.17: 69251850-69251850 on Build GRCh38
rs768592168 Chr.17: 69251914-69251914 on Build GRCh38
rs571529452 Chr.17: 69252042-69252042 on Build GRCh38
rs897160758 Chr.17: 69252261-69252261 on Build GRCh38
rs796240448 Chr.17: 69252024-69252024 on Build GRCh38
rs1024063272 Chr.17: 69252097-69252097 on Build GRCh38
rs6501313 Chr.17: 69252256-69252256 on Build GRCh38
rs888410122 Chr.17: 69251993-69251993 on Build GRCh38
rs373860253 Chr.17: 69251863-69251863 on Build GRCh38
rs1004314236 Chr.17: 69252152-69252152 on Build GRCh38
rs112048265 Chr.17: 69252023-69252023 on Build GRCh38
rs566757690 Chr.17: 69251950-69251953 on Build GRCh38
rs932810288 Chr.17: 69252212-69252212 on Build GRCh38
rs1041242161 Chr.17: 69251925-69251925 on Build GRCh38
rs970721139 Chr.17: 69252216-69252216 on Build GRCh38
rs538513495 Chr.17: 69252046-69252046 on Build GRCh38
rs912956361 Chr.17: 69252155-69252155 on Build GRCh38
rs1014316036 Chr.17: 69252293-69252293 on Build GRCh38
rs1024344837 Chr.17: 69252304-69252304 on Build GRCh38
rs771305133 Chr.17: 69251967-69251974 on Build GRCh38
rs570141113 Chr.17: 69251923-69251923 on Build GRCh38
rs567128552 Chr.17: 69252178-69252179 on Build GRCh38
rs763822136 Chr.17: 69251844-69251844 on Build GRCh38
rs777069533 Chr.17: 69251949-69251949 on Build GRCh38
rs12942723 Chr.17: 69252017-69252017 on Build GRCh38
rs759792867 Chr.17: 69252012-69252012 on Build GRCh38
rs750426352 Chr.17: 69251855-69251855 on Build GRCh38
rs1040161599 Chr.17: 69252198-69252198 on Build GRCh38
rs548730483 Chr.17: 69252174-69252174 on Build GRCh38
rs533878232 Chr.17: 69252113-69252113 on Build GRCh38
rs960167256 Chr.17: 69251954-69251958 on Build GRCh38
rs994731223 Chr.17: 69252088-69252088 on Build GRCh38
rs749663854 Chr.17: 69251882-69251882 on Build GRCh38
rs370948728 Chr.17: 69252273-69252273 on Build GRCh38
rs62080864 Chr.17: 69252170-69252170 on Build GRCh38
rs1049972526 Chr.17: 69252229-69252229 on Build GRCh38
rs1005560385 Chr.17: 69252055-69252055 on Build GRCh38
rs374104476 Chr.17: 69252021-69252022 on Build GRCh38

View Full Product Details