Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 69253337-69253873 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA5
Entrez Gene ID
23461
Gene Name
ATP binding cassette subfamily A member 5
Gene Aliases
ABC13, EST90625
UniGene
Hs.731824
Gene Chromosome Location
Chr.17: 69244435-69327182 on Build GRCh38
Transcripts
NM_018672.4 NM_172232.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6403049 c.4318A>G p.K1440E Chr.17: 69253796-69253796 on Build GRCh38
7297229 c.4415+10G>T p.? Chr.17: 69253563-69253563 on Build GRCh38
6246240 c.4304C>A p.A1435E Chr.17: 69253810-69253810 on Build GRCh38
983601 c.4316G>A p.R1439Q Chr.17: 69253798-69253798 on Build GRCh38
6424372 c.4390G>T p.D1464Y Chr.17: 69253598-69253598 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768296243 Chr.17: 69253545-69253545 on Build GRCh38
rs778193276 Chr.17: 69253512-69253512 on Build GRCh38
rs377065403 Chr.17: 69253750-69253750 on Build GRCh38
rs769593687 Chr.17: 69253527-69253531 on Build GRCh38
rs190726871 Chr.17: 69253557-69253557 on Build GRCh38
rs779363752 Chr.17: 69253589-69253589 on Build GRCh38
rs754445657 Chr.17: 69253746-69253746 on Build GRCh38
rs757572144 Chr.17: 69253663-69253663 on Build GRCh38
rs77262773 Chr.17: 69253570-69253570 on Build GRCh38
rs369202022 Chr.17: 69253798-69253798 on Build GRCh38
rs372070705 Chr.17: 69253790-69253790 on Build GRCh38
rs1006333117 Chr.17: 69253507-69253507 on Build GRCh38
rs552514689 Chr.17: 69253657-69253657 on Build GRCh38
rs752014337 Chr.17: 69253824-69253824 on Build GRCh38
rs746424183 Chr.17: 69253526-69253526 on Build GRCh38
rs887420788 Chr.17: 69253738-69253738 on Build GRCh38
rs761693989 Chr.17: 69253523-69253524 on Build GRCh38
rs765058027 Chr.17: 69253550-69253550 on Build GRCh38
rs776326867 Chr.17: 69253546-69253546 on Build GRCh38
rs776237183 Chr.17: 69253632-69253632 on Build GRCh38
rs897501458 Chr.17: 69253814-69253814 on Build GRCh38
rs769491093 Chr.17: 69253757-69253757 on Build GRCh38
rs766687347 Chr.17: 69253810-69253810 on Build GRCh38
rs9909726 Chr.17: 69253781-69253781 on Build GRCh38
rs759574855 Chr.17: 69253792-69253792 on Build GRCh38
rs780590826 Chr.17: 69253710-69253710 on Build GRCh38
rs760182920 Chr.17: 69253540-69253540 on Build GRCh38
rs185423871 Chr.17: 69253490-69253490 on Build GRCh38
rs907725902 Chr.17: 69253456-69253456 on Build GRCh38
rs534944925 Chr.17: 69253766-69253766 on Build GRCh38
rs773073518 Chr.17: 69253649-69253649 on Build GRCh38
rs946153180 Chr.17: 69253714-69253714 on Build GRCh38
rs917821865 Chr.17: 69253468-69253468 on Build GRCh38
rs1029196044 Chr.17: 69253380-69253380 on Build GRCh38
rs367707617 Chr.17: 69253536-69253536 on Build GRCh38
rs778126578 Chr.17: 69253579-69253579 on Build GRCh38
rs139242970 Chr.17: 69253648-69253648 on Build GRCh38
rs75169077 Chr.17: 69253458-69253458 on Build GRCh38
rs758761906 Chr.17: 69253809-69253809 on Build GRCh38
rs888749823 Chr.17: 69253459-69253459 on Build GRCh38
rs372565939 Chr.17: 69253706-69253706 on Build GRCh38
rs779538014 Chr.17: 69253522-69253522 on Build GRCh38
rs34951150 Chr.17: 69253399-69253399 on Build GRCh38
rs746512679 Chr.17: 69253607-69253607 on Build GRCh38
rs570521333 Chr.17: 69253755-69253755 on Build GRCh38
rs751509122 Chr.17: 69253655-69253655 on Build GRCh38
rs747557090 Chr.17: 69253711-69253711 on Build GRCh38
rs199753304 Chr.17: 69253793-69253793 on Build GRCh38
rs146689980 Chr.17: 69253597-69253597 on Build GRCh38
rs772229277 Chr.17: 69253651-69253651 on Build GRCh38
rs780614063 Chr.17: 69253841-69253841 on Build GRCh38
rs761575607 Chr.17: 69253547-69253547 on Build GRCh38
rs144095768 Chr.17: 69253656-69253656 on Build GRCh38
rs537035226 Chr.17: 69253685-69253685 on Build GRCh38
rs761485735 Chr.17: 69253641-69253641 on Build GRCh38
rs750662706 Chr.17: 69253803-69253803 on Build GRCh38
rs768206442 Chr.17: 69253617-69253617 on Build GRCh38
rs772926995 Chr.17: 69253686-69253686 on Build GRCh38
rs747651434 Chr.17: 69253844-69253844 on Build GRCh38
rs375563778 Chr.17: 69253580-69253580 on Build GRCh38
rs756327889 Chr.17: 69253577-69253577 on Build GRCh38
rs755489462 Chr.17: 69253834-69253834 on Build GRCh38
rs771439312 Chr.17: 69253527-69253527 on Build GRCh38
rs746307111 Chr.17: 69253590-69253590 on Build GRCh38
rs773430791 Chr.17: 69253744-69253744 on Build GRCh38
rs749094829 Chr.17: 69253779-69253779 on Build GRCh38
rs758750288 Chr.17: 69253707-69253707 on Build GRCh38
rs902086500 Chr.17: 69253684-69253684 on Build GRCh38
rs368600407 Chr.17: 69253653-69253653 on Build GRCh38
rs765457976 Chr.17: 69253676-69253676 on Build GRCh38
rs372523620 Chr.17: 69253799-69253799 on Build GRCh38
rs1044393607 Chr.17: 69253531-69253531 on Build GRCh38
rs1041819843 Chr.17: 69253725-69253725 on Build GRCh38
rs772643034 Chr.17: 69253595-69253595 on Build GRCh38
rs754865616 Chr.17: 69253396-69253396 on Build GRCh38
rs767438535 Chr.17: 69253566-69253566 on Build GRCh38
rs754069144 Chr.17: 69253658-69253658 on Build GRCh38
rs750288164 Chr.17: 69253551-69253551 on Build GRCh38

View Full Product Details