Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 49791364-49791910 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACAA2
Entrez Gene ID
10449
Gene Name
acetyl-CoA acyltransferase 2
Gene Aliases
DSAEC
UniGene
Hs.200136
Gene Chromosome Location
Chr.18: 49783504-49813881 on Build GRCh38
Transcripts
NM_006111.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6237114 c.867C>A p.P289P Chr.18: 49791486-49791486 on Build GRCh38
4396687 c.873C>T p.I291I Chr.18: 49791480-49791480 on Build GRCh38
1611264 c.871A>G p.I291V Chr.18: 49791482-49791482 on Build GRCh38
7353830 c.811C>T p.H271Y Chr.18: 49791542-49791542 on Build GRCh38
3526157 c.826C>T p.L276L Chr.18: 49791527-49791527 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140796418 Chr.18: 49791754-49791754 on Build GRCh38
rs998099308 Chr.18: 49791778-49791778 on Build GRCh38
rs748882477 Chr.18: 49791599-49791599 on Build GRCh38
rs888517118 Chr.18: 49791662-49791662 on Build GRCh38
rs78816994 Chr.18: 49791459-49791459 on Build GRCh38
rs148304029 Chr.18: 49791542-49791542 on Build GRCh38
rs901106207 Chr.18: 49791726-49791726 on Build GRCh38
rs1017146727 Chr.18: 49791488-49791488 on Build GRCh38
rs543656657 Chr.18: 49791535-49791535 on Build GRCh38
rs774333318 Chr.18: 49791475-49791475 on Build GRCh38
rs760436378 Chr.18: 49791593-49791594 on Build GRCh38
rs1047063834 Chr.18: 49791782-49791782 on Build GRCh38
rs143197316 Chr.18: 49791879-49791879 on Build GRCh38
rs143583716 Chr.18: 49791507-49791507 on Build GRCh38
rs988999225 Chr.18: 49791876-49791877 on Build GRCh38
rs767375969 Chr.18: 49791479-49791479 on Build GRCh38
rs1039482689 Chr.18: 49791723-49791723 on Build GRCh38
rs781744623 Chr.18: 49791577-49791577 on Build GRCh38
rs1013106626 Chr.18: 49791863-49791863 on Build GRCh38
rs958860309 Chr.18: 49791497-49791497 on Build GRCh38
rs771812881 Chr.18: 49791497-49791502 on Build GRCh38
rs985118781 Chr.18: 49791849-49791852 on Build GRCh38
rs972030627 Chr.18: 49791571-49791571 on Build GRCh38
rs915099367 Chr.18: 49791688-49791688 on Build GRCh38
rs201013632 Chr.18: 49791422-49791422 on Build GRCh38
rs568640198 Chr.18: 49791877-49791879 on Build GRCh38
rs781169602 Chr.18: 49791423-49791423 on Build GRCh38
rs867343333 Chr.18: 49791857-49791857 on Build GRCh38
rs868563880 Chr.18: 49791861-49791861 on Build GRCh38
rs773112984 Chr.18: 49791626-49791626 on Build GRCh38
rs953956491 Chr.18: 49791844-49791844 on Build GRCh38
rs773079803 Chr.18: 49791561-49791562 on Build GRCh38
rs747505009 Chr.18: 49791425-49791425 on Build GRCh38
rs753129783 Chr.18: 49791421-49791421 on Build GRCh38
rs753782335 Chr.18: 49791647-49791648 on Build GRCh38
rs1006800802 Chr.18: 49791404-49791404 on Build GRCh38
rs761073016 Chr.18: 49791780-49791780 on Build GRCh38
rs186288400 Chr.18: 49791866-49791866 on Build GRCh38
rs757866729 Chr.18: 49791567-49791567 on Build GRCh38
rs180793143 Chr.18: 49791630-49791630 on Build GRCh38
rs772535681 Chr.18: 49791805-49791805 on Build GRCh38
rs372382497 Chr.18: 49791462-49791462 on Build GRCh38
rs2298629 Chr.18: 49791709-49791709 on Build GRCh38
rs548562452 Chr.18: 49791770-49791770 on Build GRCh38
rs80081658 Chr.18: 49791616-49791616 on Build GRCh38
rs1002192183 Chr.18: 49791632-49791632 on Build GRCh38
rs751902237 Chr.18: 49791554-49791554 on Build GRCh38
rs765216114 Chr.18: 49791647-49791647 on Build GRCh38
rs374903164 Chr.18: 49791478-49791478 on Build GRCh38
rs367948616 Chr.18: 49791615-49791615 on Build GRCh38
rs142461980 Chr.18: 49791388-49791388 on Build GRCh38
rs140434637 Chr.18: 49791598-49791598 on Build GRCh38
rs558610040 Chr.18: 49791705-49791705 on Build GRCh38
rs372156067 Chr.18: 49791642-49791642 on Build GRCh38
rs745745330 Chr.18: 49791440-49791440 on Build GRCh38
rs771149248 Chr.18: 49791838-49791838 on Build GRCh38
rs753950522 Chr.18: 49791483-49791483 on Build GRCh38
rs998210620 Chr.18: 49791452-49791452 on Build GRCh38
rs887098263 Chr.18: 49791814-49791814 on Build GRCh38
rs572199184 Chr.18: 49791576-49791576 on Build GRCh38
rs906523483 Chr.18: 49791631-49791631 on Build GRCh38
rs561205872 Chr.18: 49791858-49791858 on Build GRCh38
rs761460625 Chr.18: 49791482-49791483 on Build GRCh38
rs762849309 Chr.18: 49791465-49791465 on Build GRCh38
rs765547364 Chr.18: 49791524-49791524 on Build GRCh38
rs200198861 Chr.18: 49791460-49791460 on Build GRCh38
rs377532103 Chr.18: 49791596-49791596 on Build GRCh38
rs190780040 Chr.18: 49791532-49791532 on Build GRCh38
rs766307965 Chr.18: 49791618-49791618 on Build GRCh38
rs529556 Chr.18: 49791397-49791397 on Build GRCh38

View Full Product Details