Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 51048678-51049186 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMAD4
Entrez Gene ID
4089
Gene Name
SMAD family member 4
Gene Aliases
DPC4, JIP, MADH4, MYHRS
UniGene
Hs.75862
Gene Chromosome Location
Chr.18: 51030213-51085042 on Build GRCh38
Transcripts
NM_005359.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6957607 c.308A>C p.D103A Chr.18: 51048744-51048744 on Build GRCh38
14104 c.1_*del p.0 Chr.18: 51047047-51078467 on Build GRCh38
4382624 c.290G>A p.R97H Chr.18: 51048726-51048726 on Build GRCh38
7430437 c.402A>T p.E134D Chr.18: 51048838-51048838 on Build GRCh38
5521010 c.275A>T p.H92L Chr.18: 51048711-51048711 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779069779 Chr.18: 51048835-51048835 on Build GRCh38
rs140926102 Chr.18: 51048805-51048805 on Build GRCh38
rs755862230 Chr.18: 51048826-51048826 on Build GRCh38
rs187523362 Chr.18: 51049131-51049131 on Build GRCh38
rs765550126 Chr.18: 51048860-51048861 on Build GRCh38
rs777803301 Chr.18: 51048862-51048862 on Build GRCh38
rs1028514415 Chr.18: 51048961-51048961 on Build GRCh38
rs189904125 Chr.18: 51048912-51048912 on Build GRCh38
rs771808272 Chr.18: 51048810-51048810 on Build GRCh38
rs377767324 Chr.18: 51048809-51048810 on Build GRCh38
rs754123998 Chr.18: 51048777-51048778 on Build GRCh38
rs2276163 Chr.18: 51049019-51049019 on Build GRCh38
rs200772603 Chr.18: 51048865-51048865 on Build GRCh38
rs1057070464 Chr.18: 51049048-51049048 on Build GRCh38
rs377767326 Chr.18: 51048839-51048839 on Build GRCh38
rs931112795 Chr.18: 51048923-51048923 on Build GRCh38
rs777019571 Chr.18: 51048870-51048870 on Build GRCh38
rs745565217 Chr.18: 51049067-51049067 on Build GRCh38
rs727504151 Chr.18: 51048762-51048762 on Build GRCh38
rs749922210 Chr.18: 51048871-51048872 on Build GRCh38
rs1028902012 Chr.18: 51049156-51049156 on Build GRCh38
rs751154230 Chr.18: 51048734-51048734 on Build GRCh38
rs1044255893 Chr.18: 51048938-51048938 on Build GRCh38
rs770303876 Chr.18: 51048898-51048898 on Build GRCh38
rs750172880 Chr.18: 51048807-51048807 on Build GRCh38
rs377767386 Chr.18: 51048861-51048861 on Build GRCh38
rs987851417 Chr.18: 51049116-51049116 on Build GRCh38
rs757211048 Chr.18: 51048778-51048778 on Build GRCh38
rs796782051 Chr.18: 51049000-51049000 on Build GRCh38
rs1042426277 Chr.18: 51049120-51049120 on Build GRCh38
rs896463150 Chr.18: 51049054-51049054 on Build GRCh38
rs771456293 Chr.18: 51048866-51048866 on Build GRCh38
rs755300827 Chr.18: 51048856-51048857 on Build GRCh38
rs748395067 Chr.18: 51048836-51048836 on Build GRCh38
rs377767325 Chr.18: 51048817-51048818 on Build GRCh38
rs1004249434 Chr.18: 51048879-51048879 on Build GRCh38
rs778319626 Chr.18: 51048864-51048865 on Build GRCh38
rs903949855 Chr.18: 51049122-51049122 on Build GRCh38
rs985886127 Chr.18: 51048998-51048998 on Build GRCh38
rs145988618 Chr.18: 51048790-51048790 on Build GRCh38
rs377767323 Chr.18: 51048738-51048738 on Build GRCh38
rs876660079 Chr.18: 51048733-51048733 on Build GRCh38
rs80002804 Chr.18: 51048962-51048962 on Build GRCh38
rs758809290 Chr.18: 51048862-51048863 on Build GRCh38
rs202126703 Chr.18: 51048730-51048730 on Build GRCh38
rs746178972 Chr.18: 51048883-51048883 on Build GRCh38
rs1016008247 Chr.18: 51048890-51048890 on Build GRCh38
rs762501162 Chr.18: 51048712-51048712 on Build GRCh38
rs876659844 Chr.18: 51048780-51048780 on Build GRCh38
rs961095816 Chr.18: 51048716-51048716 on Build GRCh38
rs863224401 Chr.18: 51048811-51048811 on Build GRCh38
rs201296880 Chr.18: 51048847-51048847 on Build GRCh38
rs150229208 Chr.18: 51048823-51048823 on Build GRCh38
rs182981059 Chr.18: 51049025-51049025 on Build GRCh38

View Full Product Details