Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 7847627-7848117 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EVI5L
Entrez Gene ID
115704
Gene Name
ecotropic viral integration site 5 like
Gene Aliases
-
UniGene
Hs.26870
Gene Chromosome Location
Chr.19: 7830275-7864976 on Build GRCh38
Transcripts
NM_001159944.2 NM_145245.4 XM_005272458.4 XM_017026239.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6365212 c.302G>A p.R101Q Chr.19: 7847896-7847896 on Build GRCh38
5653736 c.223G>T p.A75S Chr.19: 7847817-7847817 on Build GRCh38
7249522 c.327+9C>T p.? Chr.19: 7847930-7847930 on Build GRCh38
397042 c.276G>C p.R92R Chr.19: 7847870-7847870 on Build GRCh38
5722805 c.230C>T p.S77F Chr.19: 7847824-7847824 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1000784579 Chr.19: 7848073-7848073 on Build GRCh38
rs754679538 Chr.19: 7847850-7847850 on Build GRCh38
rs546615764 Chr.19: 7847931-7847931 on Build GRCh38
rs898810024 Chr.19: 7847991-7847991 on Build GRCh38
rs779624210 Chr.19: 7847898-7847898 on Build GRCh38
rs144010004 Chr.19: 7847960-7847960 on Build GRCh38
rs199527137 Chr.19: 7847741-7847741 on Build GRCh38
rs775424127 Chr.19: 7847709-7847709 on Build GRCh38
rs1042278666 Chr.19: 7848046-7848046 on Build GRCh38
rs1007662081 Chr.19: 7847819-7847819 on Build GRCh38
rs74651651 Chr.19: 7847982-7847982 on Build GRCh38
rs185612344 Chr.19: 7847934-7847934 on Build GRCh38
rs927891889 Chr.19: 7847922-7847922 on Build GRCh38
rs756371946 Chr.19: 7847758-7847758 on Build GRCh38
rs1010989603 Chr.19: 7847672-7847672 on Build GRCh38
rs868044398 Chr.19: 7847802-7847802 on Build GRCh38
rs753092741 Chr.19: 7847745-7847745 on Build GRCh38
rs917097907 Chr.19: 7847923-7847923 on Build GRCh38
rs771840893 Chr.19: 7847777-7847777 on Build GRCh38
rs1026048912 Chr.19: 7848003-7848003 on Build GRCh38
rs938036659 Chr.19: 7847651-7847651 on Build GRCh38
rs1013959667 Chr.19: 7847654-7847654 on Build GRCh38
rs373550043 Chr.19: 7847774-7847774 on Build GRCh38
rs749811652 Chr.19: 7847687-7847687 on Build GRCh38
rs375712197 Chr.19: 7847776-7847776 on Build GRCh38
rs778537610 Chr.19: 7847773-7847773 on Build GRCh38
rs545076372 Chr.19: 7848076-7848076 on Build GRCh38
rs200105663 Chr.19: 7847953-7847953 on Build GRCh38
rs767451220 Chr.19: 7847957-7847957 on Build GRCh38
rs902438611 Chr.19: 7848068-7848068 on Build GRCh38
rs893932597 Chr.19: 7847671-7847671 on Build GRCh38
rs367839369 Chr.19: 7847879-7847879 on Build GRCh38
rs1055112177 Chr.19: 7847670-7847670 on Build GRCh38
rs372069618 Chr.19: 7847924-7847924 on Build GRCh38
rs761718634 Chr.19: 7847815-7847815 on Build GRCh38
rs528561163 Chr.19: 7847703-7847703 on Build GRCh38
rs990851989 Chr.19: 7847927-7847927 on Build GRCh38
rs775658223 Chr.19: 7847959-7847959 on Build GRCh38
rs763967496 Chr.19: 7847711-7847711 on Build GRCh38
rs573951961 Chr.19: 7847989-7847989 on Build GRCh38
rs140398803 Chr.19: 7847852-7847852 on Build GRCh38
rs769705820 Chr.19: 7847938-7847938 on Build GRCh38
rs370414994 Chr.19: 7847778-7847778 on Build GRCh38
rs764314362 Chr.19: 7847891-7847891 on Build GRCh38
rs1007136576 Chr.19: 7847974-7847974 on Build GRCh38
rs748002704 Chr.19: 7847937-7847937 on Build GRCh38
rs749715262 Chr.19: 7847760-7847760 on Build GRCh38
rs908916755 Chr.19: 7847996-7847996 on Build GRCh38
rs950377809 Chr.19: 7847975-7847975 on Build GRCh38
rs761843617 Chr.19: 7847716-7847716 on Build GRCh38
rs1034851314 Chr.19: 7848031-7848031 on Build GRCh38
rs1030298860 Chr.19: 7847782-7847782 on Build GRCh38
rs559312006 Chr.19: 7848008-7848008 on Build GRCh38
rs771585408 Chr.19: 7847761-7847761 on Build GRCh38
rs2307003 Chr.19: 7847930-7847930 on Build GRCh38
rs140227216 Chr.19: 7847710-7847710 on Build GRCh38
rs950475753 Chr.19: 7848009-7848009 on Build GRCh38
rs754276273 Chr.19: 7847895-7847895 on Build GRCh38
rs573034029 Chr.19: 7848013-7848013 on Build GRCh38
rs201715914 Chr.19: 7847954-7847954 on Build GRCh38
rs982391362 Chr.19: 7848010-7848010 on Build GRCh38
rs371567316 Chr.19: 7847744-7847744 on Build GRCh38
rs917741594 Chr.19: 7848017-7848017 on Build GRCh38
rs757837977 Chr.19: 7847896-7847896 on Build GRCh38
rs146677119 Chr.19: 7847798-7847798 on Build GRCh38
rs959558567 Chr.19: 7848084-7848084 on Build GRCh38
rs768297573 Chr.19: 7847781-7847781 on Build GRCh38
rs774026921 Chr.19: 7847690-7847690 on Build GRCh38
rs981952538 Chr.19: 7847907-7847907 on Build GRCh38
rs144945165 Chr.19: 7847997-7847997 on Build GRCh38
rs193137808 Chr.19: 7847873-7847873 on Build GRCh38
rs564757821 Chr.19: 7848086-7848086 on Build GRCh38
rs779645750 Chr.19: 7847929-7847929 on Build GRCh38
rs752791877 Chr.19: 7847874-7847874 on Build GRCh38
rs770495881 Chr.19: 7847688-7847688 on Build GRCh38
rs114480200 Chr.19: 7847956-7847956 on Build GRCh38
rs949262068 Chr.19: 7848029-7848029 on Build GRCh38
rs759213228 Chr.19: 7847691-7847691 on Build GRCh38
rs753652084 Chr.19: 7847715-7847715 on Build GRCh38
rs766398564 Chr.19: 7847721-7847721 on Build GRCh38

View Full Product Details