Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 7849875-7850367 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EVI5L
Entrez Gene ID
115704
Gene Name
ecotropic viral integration site 5 like
Gene Aliases
-
UniGene
Hs.26870
Gene Chromosome Location
Chr.19: 7830275-7864976 on Build GRCh38
Transcripts
NM_001159944.2 NM_145245.4 XM_005272458.4 XM_017026239.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7129040 c.715C>A p.L239I Chr.19: 7850084-7850084 on Build GRCh38
5524915 c.673G>A p.E225K Chr.19: 7850042-7850042 on Build GRCh38
5793614 c.692T>A p.L231H Chr.19: 7850061-7850061 on Build GRCh38
6292415 c.750G>A p.L250L Chr.19: 7850119-7850119 on Build GRCh38
5857120 c.652G>T p.V218L Chr.19: 7850021-7850021 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747444296 Chr.19: 7850029-7850029 on Build GRCh38
rs971826361 Chr.19: 7850249-7850249 on Build GRCh38
rs1057128082 Chr.19: 7850182-7850182 on Build GRCh38
rs762669875 Chr.19: 7850081-7850081 on Build GRCh38
rs375512695 Chr.19: 7850150-7850150 on Build GRCh38
rs149368767 Chr.19: 7850283-7850283 on Build GRCh38
rs28420566 Chr.19: 7849992-7849992 on Build GRCh38
rs1032185736 Chr.19: 7850282-7850282 on Build GRCh38
rs751796861 Chr.19: 7850143-7850143 on Build GRCh38
rs912325406 Chr.19: 7850012-7850012 on Build GRCh38
rs904667342 Chr.19: 7850242-7850242 on Build GRCh38
rs1004064138 Chr.19: 7850180-7850180 on Build GRCh38
rs138727675 Chr.19: 7850187-7850187 on Build GRCh38
rs1023423764 Chr.19: 7849956-7849956 on Build GRCh38
rs759460493 Chr.19: 7850099-7850099 on Build GRCh38
rs200789450 Chr.19: 7850134-7850134 on Build GRCh38
rs750596733 Chr.19: 7850023-7850023 on Build GRCh38
rs761290465 Chr.19: 7849976-7849976 on Build GRCh38
rs116138693 Chr.19: 7850211-7850211 on Build GRCh38
rs751446273 Chr.19: 7849988-7849988 on Build GRCh38
rs372190141 Chr.19: 7850080-7850080 on Build GRCh38
rs574129042 Chr.19: 7849900-7849900 on Build GRCh38
rs187344289 Chr.19: 7850039-7850039 on Build GRCh38
rs556143521 Chr.19: 7850296-7850296 on Build GRCh38
rs758668920 Chr.19: 7850024-7850024 on Build GRCh38
rs757495280 Chr.19: 7849955-7849955 on Build GRCh38
rs143142461 Chr.19: 7850086-7850086 on Build GRCh38
rs781641526 Chr.19: 7850167-7850167 on Build GRCh38
rs548557570 Chr.19: 7850159-7850159 on Build GRCh38
rs924785000 Chr.19: 7849977-7849977 on Build GRCh38
rs779059964 Chr.19: 7849957-7849957 on Build GRCh38
rs752538532 Chr.19: 7850111-7850111 on Build GRCh38
rs532863835 Chr.19: 7849990-7849990 on Build GRCh38
rs753938272 Chr.19: 7849991-7849991 on Build GRCh38
rs780800302 Chr.19: 7850143-7850158 on Build GRCh38
rs1013129536 Chr.19: 7849921-7849921 on Build GRCh38
rs1016881673 Chr.19: 7849915-7849915 on Build GRCh38
rs555214968 Chr.19: 7849939-7849939 on Build GRCh38
rs892030687 Chr.19: 7850333-7850333 on Build GRCh38
rs748600497 Chr.19: 7850135-7850150 on Build GRCh38
rs759513871 Chr.19: 7849989-7849989 on Build GRCh38
rs375165078 Chr.19: 7849986-7849986 on Build GRCh38
rs990281405 Chr.19: 7850295-7850295 on Build GRCh38
rs981525421 Chr.19: 7850250-7850250 on Build GRCh38
rs769143750 Chr.19: 7850058-7850058 on Build GRCh38
rs143832251 Chr.19: 7849985-7849985 on Build GRCh38
rs931235647 Chr.19: 7850100-7850100 on Build GRCh38
rs781496751 Chr.19: 7850049-7850049 on Build GRCh38
rs377173331 Chr.19: 7850135-7850135 on Build GRCh38
rs760686773 Chr.19: 7850119-7850119 on Build GRCh38
rs568606721 Chr.19: 7850183-7850183 on Build GRCh38
rs886924027 Chr.19: 7850174-7850174 on Build GRCh38
rs764882517 Chr.19: 7849979-7849979 on Build GRCh38
rs577083649 Chr.19: 7850234-7850234 on Build GRCh38
rs774283177 Chr.19: 7850089-7850089 on Build GRCh38
rs776204020 Chr.19: 7849975-7849975 on Build GRCh38
rs779303413 Chr.19: 7849994-7849994 on Build GRCh38
rs564664184 Chr.19: 7849970-7849971 on Build GRCh38
rs367946414 Chr.19: 7849946-7849946 on Build GRCh38
rs780317405 Chr.19: 7850138-7850138 on Build GRCh38
rs962676419 Chr.19: 7849933-7849933 on Build GRCh38
rs545130237 Chr.19: 7849942-7849942 on Build GRCh38
rs111420117 Chr.19: 7850169-7850169 on Build GRCh38
rs376359329 Chr.19: 7850275-7850275 on Build GRCh38
rs780366498 Chr.19: 7850027-7850027 on Build GRCh38
rs781123718 Chr.19: 7850332-7850332 on Build GRCh38
rs527377964 Chr.19: 7849970-7849970 on Build GRCh38
rs895881100 Chr.19: 7849912-7849912 on Build GRCh38
rs751471380 Chr.19: 7850191-7850191 on Build GRCh38
rs368268597 Chr.19: 7849960-7849960 on Build GRCh38
rs765209807 Chr.19: 7850130-7850130 on Build GRCh38
rs748697117 Chr.19: 7850054-7850054 on Build GRCh38
rs747492093 Chr.19: 7849972-7849972 on Build GRCh38
rs138346986 Chr.19: 7850107-7850107 on Build GRCh38
rs756859436 Chr.19: 7850134-7850135 on Build GRCh38
rs561494343 Chr.19: 7849910-7849910 on Build GRCh38
rs1023130481 Chr.19: 7849959-7849959 on Build GRCh38
rs745348232 Chr.19: 7850156-7850157 on Build GRCh38
rs769196716 Chr.19: 7850048-7850048 on Build GRCh38

View Full Product Details