Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 8075044-8075471 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBN3
Entrez Gene ID
84467
Gene Name
fibrillin 3
Gene Aliases
-
UniGene
Hs.370362
Gene Chromosome Location
Chr.19: 8065402-8149846 on Build GRCh38
Transcripts
NM_001321431.1 NM_032447.4 XM_017027372.1 XM_017027373.1 XM_017027374.1 XM_017027375.1 XM_017027376.1 XM_017027377.1 XM_017027378.1 XM_017027379.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7167066 c.7583-1G>C p.? Chr.19: 8075191-8075191 on Build GRCh38
266525 c.7569C>T p.G2523G Chr.19: 8075296-8075296 on Build GRCh38
3681114 c.7517C>T p.P2506L Chr.19: 8075348-8075348 on Build GRCh38
3693232 c.7650C>T p.R2550R Chr.19: 8075123-8075123 on Build GRCh38
1397710 c.7529G>A p.R2510H Chr.19: 8075336-8075336 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749575255 Chr.19: 8075368-8075368 on Build GRCh38
rs753699910 Chr.19: 8075257-8075257 on Build GRCh38
rs148480582 Chr.19: 8075350-8075350 on Build GRCh38
rs761619910 Chr.19: 8075429-8075429 on Build GRCh38
rs757071505 Chr.19: 8075258-8075258 on Build GRCh38
rs766891455 Chr.19: 8075238-8075238 on Build GRCh38
rs758216059 Chr.19: 8075447-8075447 on Build GRCh38
rs777679709 Chr.19: 8075168-8075168 on Build GRCh38
rs778921959 Chr.19: 8075347-8075347 on Build GRCh38
rs778056837 Chr.19: 8075367-8075367 on Build GRCh38
rs760808197 Chr.19: 8075112-8075112 on Build GRCh38
rs748278526 Chr.19: 8075359-8075359 on Build GRCh38
rs749013131 Chr.19: 8075171-8075171 on Build GRCh38
rs771469177 Chr.19: 8075373-8075373 on Build GRCh38
rs372140427 Chr.19: 8075222-8075222 on Build GRCh38
rs759004864 Chr.19: 8075133-8075133 on Build GRCh38
rs774393336 Chr.19: 8075199-8075199 on Build GRCh38
rs139865578 Chr.19: 8075302-8075302 on Build GRCh38
rs749313245 Chr.19: 8075096-8075096 on Build GRCh38
rs367900131 Chr.19: 8075173-8075173 on Build GRCh38
rs200811952 Chr.19: 8075247-8075247 on Build GRCh38
rs770082761 Chr.19: 8075281-8075281 on Build GRCh38
rs142718485 Chr.19: 8075372-8075372 on Build GRCh38
rs866464002 Chr.19: 8075298-8075298 on Build GRCh38
rs548835102 Chr.19: 8075243-8075243 on Build GRCh38
rs757122800 Chr.19: 8075338-8075338 on Build GRCh38
rs774745783 Chr.19: 8075371-8075371 on Build GRCh38
rs748156927 Chr.19: 8075279-8075279 on Build GRCh38
rs112479210 Chr.19: 8075227-8075227 on Build GRCh38
rs897690137 Chr.19: 8075441-8075441 on Build GRCh38
rs200311384 Chr.19: 8075163-8075163 on Build GRCh38
rs540174603 Chr.19: 8075336-8075336 on Build GRCh38
rs747955852 Chr.19: 8075091-8075091 on Build GRCh38
rs373991096 Chr.19: 8075203-8075203 on Build GRCh38
rs750752355 Chr.19: 8075131-8075131 on Build GRCh38
rs35477781 Chr.19: 8075153-8075153 on Build GRCh38
rs922967358 Chr.19: 8075205-8075205 on Build GRCh38
rs780832638 Chr.19: 8075154-8075154 on Build GRCh38
rs376017916 Chr.19: 8075164-8075164 on Build GRCh38
rs139725223 Chr.19: 8075111-8075111 on Build GRCh38
rs779827993 Chr.19: 8075352-8075352 on Build GRCh38
rs528149973 Chr.19: 8075266-8075266 on Build GRCh38
rs765575981 Chr.19: 8075125-8075125 on Build GRCh38
rs142975316 Chr.19: 8075124-8075124 on Build GRCh38
rs776946199 Chr.19: 8075114-8075114 on Build GRCh38
rs375723933 Chr.19: 8075337-8075337 on Build GRCh38
rs764444197 Chr.19: 8075113-8075113 on Build GRCh38
rs201413560 Chr.19: 8075264-8075264 on Build GRCh38
rs755837186 Chr.19: 8075248-8075248 on Build GRCh38
rs977083740 Chr.19: 8075194-8075194 on Build GRCh38
rs767987720 Chr.19: 8075317-8075317 on Build GRCh38
rs145316149 Chr.19: 8075348-8075348 on Build GRCh38
rs753825866 Chr.19: 8075297-8075297 on Build GRCh38
rs376718909 Chr.19: 8075361-8075362 on Build GRCh38
rs1050797029 Chr.19: 8075218-8075218 on Build GRCh38
rs200805247 Chr.19: 8075272-8075272 on Build GRCh38
rs769702519 Chr.19: 8075362-8075362 on Build GRCh38
rs774692537 Chr.19: 8075308-8075308 on Build GRCh38
rs766838172 Chr.19: 8075141-8075141 on Build GRCh38
rs191067028 Chr.19: 8075392-8075392 on Build GRCh38
rs1029020725 Chr.19: 8075259-8075259 on Build GRCh38
rs777258686 Chr.19: 8075249-8075249 on Build GRCh38
rs769255273 Chr.19: 8075294-8075303 on Build GRCh38
rs760294678 Chr.19: 8075080-8075080 on Build GRCh38
rs773678898 Chr.19: 8075228-8075228 on Build GRCh38
rs750286906 Chr.19: 8075438-8075438 on Build GRCh38
rs764963517 Chr.19: 8075433-8075433 on Build GRCh38
rs202000856 Chr.19: 8075223-8075223 on Build GRCh38
rs754484605 Chr.19: 8075245-8075245 on Build GRCh38
rs571052941 Chr.19: 8075326-8075326 on Build GRCh38
rs760064722 Chr.19: 8075333-8075333 on Build GRCh38
rs771128192 Chr.19: 8075370-8075370 on Build GRCh38
rs373710480 Chr.19: 8075382-8075382 on Build GRCh38
rs929372006 Chr.19: 8075448-8075448 on Build GRCh38
rs945897981 Chr.19: 8075443-8075443 on Build GRCh38
rs61729621 Chr.19: 8075093-8075093 on Build GRCh38
rs147665635 Chr.19: 8075393-8075393 on Build GRCh38
rs993575178 Chr.19: 8075253-8075253 on Build GRCh38
rs763183824 Chr.19: 8075232-8075232 on Build GRCh38
rs201291150 Chr.19: 8075363-8075363 on Build GRCh38
rs933211608 Chr.19: 8075207-8075207 on Build GRCh38
rs780779632 Chr.19: 8075086-8075086 on Build GRCh38
rs138062309 Chr.19: 8075146-8075146 on Build GRCh38
rs143120483 Chr.19: 8075303-8075303 on Build GRCh38
rs199739474 Chr.19: 8075074-8075074 on Build GRCh38
rs760247633 Chr.19: 8075384-8075384 on Build GRCh38
rs150872554 Chr.19: 8075099-8075099 on Build GRCh38
rs760093631 Chr.19: 8075312-8075312 on Build GRCh38
rs768799159 Chr.19: 8075215-8075215 on Build GRCh38
rs142379709 Chr.19: 8075428-8075428 on Build GRCh38

View Full Product Details