Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35154536-35155042 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FXYD5
Entrez Gene ID
53827
Gene Name
FXYD domain containing ion transport regulator 5
Gene Aliases
DYSAD, HSPC113, IWU1, KCT1, OIT2, PRO6241, RIC
UniGene
Hs.333418
Gene Chromosome Location
Chr.19: 35154722-35169885 on Build GRCh38
Transcripts
NM_001164605.1 NM_001320912.1 NM_001320913.1 NM_014164.5 NM_144779.2 NR_028406.1

SNP Detail

SNP ID Chromosome Location
rs937983978 Chr.19: 35154915-35154915 on Build GRCh38
rs969179111 Chr.19: 35154887-35154887 on Build GRCh38
rs917772185 Chr.19: 35154964-35154964 on Build GRCh38
rs576708519 Chr.19: 35154711-35154711 on Build GRCh38
rs545660948 Chr.19: 35154844-35154844 on Build GRCh38
rs532472509 Chr.19: 35154840-35154840 on Build GRCh38
rs984585159 Chr.19: 35154788-35154788 on Build GRCh38
rs898224661 Chr.19: 35154778-35154778 on Build GRCh38
rs976045542 Chr.19: 35154749-35154749 on Build GRCh38
rs910007485 Chr.19: 35154809-35154809 on Build GRCh38
rs892649447 Chr.19: 35154612-35154612 on Build GRCh38
rs914605220 Chr.19: 35154697-35154697 on Build GRCh38
rs192821563 Chr.19: 35154646-35154646 on Build GRCh38
rs1056304097 Chr.19: 35154733-35154733 on Build GRCh38
rs779960061 Chr.19: 35154979-35154979 on Build GRCh38
rs995194290 Chr.19: 35154786-35154786 on Build GRCh38
rs901523700 Chr.19: 35154982-35154982 on Build GRCh38
rs939601072 Chr.19: 35154586-35154586 on Build GRCh38
rs886139 Chr.19: 35154566-35154566 on Build GRCh38
rs898257479 Chr.19: 35154978-35154978 on Build GRCh38
rs947352155 Chr.19: 35154700-35154700 on Build GRCh38
rs941447156 Chr.19: 35154813-35154813 on Build GRCh38
rs1036720286 Chr.19: 35154762-35154762 on Build GRCh38
rs540386443 Chr.19: 35154757-35154757 on Build GRCh38
rs541459666 Chr.19: 35154676-35154676 on Build GRCh38
rs73041999 Chr.19: 35154910-35154910 on Build GRCh38
rs903602044 Chr.19: 35154707-35154707 on Build GRCh38
rs997151855 Chr.19: 35155006-35155006 on Build GRCh38
rs750176740 Chr.19: 35154878-35154878 on Build GRCh38
rs552615838 Chr.19: 35154859-35154859 on Build GRCh38
rs569283784 Chr.19: 35154904-35154904 on Build GRCh38
rs950611958 Chr.19: 35155011-35155011 on Build GRCh38
rs1008892343 Chr.19: 35154835-35154835 on Build GRCh38
rs1007674834 Chr.19: 35154710-35154710 on Build GRCh38
rs966382139 Chr.19: 35154734-35154734 on Build GRCh38
rs1026239564 Chr.19: 35155008-35155008 on Build GRCh38
rs1056296381 Chr.19: 35154943-35154943 on Build GRCh38
rs1633925 Chr.19: 35155012-35155012 on Build GRCh38
rs112352066 Chr.19: 35154914-35154914 on Build GRCh38
rs548648170 Chr.19: 35154977-35154977 on Build GRCh38
rs1003945546 Chr.19: 35154760-35154760 on Build GRCh38
rs1016085058 Chr.19: 35155015-35155015 on Build GRCh38
rs1028373146 Chr.19: 35154820-35154820 on Build GRCh38
rs912694735 Chr.19: 35154665-35154665 on Build GRCh38
rs988919393 Chr.19: 35154863-35154863 on Build GRCh38
rs1043863039 Chr.19: 35154913-35154913 on Build GRCh38
rs1009658573 Chr.19: 35154635-35154635 on Build GRCh38
rs2451992 Chr.19: 35154824-35154824 on Build GRCh38
rs954411016 Chr.19: 35154573-35154573 on Build GRCh38
rs914678586 Chr.19: 35154879-35154879 on Build GRCh38
rs896322697 Chr.19: 35154739-35154739 on Build GRCh38
rs889441289 Chr.19: 35155010-35155010 on Build GRCh38
rs956180451 Chr.19: 35154861-35154861 on Build GRCh38
rs530329123 Chr.19: 35154721-35154721 on Build GRCh38
rs905124107 Chr.19: 35154715-35154715 on Build GRCh38
rs981204059 Chr.19: 35154847-35154847 on Build GRCh38
rs62111371 Chr.19: 35154974-35154974 on Build GRCh38
rs1028590345 Chr.19: 35154571-35154571 on Build GRCh38
rs1049443804 Chr.19: 35155004-35155004 on Build GRCh38
rs934770687 Chr.19: 35154671-35154671 on Build GRCh38
rs1045070212 Chr.19: 35154563-35154563 on Build GRCh38
rs879655375 Chr.19: 35154612-35154612 on Build GRCh38

View Full Product Details