Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35758016-35758558 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PROSER3
Entrez Gene ID
148137
Gene Name
proline and serine rich 3
Gene Aliases
C19orf55
UniGene
Hs.527982
Gene Chromosome Location
Chr.19: 35758139-35770115 on Build GRCh38
Transcripts
NM_001039887.2 XM_011526528.1 XM_011526529.2 XM_011526530.1 XM_011526531.1 XM_011526532.1 XM_011526533.2 XM_011526534.1 XM_011526535.2 XM_011526536.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4140536 c.2T>C p.M1T Chr.19: 35758217-35758217 on Build GRCh38
4140537 c.2T>C p.M1T Chr.19: 35758217-35758217 on Build GRCh38
4140535 c.2T>C p.M1T Chr.19: 35758217-35758217 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs776847103 Chr.19: 35758198-35758198 on Build GRCh38
rs188604352 Chr.19: 35758419-35758419 on Build GRCh38
rs991617876 Chr.19: 35758391-35758391 on Build GRCh38
rs376726005 Chr.19: 35758516-35758516 on Build GRCh38
rs987420037 Chr.19: 35758325-35758325 on Build GRCh38
rs751301719 Chr.19: 35758249-35758249 on Build GRCh38
rs531096426 Chr.19: 35758160-35758160 on Build GRCh38
rs986191970 Chr.19: 35758094-35758094 on Build GRCh38
rs569704049 Chr.19: 35758426-35758426 on Build GRCh38
rs900251897 Chr.19: 35758529-35758529 on Build GRCh38
rs1042676098 Chr.19: 35758487-35758487 on Build GRCh38
rs574894145 Chr.19: 35758096-35758096 on Build GRCh38
rs373439591 Chr.19: 35758257-35758257 on Build GRCh38
rs1000726103 Chr.19: 35758288-35758288 on Build GRCh38
rs559809348 Chr.19: 35758093-35758093 on Build GRCh38
rs960643503 Chr.19: 35758088-35758088 on Build GRCh38
rs962173850 Chr.19: 35758063-35758063 on Build GRCh38
rs548356020 Chr.19: 35758357-35758357 on Build GRCh38
rs781546891 Chr.19: 35758402-35758402 on Build GRCh38
rs376371218 Chr.19: 35758277-35758277 on Build GRCh38
rs749278407 Chr.19: 35758173-35758173 on Build GRCh38
rs1027775440 Chr.19: 35758081-35758081 on Build GRCh38
rs1021247494 Chr.19: 35758100-35758100 on Build GRCh38
rs765385863 Chr.19: 35758206-35758206 on Build GRCh38
rs771870086 Chr.19: 35758276-35758276 on Build GRCh38
rs374829741 Chr.19: 35758238-35758238 on Build GRCh38
rs368833431 Chr.19: 35758255-35758255 on Build GRCh38
rs576588823 Chr.19: 35758490-35758490 on Build GRCh38
rs557322715 Chr.19: 35758303-35758304 on Build GRCh38
rs908811661 Chr.19: 35758359-35758359 on Build GRCh38
rs940206213 Chr.19: 35758188-35758188 on Build GRCh38
rs1020788793 Chr.19: 35758311-35758311 on Build GRCh38
rs889318169 Chr.19: 35758512-35758512 on Build GRCh38
rs898081011 Chr.19: 35758240-35758240 on Build GRCh38
rs979237720 Chr.19: 35758133-35758133 on Build GRCh38
rs552482889 Chr.19: 35758183-35758183 on Build GRCh38
rs774181765 Chr.19: 35758191-35758191 on Build GRCh38
rs757477530 Chr.19: 35758273-35758273 on Build GRCh38
rs1027854692 Chr.19: 35758268-35758268 on Build GRCh38
rs531996734 Chr.19: 35758235-35758235 on Build GRCh38
rs926518453 Chr.19: 35758381-35758381 on Build GRCh38
rs979669572 Chr.19: 35758318-35758318 on Build GRCh38
rs921496423 Chr.19: 35758157-35758157 on Build GRCh38
rs763561639 Chr.19: 35758209-35758209 on Build GRCh38
rs1032173789 Chr.19: 35758527-35758527 on Build GRCh38
rs778818381 Chr.19: 35758256-35758256 on Build GRCh38
rs946530612 Chr.19: 35758112-35758112 on Build GRCh38
rs1019013134 Chr.19: 35758282-35758282 on Build GRCh38
rs938117518 Chr.19: 35758409-35758409 on Build GRCh38
rs964452405 Chr.19: 35758301-35758301 on Build GRCh38
rs781640812 Chr.19: 35758232-35758232 on Build GRCh38
rs779655719 Chr.19: 35758239-35758239 on Build GRCh38
rs1056200228 Chr.19: 35758197-35758197 on Build GRCh38
rs769123709 Chr.19: 35758445-35758445 on Build GRCh38
rs3746275 Chr.19: 35758296-35758296 on Build GRCh38
rs752035115 Chr.19: 35758225-35758225 on Build GRCh38
rs898069600 Chr.19: 35758428-35758428 on Build GRCh38
rs934273603 Chr.19: 35758164-35758164 on Build GRCh38
rs966478601 Chr.19: 35758102-35758102 on Build GRCh38
rs964325094 Chr.19: 35758113-35758113 on Build GRCh38
rs577273493 Chr.19: 35758470-35758470 on Build GRCh38
rs1018673212 Chr.19: 35758526-35758526 on Build GRCh38
rs372100420 Chr.19: 35758210-35758210 on Build GRCh38
rs1032024781 Chr.19: 35758292-35758292 on Build GRCh38
rs974394768 Chr.19: 35758143-35758143 on Build GRCh38
rs564514714 Chr.19: 35758233-35758233 on Build GRCh38
rs940154908 Chr.19: 35758370-35758370 on Build GRCh38
rs752683575 Chr.19: 35758207-35758207 on Build GRCh38
rs896444493 Chr.19: 35758450-35758450 on Build GRCh38
rs937405259 Chr.19: 35758388-35758388 on Build GRCh38
rs550909936 Chr.19: 35758234-35758234 on Build GRCh38
rs192896473 Chr.19: 35758109-35758109 on Build GRCh38
rs58561864 Chr.19: 35758074-35758074 on Build GRCh38
rs901604623 Chr.19: 35758531-35758531 on Build GRCh38
rs913834608 Chr.19: 35758111-35758111 on Build GRCh38
rs779383153 Chr.19: 35758169-35758169 on Build GRCh38
rs950153127 Chr.19: 35758389-35758389 on Build GRCh38
rs577337169 Chr.19: 35758040-35758040 on Build GRCh38
rs372138291 Chr.19: 35758180-35758180 on Build GRCh38
rs569899254 Chr.19: 35758373-35758373 on Build GRCh38
rs536008462 Chr.19: 35758312-35758312 on Build GRCh38
rs757697249 Chr.19: 35758231-35758231 on Build GRCh38
rs965653643 Chr.19: 35758290-35758290 on Build GRCh38
rs566283212 Chr.19: 35758193-35758193 on Build GRCh38
rs974265984 Chr.19: 35758372-35758372 on Build GRCh38
rs759622783 Chr.19: 35758203-35758203 on Build GRCh38
rs542410516 Chr.19: 35758099-35758099 on Build GRCh38
rs563772067 Chr.19: 35758104-35758104 on Build GRCh38
rs917354512 Chr.19: 35758408-35758408 on Build GRCh38

View Full Product Details