Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35765087-35765621 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PROSER3
Entrez Gene ID
148137
Gene Name
proline and serine rich 3
Gene Aliases
C19orf55
UniGene
Hs.527982
Gene Chromosome Location
Chr.19: 35758139-35770115 on Build GRCh38
Transcripts
NM_001039887.2 XM_011526528.1 XM_011526529.2 XM_011526530.1 XM_011526531.1 XM_011526532.1 XM_011526533.2 XM_011526534.1 XM_011526535.2 XM_011526536.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6614930 c.720G>A p.S240S Chr.19: 35765127-35765127 on Build GRCh38
5627643 c.644C>A p.S215Y Chr.19: 35765087-35765087 on Build GRCh38
4911879 c.695T>C p.I232T Chr.19: 35765138-35765138 on Build GRCh38
6614931 c.684G>A p.S228S Chr.19: 35765127-35765127 on Build GRCh38
3796978 c.685G>A p.E229K Chr.19: 35765128-35765128 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs541082568 Chr.19: 35765424-35765424 on Build GRCh38
rs936522802 Chr.19: 35765357-35765357 on Build GRCh38
rs575516357 Chr.19: 35765329-35765329 on Build GRCh38
rs574650122 Chr.19: 35765252-35765252 on Build GRCh38
rs1034539135 Chr.19: 35765172-35765172 on Build GRCh38
rs897297439 Chr.19: 35765426-35765426 on Build GRCh38
rs751240968 Chr.19: 35765204-35765204 on Build GRCh38
rs913326033 Chr.19: 35765335-35765335 on Build GRCh38
rs183176244 Chr.19: 35765271-35765271 on Build GRCh38
rs370565442 Chr.19: 35765128-35765128 on Build GRCh38
rs1024890971 Chr.19: 35765495-35765495 on Build GRCh38
rs946040679 Chr.19: 35765339-35765339 on Build GRCh38
rs762003029 Chr.19: 35765147-35765147 on Build GRCh38
rs976824656 Chr.19: 35765334-35765334 on Build GRCh38
rs1036085978 Chr.19: 35765391-35765391 on Build GRCh38
rs186760248 Chr.19: 35765407-35765407 on Build GRCh38
rs931685757 Chr.19: 35765571-35765571 on Build GRCh38
rs1051517911 Chr.19: 35765580-35765582 on Build GRCh38
rs911531513 Chr.19: 35765582-35765585 on Build GRCh38
rs1027318519 Chr.19: 35765427-35765427 on Build GRCh38
rs940357134 Chr.19: 35765594-35765598 on Build GRCh38
rs749820830 Chr.19: 35765133-35765133 on Build GRCh38
rs192352490 Chr.19: 35765176-35765176 on Build GRCh38
rs1007398117 Chr.19: 35765447-35765447 on Build GRCh38
rs950215085 Chr.19: 35765540-35765540 on Build GRCh38
rs868550892 Chr.19: 35765237-35765237 on Build GRCh38
rs996101411 Chr.19: 35765433-35765433 on Build GRCh38
rs138303132 Chr.19: 35765537-35765537 on Build GRCh38
rs559047631 Chr.19: 35765383-35765383 on Build GRCh38
rs377661666 Chr.19: 35765536-35765536 on Build GRCh38
rs548393423 Chr.19: 35765558-35765558 on Build GRCh38
rs765213243 Chr.19: 35765177-35765177 on Build GRCh38
rs900144714 Chr.19: 35765143-35765143 on Build GRCh38
rs62112161 Chr.19: 35765256-35765256 on Build GRCh38
rs746948640 Chr.19: 35765421-35765421 on Build GRCh38
rs1018821443 Chr.19: 35765451-35765451 on Build GRCh38
rs977957540 Chr.19: 35765524-35765524 on Build GRCh38
rs773513556 Chr.19: 35765150-35765150 on Build GRCh38
rs368325986 Chr.19: 35765127-35765127 on Build GRCh38
rs890129247 Chr.19: 35765422-35765422 on Build GRCh38
rs201009511 Chr.19: 35765126-35765126 on Build GRCh38
rs770939502 Chr.19: 35765476-35765476 on Build GRCh38
rs759474783 Chr.19: 35765528-35765528 on Build GRCh38
rs1014838270 Chr.19: 35765390-35765390 on Build GRCh38
rs967890323 Chr.19: 35765482-35765482 on Build GRCh38
rs754241472 Chr.19: 35765150-35765150 on Build GRCh38
rs756483441 Chr.19: 35765205-35765205 on Build GRCh38
rs777246418 Chr.19: 35765182-35765182 on Build GRCh38
rs776576651 Chr.19: 35765512-35765512 on Build GRCh38
rs200193402 Chr.19: 35765108-35765108 on Build GRCh38
rs966279871 Chr.19: 35765331-35765331 on Build GRCh38
rs965882627 Chr.19: 35765457-35765457 on Build GRCh38
rs757961573 Chr.19: 35765179-35765179 on Build GRCh38
rs532029960 Chr.19: 35765423-35765423 on Build GRCh38
rs747821810 Chr.19: 35765322-35765322 on Build GRCh38
rs4806203 Chr.19: 35765340-35765340 on Build GRCh38
rs978754912 Chr.19: 35765511-35765511 on Build GRCh38
rs1042234369 Chr.19: 35765112-35765112 on Build GRCh38
rs376874928 Chr.19: 35765175-35765175 on Build GRCh38
rs895951973 Chr.19: 35765384-35765384 on Build GRCh38
rs779605436 Chr.19: 35765225-35765225 on Build GRCh38
rs926174081 Chr.19: 35765342-35765342 on Build GRCh38
rs961693799 Chr.19: 35765222-35765222 on Build GRCh38
rs974156587 Chr.19: 35765215-35765215 on Build GRCh38
rs774561163 Chr.19: 35765146-35765146 on Build GRCh38
rs921701431 Chr.19: 35765562-35765565 on Build GRCh38
rs917823218 Chr.19: 35765313-35765313 on Build GRCh38
rs988393516 Chr.19: 35765461-35765461 on Build GRCh38
rs769354991 Chr.19: 35765140-35765140 on Build GRCh38
rs766173272 Chr.19: 35765156-35765156 on Build GRCh38
rs769264885 Chr.19: 35765223-35765223 on Build GRCh38
rs371097615 Chr.19: 35765213-35765213 on Build GRCh38
rs557173124 Chr.19: 35765328-35765328 on Build GRCh38
rs777526573 Chr.19: 35765124-35765124 on Build GRCh38
rs558892596 Chr.19: 35765154-35765154 on Build GRCh38
rs367725676 Chr.19: 35765207-35765207 on Build GRCh38
rs145805491 Chr.19: 35765435-35765435 on Build GRCh38
rs937615296 Chr.19: 35765362-35765362 on Build GRCh38
rs953321567 Chr.19: 35765264-35765264 on Build GRCh38
rs931660657 Chr.19: 35765408-35765408 on Build GRCh38
rs1028568275 Chr.19: 35765170-35765170 on Build GRCh38

View Full Product Details