The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000924.3 | NP_000915.1 | 6 - 7 | 983 | 70 |
NM_001165975.2 | NP_001159447.1 | 5 - 6 | 671 | 70 | |
NM_001288768.1 | NP_001275697.1 | 6 - 7 | 1045 | 70 | |
NM_001288769.1 | NP_001275698.1 | 5 - 6 | 603 | 70 | |
NM_001315534.1 | NP_001302463.1 | 5 - 6 | 574 | 70 | |
NM_001315535.1 | NP_001302464.1 | 3 - 4 | 421 | 70 | |
XM_011538456.1 | XP_011536758.1 | 7 - 8 | 933 | 70 | |
XM_017019432.1 | XP_016874921.1 | 5 - 6 | 502 | 70 | |
XM_017019433.1 | XP_016874922.1 | 5 - 6 | 1190 | 70 | |
GenBank mRNA | AJ401609.1 | - | 5 - 6 | 530 | 70 |
AK126899.1 | - | 3 - 4 | 364 | 70 | |
AK296422.1 | - | 5 - 6 | 603 | 70 | |
AK302899.1 | - | 6 - 7 | 772 | 70 | |
AK302931.1 | - | 5 - 6 | 658 | 70 | |
BC032226.1 | - | 6 - 7 | 756 | 70 | |
BX538092.1 | - | 5 - 6 | 574 | 70 | |
U56976.1 | - | 5 - 6 | 624 | 70 | |
U86078.1 | - | 6 - 7 | 703 | 70 |